Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27070407-27070920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL5
Entrez Gene ID
60509
Gene Name
ATP/GTP binding protein like 5
Gene Aliases
CCP5, RP75
UniGene
Hs.138207
Gene Chromosome Location
Chr.2: 27051423-27070622 on Build GRCh38
Transcripts
NM_021831.5 NR_104246.1 XM_005264477.2 NM_001134693.1

Gene Symbol
OST4
Entrez Gene ID
100128731
Gene Name
oligosaccharyltransferase complex subunit 4, non-catalytic
Gene Aliases
-
UniGene
Hs.502948
Gene Chromosome Location
Chr.2: 27070472-27071699 on Build GRCh38
Transcripts
NM_021831.5 NR_104246.1 XM_005264477.2 NM_001134693.1

SNP Detail

SNP ID Chromosome Location
rs1047417783 Chr.2: 27070851-27070851 on Build GRCh38
rs905379393 Chr.2: 27070550-27070550 on Build GRCh38
rs971271076 Chr.2: 27070831-27070831 on Build GRCh38
rs910248134 Chr.2: 27070872-27070872 on Build GRCh38
rs1032898030 Chr.2: 27070475-27070475 on Build GRCh38
rs755758224 Chr.2: 27070541-27070541 on Build GRCh38
rs550879873 Chr.2: 27070438-27070438 on Build GRCh38
rs910346533 Chr.2: 27070722-27070722 on Build GRCh38
rs747761255 Chr.2: 27070551-27070551 on Build GRCh38
rs984418831 Chr.2: 27070567-27070567 on Build GRCh38
rs376938680 Chr.2: 27070612-27070612 on Build GRCh38
rs533896731 Chr.2: 27070517-27070517 on Build GRCh38
rs779584186 Chr.2: 27070543-27070543 on Build GRCh38
rs908903510 Chr.2: 27070852-27070852 on Build GRCh38
rs918975927 Chr.2: 27070546-27070546 on Build GRCh38
rs1042490260 Chr.2: 27070659-27070659 on Build GRCh38
rs991361680 Chr.2: 27070509-27070509 on Build GRCh38
rs181742210 Chr.2: 27070854-27070854 on Build GRCh38
rs958574586 Chr.2: 27070490-27070490 on Build GRCh38
rs567488248 Chr.2: 27070602-27070602 on Build GRCh38
rs753336103 Chr.2: 27070557-27070557 on Build GRCh38
rs1012813108 Chr.2: 27070787-27070787 on Build GRCh38
rs1038631246 Chr.2: 27070858-27070858 on Build GRCh38
rs1002379907 Chr.2: 27070894-27070894 on Build GRCh38
rs928957075 Chr.2: 27070510-27070510 on Build GRCh38
rs895140003 Chr.2: 27070745-27070745 on Build GRCh38
rs887352371 Chr.2: 27070539-27070539 on Build GRCh38
rs945512724 Chr.2: 27070636-27070636 on Build GRCh38
rs979661366 Chr.2: 27070446-27070446 on Build GRCh38
rs1020939475 Chr.2: 27070595-27070595 on Build GRCh38
rs1033679520 Chr.2: 27070728-27070728 on Build GRCh38
rs865817608 Chr.2: 27070593-27070593 on Build GRCh38
rs754477135 Chr.2: 27070574-27070574 on Build GRCh38
rs577867571 Chr.2: 27070839-27070839 on Build GRCh38
rs113173636 Chr.2: 27070724-27070724 on Build GRCh38
rs532815797 Chr.2: 27070483-27070483 on Build GRCh38
rs569193549 Chr.2: 27070445-27070445 on Build GRCh38
rs558145217 Chr.2: 27070885-27070885 on Build GRCh38
rs1026067540 Chr.2: 27070849-27070849 on Build GRCh38
rs930305651 Chr.2: 27070556-27070556 on Build GRCh38
rs1024620899 Chr.2: 27070796-27070796 on Build GRCh38
rs780672862 Chr.2: 27070497-27070497 on Build GRCh38
rs566486761 Chr.2: 27070516-27070516 on Build GRCh38
rs1000791296 Chr.2: 27070469-27070469 on Build GRCh38
rs1000822366 Chr.2: 27070677-27070677 on Build GRCh38
rs555453215 Chr.2: 27070528-27070528 on Build GRCh38
rs972275979 Chr.2: 27070838-27070838 on Build GRCh38
rs984532784 Chr.2: 27070857-27070857 on Build GRCh38
rs551352170 Chr.2: 27070511-27070511 on Build GRCh38
rs960656689 Chr.2: 27070587-27070587 on Build GRCh38
rs902034045 Chr.2: 27070624-27070624 on Build GRCh38
rs964582692 Chr.2: 27070730-27070730 on Build GRCh38
rs576376692 Chr.2: 27070850-27070851 on Build GRCh38
rs943087106 Chr.2: 27070496-27070496 on Build GRCh38
rs556530355 Chr.2: 27070676-27070676 on Build GRCh38
rs538002310 Chr.2: 27070607-27070607 on Build GRCh38

View Full Product Details