Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27445847-27446343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT172
Entrez Gene ID
26160
Gene Name
intraflagellar transport 172
Gene Aliases
BBS20, NPHP17, RP71, SLB, SRTD10, osm-1, wim
UniGene
Hs.127401
Gene Chromosome Location
Chr.2: 27444373-27489819 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
UniGene
Hs.59509
Gene Chromosome Location
Chr.2: 27442366-27446481 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4826416 c.4675A>G p.R1559G Chr.2: 27446340-27446340 on Build GRCh38
281864 c.4721A>G p.D1574G Chr.2: 27446294-27446294 on Build GRCh38
5058856 c.4787T>G p.F1596C Chr.2: 27445957-27445957 on Build GRCh38
6261122 c.4701C>A p.H1567Q Chr.2: 27446314-27446314 on Build GRCh38
6684031 c.4672G>A p.A1558T Chr.2: 27446343-27446343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186020523 Chr.2: 27445949-27445949 on Build GRCh38
rs778725103 Chr.2: 27446244-27446244 on Build GRCh38
rs760183362 Chr.2: 27445993-27445993 on Build GRCh38
rs780339054 Chr.2: 27445934-27445934 on Build GRCh38
rs200328536 Chr.2: 27446032-27446032 on Build GRCh38
rs114253314 Chr.2: 27446150-27446150 on Build GRCh38
rs768149033 Chr.2: 27446260-27446260 on Build GRCh38
rs773088930 Chr.2: 27445900-27445900 on Build GRCh38
rs759461544 Chr.2: 27445919-27445919 on Build GRCh38
rs745459521 Chr.2: 27446033-27446036 on Build GRCh38
rs763680759 Chr.2: 27446004-27446004 on Build GRCh38
rs773569919 Chr.2: 27445969-27445969 on Build GRCh38
rs375261962 Chr.2: 27446251-27446251 on Build GRCh38
rs541271340 Chr.2: 27446107-27446107 on Build GRCh38
rs747003975 Chr.2: 27445882-27445882 on Build GRCh38
rs770189360 Chr.2: 27445897-27445897 on Build GRCh38
rs753517897 Chr.2: 27446005-27446005 on Build GRCh38
rs786205855 Chr.2: 27446314-27446314 on Build GRCh38
rs756273133 Chr.2: 27446223-27446223 on Build GRCh38
rs369307448 Chr.2: 27446229-27446229 on Build GRCh38
rs774056784 Chr.2: 27445973-27445973 on Build GRCh38
rs61742074 Chr.2: 27446270-27446270 on Build GRCh38
rs1042644207 Chr.2: 27445968-27445968 on Build GRCh38
rs139979160 Chr.2: 27446133-27446135 on Build GRCh38
rs771110267 Chr.2: 27446235-27446235 on Build GRCh38
rs982399754 Chr.2: 27446002-27446003 on Build GRCh38
rs768767140 Chr.2: 27445883-27445883 on Build GRCh38
rs115716101 Chr.2: 27445933-27445933 on Build GRCh38
rs369400518 Chr.2: 27445932-27445932 on Build GRCh38
rs780617403 Chr.2: 27445881-27445881 on Build GRCh38
rs761437797 Chr.2: 27446017-27446017 on Build GRCh38
rs748974533 Chr.2: 27445970-27445970 on Build GRCh38
rs181365583 Chr.2: 27445906-27445906 on Build GRCh38
rs756750247 Chr.2: 27445919-27445919 on Build GRCh38
rs761504464 Chr.2: 27446282-27446282 on Build GRCh38
rs894833948 Chr.2: 27446041-27446041 on Build GRCh38
rs148228238 Chr.2: 27446025-27446025 on Build GRCh38
rs371490518 Chr.2: 27445910-27445910 on Build GRCh38
rs748257330 Chr.2: 27445956-27445956 on Build GRCh38
rs751918930 Chr.2: 27446034-27446034 on Build GRCh38
rs994445963 Chr.2: 27446155-27446155 on Build GRCh38
rs376189412 Chr.2: 27446137-27446139 on Build GRCh38
rs878941908 Chr.2: 27446234-27446234 on Build GRCh38
rs915032282 Chr.2: 27446318-27446318 on Build GRCh38
rs774356097 Chr.2: 27445917-27445917 on Build GRCh38
rs369487724 Chr.2: 27446252-27446252 on Build GRCh38
rs755462662 Chr.2: 27446040-27446040 on Build GRCh38
rs558185038 Chr.2: 27446213-27446213 on Build GRCh38
rs749498090 Chr.2: 27446231-27446231 on Build GRCh38
rs190647362 Chr.2: 27446029-27446029 on Build GRCh38
rs770097378 Chr.2: 27445965-27445965 on Build GRCh38
rs199667794 Chr.2: 27445920-27445920 on Build GRCh38
rs755477787 Chr.2: 27445951-27445951 on Build GRCh38
rs377200724 Chr.2: 27445952-27445952 on Build GRCh38
rs772502337 Chr.2: 27446238-27446239 on Build GRCh38
rs781771585 Chr.2: 27446209-27446209 on Build GRCh38
rs1006823501 Chr.2: 27446196-27446196 on Build GRCh38
rs759979976 Chr.2: 27446312-27446312 on Build GRCh38
rs748832823 Chr.2: 27445906-27445907 on Build GRCh38
rs188659223 Chr.2: 27445884-27445884 on Build GRCh38
rs1026821400 Chr.2: 27446167-27446167 on Build GRCh38
rs764818890 Chr.2: 27446283-27446283 on Build GRCh38
rs1030462015 Chr.2: 27446272-27446272 on Build GRCh38
rs786205856 Chr.2: 27445929-27445929 on Build GRCh38
rs768909809 Chr.2: 27446098-27446098 on Build GRCh38
rs573058783 Chr.2: 27446154-27446154 on Build GRCh38
rs571624967 Chr.2: 27446299-27446299 on Build GRCh38
rs944310654 Chr.2: 27446115-27446115 on Build GRCh38
rs776563892 Chr.2: 27446247-27446247 on Build GRCh38
rs372748754 Chr.2: 27446091-27446091 on Build GRCh38
rs772023412 Chr.2: 27445976-27445976 on Build GRCh38
rs897338264 Chr.2: 27446124-27446124 on Build GRCh38
rs770815937 Chr.2: 27445879-27445879 on Build GRCh38
rs371445480 Chr.2: 27446026-27446026 on Build GRCh38
rs971103505 Chr.2: 27446204-27446204 on Build GRCh38
rs778607708 Chr.2: 27446030-27446037 on Build GRCh38
rs755717990 Chr.2: 27445931-27445931 on Build GRCh38
rs774941853 Chr.2: 27445985-27445985 on Build GRCh38
rs770759155 Chr.2: 27445971-27445971 on Build GRCh38
rs970633023 Chr.2: 27445994-27445994 on Build GRCh38
rs759499880 Chr.2: 27445974-27445974 on Build GRCh38
rs923806433 Chr.2: 27446022-27446022 on Build GRCh38
rs141196620 Chr.2: 27446173-27446173 on Build GRCh38
rs752257485 Chr.2: 27445928-27445928 on Build GRCh38
rs1041663658 Chr.2: 27446116-27446116 on Build GRCh38
rs775669156 Chr.2: 27446249-27446249 on Build GRCh38

View Full Product Details