Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27577242-27577750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf16
Entrez Gene ID
84226
Gene Name
chromosome 2 open reading frame 16
Gene Aliases
-
UniGene
Hs.131021
Gene Chromosome Location
Chr.2: 27576522-27582722 on Build GRCh38
Transcripts
NM_032266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5078189 c.1015T>C p.S339P Chr.2: 27577587-27577587 on Build GRCh38
4560653 c.851G>A p.R284K Chr.2: 27577423-27577423 on Build GRCh38
5701725 c.674_675CA>AG p.P225Q Chr.2: 27577246-27577247 on Build GRCh38
1019749 c.762G>A p.K254K Chr.2: 27577334-27577334 on Build GRCh38
6158096 c.772C>A p.Q258K Chr.2: 27577344-27577344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779096554 Chr.2: 27577349-27577350 on Build GRCh38
rs773169069 Chr.2: 27577423-27577423 on Build GRCh38
rs200949260 Chr.2: 27577626-27577626 on Build GRCh38
rs776054946 Chr.2: 27577688-27577692 on Build GRCh38
rs200280226 Chr.2: 27577302-27577302 on Build GRCh38
rs759807559 Chr.2: 27577606-27577606 on Build GRCh38
rs759292446 Chr.2: 27577461-27577461 on Build GRCh38
rs746215772 Chr.2: 27577362-27577362 on Build GRCh38
rs754923529 Chr.2: 27577267-27577267 on Build GRCh38
rs765273018 Chr.2: 27577316-27577316 on Build GRCh38
rs34687163 Chr.2: 27577320-27577321 on Build GRCh38
rs865896322 Chr.2: 27577579-27577579 on Build GRCh38
rs549844411 Chr.2: 27577638-27577638 on Build GRCh38
rs766607597 Chr.2: 27577436-27577436 on Build GRCh38
rs769285147 Chr.2: 27577290-27577290 on Build GRCh38
rs945826851 Chr.2: 27577336-27577336 on Build GRCh38
rs79199753 Chr.2: 27577456-27577456 on Build GRCh38
rs769191389 Chr.2: 27577385-27577385 on Build GRCh38
rs778678793 Chr.2: 27577537-27577537 on Build GRCh38
rs781133084 Chr.2: 27577341-27577341 on Build GRCh38
rs756932913 Chr.2: 27577525-27577525 on Build GRCh38
rs752452500 Chr.2: 27577618-27577618 on Build GRCh38
rs770215020 Chr.2: 27577570-27577570 on Build GRCh38
rs758607103 Chr.2: 27577322-27577322 on Build GRCh38
rs561517202 Chr.2: 27577418-27577418 on Build GRCh38
rs772507454 Chr.2: 27577452-27577456 on Build GRCh38
rs767236969 Chr.2: 27577613-27577613 on Build GRCh38
rs763972463 Chr.2: 27577632-27577632 on Build GRCh38
rs923994754 Chr.2: 27577338-27577338 on Build GRCh38
rs866349708 Chr.2: 27577437-27577437 on Build GRCh38
rs897337644 Chr.2: 27577453-27577453 on Build GRCh38
rs756013829 Chr.2: 27577490-27577490 on Build GRCh38
rs372273777 Chr.2: 27577647-27577647 on Build GRCh38
rs765741039 Chr.2: 27577711-27577711 on Build GRCh38
rs769130005 Chr.2: 27577552-27577552 on Build GRCh38
rs76077128 Chr.2: 27577314-27577314 on Build GRCh38
rs780616042 Chr.2: 27577270-27577270 on Build GRCh38
rs755622636 Chr.2: 27577354-27577354 on Build GRCh38
rs994268451 Chr.2: 27577477-27577477 on Build GRCh38
rs139534811 Chr.2: 27577296-27577296 on Build GRCh38
rs368267439 Chr.2: 27577642-27577642 on Build GRCh38
rs202171403 Chr.2: 27577708-27577708 on Build GRCh38
rs754873095 Chr.2: 27577340-27577340 on Build GRCh38
rs761929727 Chr.2: 27577303-27577303 on Build GRCh38
rs750653893 Chr.2: 27577321-27577321 on Build GRCh38
rs770981012 Chr.2: 27577669-27577669 on Build GRCh38
rs763049275 Chr.2: 27577587-27577587 on Build GRCh38
rs776006855 Chr.2: 27577300-27577300 on Build GRCh38
rs1027159955 Chr.2: 27577288-27577288 on Build GRCh38
rs374394016 Chr.2: 27577297-27577297 on Build GRCh38
rs772902411 Chr.2: 27577294-27577294 on Build GRCh38
rs781476417 Chr.2: 27577559-27577559 on Build GRCh38
rs17006143 Chr.2: 27577641-27577641 on Build GRCh38
rs753313270 Chr.2: 27577515-27577515 on Build GRCh38
rs376678059 Chr.2: 27577657-27577657 on Build GRCh38
rs771036151 Chr.2: 27577593-27577593 on Build GRCh38
rs199971484 Chr.2: 27577371-27577371 on Build GRCh38
rs370067086 Chr.2: 27577412-27577412 on Build GRCh38
rs770708739 Chr.2: 27577372-27577372 on Build GRCh38
rs752630232 Chr.2: 27577345-27577345 on Build GRCh38
rs565366105 Chr.2: 27577680-27577680 on Build GRCh38
rs143452806 Chr.2: 27577334-27577334 on Build GRCh38
rs952989347 Chr.2: 27577539-27577539 on Build GRCh38
rs1055566475 Chr.2: 27577383-27577383 on Build GRCh38
rs777246659 Chr.2: 27577407-27577407 on Build GRCh38
rs1027212185 Chr.2: 27577486-27577486 on Build GRCh38
rs578053563 Chr.2: 27577325-27577325 on Build GRCh38
rs774530891 Chr.2: 27577598-27577598 on Build GRCh38
rs751312239 Chr.2: 27577454-27577454 on Build GRCh38
rs767345265 Chr.2: 27577474-27577474 on Build GRCh38
rs376483751 Chr.2: 27577573-27577573 on Build GRCh38
rs371389804 Chr.2: 27577664-27577664 on Build GRCh38
rs777282378 Chr.2: 27577505-27577505 on Build GRCh38
rs748621531 Chr.2: 27577563-27577563 on Build GRCh38
rs747727945 Chr.2: 27577287-27577287 on Build GRCh38
rs998065515 Chr.2: 27577538-27577538 on Build GRCh38
rs769912897 Chr.2: 27577413-27577413 on Build GRCh38
rs1043006257 Chr.2: 27577358-27577358 on Build GRCh38
rs747398801 Chr.2: 27577373-27577373 on Build GRCh38
rs531851124 Chr.2: 27577475-27577475 on Build GRCh38
rs143854816 Chr.2: 27577356-27577356 on Build GRCh38
rs374985722 Chr.2: 27577428-27577428 on Build GRCh38
rs374225053 Chr.2: 27577628-27577628 on Build GRCh38
rs939424826 Chr.2: 27577364-27577364 on Build GRCh38
rs367710159 Chr.2: 27577662-27577662 on Build GRCh38
rs1014127877 Chr.2: 27577416-27577416 on Build GRCh38

View Full Product Details