Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 28413802-28414309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOSL2
Entrez Gene ID
2355
Gene Name
FOS like 2, AP-1 transcription factor subunit
Gene Aliases
FRA2
UniGene
Hs.220971
Gene Chromosome Location
Chr.2: 28392759-28417312 on Build GRCh38
Transcripts
NM_005253.3 XM_006711976.2 XM_006711977.3 XM_017003737.1

SNP Detail

SNP ID Chromosome Location
rs2279990 Chr.2: 28413873-28413873 on Build GRCh38
rs1040182992 Chr.2: 28414229-28414229 on Build GRCh38
rs1009954516 Chr.2: 28414070-28414070 on Build GRCh38
rs921799877 Chr.2: 28414247-28414247 on Build GRCh38
rs762079686 Chr.2: 28414172-28414173 on Build GRCh38
rs551221719 Chr.2: 28414276-28414276 on Build GRCh38
rs1046687007 Chr.2: 28414146-28414146 on Build GRCh38
rs967020009 Chr.2: 28413991-28413991 on Build GRCh38
rs370380022 Chr.2: 28414284-28414284 on Build GRCh38
rs747054048 Chr.2: 28414157-28414157 on Build GRCh38
rs184499178 Chr.2: 28413924-28413924 on Build GRCh38
rs991645221 Chr.2: 28414108-28414108 on Build GRCh38
rs368166363 Chr.2: 28413892-28413892 on Build GRCh38
rs564770709 Chr.2: 28413935-28413935 on Build GRCh38
rs949549712 Chr.2: 28414145-28414145 on Build GRCh38
rs1000056026 Chr.2: 28414267-28414267 on Build GRCh38
rs949703210 Chr.2: 28413835-28413835 on Build GRCh38
rs562022145 Chr.2: 28414107-28414107 on Build GRCh38
rs967238013 Chr.2: 28414260-28414260 on Build GRCh38
rs776395832 Chr.2: 28413900-28413900 on Build GRCh38
rs368760725 Chr.2: 28414224-28414224 on Build GRCh38
rs916946705 Chr.2: 28414281-28414281 on Build GRCh38
rs975897517 Chr.2: 28414245-28414245 on Build GRCh38
rs978372691 Chr.2: 28414079-28414079 on Build GRCh38
rs768885340 Chr.2: 28414170-28414170 on Build GRCh38
rs572834271 Chr.2: 28414264-28414265 on Build GRCh38
rs566800110 Chr.2: 28414283-28414283 on Build GRCh38
rs888424727 Chr.2: 28414183-28414183 on Build GRCh38
rs1020123095 Chr.2: 28413978-28413978 on Build GRCh38
rs900409388 Chr.2: 28413919-28413919 on Build GRCh38
rs529424267 Chr.2: 28414152-28414152 on Build GRCh38
rs1047184523 Chr.2: 28413833-28413833 on Build GRCh38
rs550680485 Chr.2: 28414160-28414160 on Build GRCh38
rs1032718957 Chr.2: 28414271-28414271 on Build GRCh38
rs568686120 Chr.2: 28414184-28414184 on Build GRCh38
rs763775249 Chr.2: 28414231-28414231 on Build GRCh38
rs930066431 Chr.2: 28413830-28413830 on Build GRCh38
rs991262302 Chr.2: 28414278-28414278 on Build GRCh38
rs546488990 Chr.2: 28413925-28413925 on Build GRCh38
rs181203381 Chr.2: 28414254-28414254 on Build GRCh38
rs762690573 Chr.2: 28414185-28414185 on Build GRCh38
rs866501050 Chr.2: 28414218-28414218 on Build GRCh38
rs547009818 Chr.2: 28414096-28414096 on Build GRCh38
rs189987815 Chr.2: 28414131-28414131 on Build GRCh38
rs888511190 Chr.2: 28413915-28413915 on Build GRCh38
rs775493248 Chr.2: 28413994-28413994 on Build GRCh38
rs780114265 Chr.2: 28414043-28414043 on Build GRCh38
rs951461224 Chr.2: 28414188-28414188 on Build GRCh38
rs892828197 Chr.2: 28413966-28413966 on Build GRCh38
rs752456290 Chr.2: 28414104-28414104 on Build GRCh38
rs982842905 Chr.2: 28414194-28414194 on Build GRCh38
rs897167074 Chr.2: 28413895-28413895 on Build GRCh38
rs925717844 Chr.2: 28414100-28414100 on Build GRCh38
rs974763483 Chr.2: 28414162-28414162 on Build GRCh38
rs929975296 Chr.2: 28414159-28414159 on Build GRCh38
rs1040288132 Chr.2: 28413897-28413897 on Build GRCh38
rs759420095 Chr.2: 28413903-28413903 on Build GRCh38
rs539253070 Chr.2: 28414255-28414255 on Build GRCh38
rs894641597 Chr.2: 28414285-28414285 on Build GRCh38
rs1020008948 Chr.2: 28414095-28414095 on Build GRCh38
rs77148777 Chr.2: 28413949-28413949 on Build GRCh38
rs965582666 Chr.2: 28413947-28413947 on Build GRCh38
rs1027092623 Chr.2: 28413906-28413906 on Build GRCh38
rs1018129141 Chr.2: 28413941-28413941 on Build GRCh38

View Full Product Details