Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32170969-32171490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A6
Entrez Gene ID
55676
Gene Name
solute carrier family 30 member 6
Gene Aliases
MST103, MSTP103, ZNT6
UniGene
Hs.23248
Gene Chromosome Location
Chr.2: 32165841-32224806 on Build GRCh38
Transcripts
NM_001193513.1 NM_001193514.1 NM_001193515.1 NM_017964.3 XM_006712048.3 XM_011532959.2 XM_011532960.2 XM_011532961.2 XM_011532962.2 XM_011532963.2 XM_017004462.1 XM_017004463.1 XM_017004464.1 XM_017004465.1 XM_017004466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2942042 c.40delT p.F15fs*11 Chr.2: 32171323-32171323 on Build GRCh38
2942041 c.23G>A p.R8Q Chr.2: 32171306-32171306 on Build GRCh38
1185873 c.28C>A p.P10T Chr.2: 32171311-32171311 on Build GRCh38
5058874 c.85C>T p.R29* Chr.2: 32171368-32171368 on Build GRCh38
2942044 c.86G>A p.R29Q Chr.2: 32171369-32171369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007060296 Chr.2: 32171173-32171173 on Build GRCh38
rs904393777 Chr.2: 32171134-32171134 on Build GRCh38
rs1057209407 Chr.2: 32171086-32171086 on Build GRCh38
rs768453562 Chr.2: 32171402-32171402 on Build GRCh38
rs770211457 Chr.2: 32171261-32171261 on Build GRCh38
rs961431840 Chr.2: 32171272-32171272 on Build GRCh38
rs567537876 Chr.2: 32171312-32171312 on Build GRCh38
rs543490397 Chr.2: 32171408-32171408 on Build GRCh38
rs1028774385 Chr.2: 32171225-32171225 on Build GRCh38
rs531896252 Chr.2: 32171097-32171097 on Build GRCh38
rs1013119904 Chr.2: 32171112-32171112 on Build GRCh38
rs764083767 Chr.2: 32171316-32171316 on Build GRCh38
rs151181468 Chr.2: 32171368-32171368 on Build GRCh38
rs761771699 Chr.2: 32171417-32171417 on Build GRCh38
rs1039874196 Chr.2: 32171453-32171453 on Build GRCh38
rs758094701 Chr.2: 32171351-32171351 on Build GRCh38
rs752766021 Chr.2: 32171462-32171462 on Build GRCh38
rs766828532 Chr.2: 32171014-32171014 on Build GRCh38
rs375008557 Chr.2: 32171244-32171244 on Build GRCh38
rs980594409 Chr.2: 32171307-32171307 on Build GRCh38
rs745547656 Chr.2: 32171270-32171270 on Build GRCh38
rs939581268 Chr.2: 32171084-32171084 on Build GRCh38
rs774503688 Chr.2: 32171306-32171306 on Build GRCh38
rs78059008 Chr.2: 32171123-32171123 on Build GRCh38
rs929623459 Chr.2: 32171394-32171394 on Build GRCh38
rs776341538 Chr.2: 32171403-32171403 on Build GRCh38
rs1050831790 Chr.2: 32171151-32171151 on Build GRCh38
rs1050715788 Chr.2: 32171432-32171432 on Build GRCh38
rs950217697 Chr.2: 32171002-32171003 on Build GRCh38
rs775402569 Chr.2: 32171313-32171313 on Build GRCh38
rs773649298 Chr.2: 32171279-32171279 on Build GRCh38
rs752391495 Chr.2: 32171241-32171241 on Build GRCh38
rs764733512 Chr.2: 32171341-32171341 on Build GRCh38
rs770385337 Chr.2: 32171296-32171296 on Build GRCh38
rs757924302 Chr.2: 32171289-32171289 on Build GRCh38
rs763624284 Chr.2: 32171323-32171323 on Build GRCh38
rs912235486 Chr.2: 32171433-32171433 on Build GRCh38
rs903704094 Chr.2: 32171193-32171193 on Build GRCh38
rs534453447 Chr.2: 32171369-32171369 on Build GRCh38
rs543325005 Chr.2: 32171072-32171072 on Build GRCh38
rs563213555 Chr.2: 32171094-32171094 on Build GRCh38
rs763995504 Chr.2: 32171245-32171245 on Build GRCh38
rs79234407 Chr.2: 32171251-32171251 on Build GRCh38
rs145648952 Chr.2: 32171342-32171342 on Build GRCh38
rs756754390 Chr.2: 32171266-32171266 on Build GRCh38
rs890914745 Chr.2: 32171166-32171166 on Build GRCh38
rs778222757 Chr.2: 32171359-32171359 on Build GRCh38
rs374555800 Chr.2: 32171301-32171301 on Build GRCh38
rs141776327 Chr.2: 32171129-32171129 on Build GRCh38
rs200625427 Chr.2: 32171391-32171391 on Build GRCh38
rs998076666 Chr.2: 32171217-32171217 on Build GRCh38
rs772134463 Chr.2: 32171311-32171311 on Build GRCh38
rs928221379 Chr.2: 32171022-32171022 on Build GRCh38
rs565895310 Chr.2: 32171153-32171153 on Build GRCh38
rs756546731 Chr.2: 32171357-32171357 on Build GRCh38
rs748416148 Chr.2: 32171293-32171293 on Build GRCh38
rs760576412 Chr.2: 32171392-32171392 on Build GRCh38
rs527808026 Chr.2: 32171176-32171176 on Build GRCh38
rs765112957 Chr.2: 32171418-32171418 on Build GRCh38
rs879003991 Chr.2: 32171413-32171413 on Build GRCh38
rs370423424 Chr.2: 32171352-32171352 on Build GRCh38
rs779070448 Chr.2: 32171373-32171373 on Build GRCh38
rs537608531 Chr.2: 32171449-32171449 on Build GRCh38
rs377700984 Chr.2: 32171314-32171314 on Build GRCh38
rs548863982 Chr.2: 32171385-32171385 on Build GRCh38
rs980327389 Chr.2: 32171020-32171020 on Build GRCh38
rs867006653 Chr.2: 32171384-32171384 on Build GRCh38
rs142115336 Chr.2: 32171305-32171305 on Build GRCh38
rs1042672520 Chr.2: 32171393-32171393 on Build GRCh38
rs927458825 Chr.2: 32171041-32171041 on Build GRCh38
rs745911029 Chr.2: 32171378-32171378 on Build GRCh38
rs1039043287 Chr.2: 32171093-32171093 on Build GRCh38
rs778595498 Chr.2: 32171269-32171269 on Build GRCh38
rs998657107 Chr.2: 32171147-32171147 on Build GRCh38
rs753763685 Chr.2: 32171327-32171327 on Build GRCh38
rs761681616 Chr.2: 32171329-32171329 on Build GRCh38
rs781644651 Chr.2: 32171290-32171290 on Build GRCh38
rs1031441201 Chr.2: 32171172-32171172 on Build GRCh38
rs942442724 Chr.2: 32171437-32171437 on Build GRCh38
rs963234697 Chr.2: 32171345-32171345 on Build GRCh38

View Full Product Details