Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37250424-37250933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKD3
Entrez Gene ID
23683
Gene Name
protein kinase D3
Gene Aliases
EPK2, PKC-NU, PKD3, PRKCN, nPKC-NU
UniGene
Hs.660757
Gene Chromosome Location
Chr.2: 37250502-37324803 on Build GRCh38
Transcripts
NM_005813.4 XM_005264237.3 XM_005264238.3 XM_017003741.1 XM_017003742.1 XM_017003743.1 XM_017003744.1

SNP Detail

SNP ID Chromosome Location
rs987021210 Chr.2: 37250717-37250717 on Build GRCh38
rs986565551 Chr.2: 37250823-37250823 on Build GRCh38
rs750105574 Chr.2: 37250577-37250577 on Build GRCh38
rs397941182 Chr.2: 37250780-37250781 on Build GRCh38
rs11449539 Chr.2: 37250773-37250774 on Build GRCh38
rs939713470 Chr.2: 37250572-37250572 on Build GRCh38
rs1033399324 Chr.2: 37250700-37250700 on Build GRCh38
rs8243 Chr.2: 37250651-37250651 on Build GRCh38
rs550500658 Chr.2: 37250738-37250738 on Build GRCh38
rs1024403813 Chr.2: 37250669-37250669 on Build GRCh38
rs561619293 Chr.2: 37250594-37250594 on Build GRCh38
rs142689216 Chr.2: 37250828-37250828 on Build GRCh38
rs1024442507 Chr.2: 37250574-37250574 on Build GRCh38
rs564837742 Chr.2: 37250526-37250527 on Build GRCh38
rs903226844 Chr.2: 37250688-37250688 on Build GRCh38
rs9318 Chr.2: 37250771-37250771 on Build GRCh38
rs933808761 Chr.2: 37250534-37250534 on Build GRCh38
rs191184528 Chr.2: 37250573-37250573 on Build GRCh38
rs1032943046 Chr.2: 37250549-37250549 on Build GRCh38
rs541490119 Chr.2: 37250481-37250481 on Build GRCh38
rs564792147 Chr.2: 37250609-37250609 on Build GRCh38
rs973834462 Chr.2: 37250896-37250896 on Build GRCh38
rs767819072 Chr.2: 37250543-37250543 on Build GRCh38
rs1014422768 Chr.2: 37250667-37250667 on Build GRCh38
rs764381522 Chr.2: 37250652-37250652 on Build GRCh38
rs903084325 Chr.2: 37250523-37250523 on Build GRCh38
rs954320130 Chr.2: 37250673-37250673 on Build GRCh38
rs891223671 Chr.2: 37250467-37250467 on Build GRCh38
rs574102555 Chr.2: 37250461-37250461 on Build GRCh38
rs919607835 Chr.2: 37250900-37250900 on Build GRCh38
rs1042451564 Chr.2: 37250517-37250518 on Build GRCh38
rs1015239947 Chr.2: 37250633-37250633 on Build GRCh38
rs79585227 Chr.2: 37250509-37250509 on Build GRCh38
rs974704875 Chr.2: 37250665-37250666 on Build GRCh38
rs998886286 Chr.2: 37250690-37250690 on Build GRCh38
rs955244037 Chr.2: 37250805-37250805 on Build GRCh38
rs1028195686 Chr.2: 37250804-37250804 on Build GRCh38
rs1054743548 Chr.2: 37250868-37250873 on Build GRCh38
rs908294378 Chr.2: 37250561-37250561 on Build GRCh38
rs568425740 Chr.2: 37250748-37250748 on Build GRCh38
rs986554278 Chr.2: 37250537-37250537 on Build GRCh38
rs1046927 Chr.2: 37250862-37250862 on Build GRCh38
rs921529346 Chr.2: 37250666-37250666 on Build GRCh38
rs183795605 Chr.2: 37250583-37250583 on Build GRCh38
rs1042400166 Chr.2: 37250785-37250785 on Build GRCh38
rs957382172 Chr.2: 37250711-37250711 on Build GRCh38
rs912648517 Chr.2: 37250740-37250740 on Build GRCh38

View Full Product Details