Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38733964-38734501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALM
Entrez Gene ID
130589
Gene Name
galactose mutarotase
Gene Aliases
BLOCK25, GLAT, HEL-S-63p, IBD1
UniGene
Hs.435012
Gene Chromosome Location
Chr.2: 38665910-38734767 on Build GRCh38
Transcripts
NM_138801.2

SNP Detail

SNP ID Chromosome Location
rs1037054545 Chr.2: 38734339-38734339 on Build GRCh38
rs986549215 Chr.2: 38734191-38734191 on Build GRCh38
rs113589850 Chr.2: 38734272-38734272 on Build GRCh38
rs1015575813 Chr.2: 38734007-38734007 on Build GRCh38
rs113777936 Chr.2: 38734262-38734262 on Build GRCh38
rs536120608 Chr.2: 38734174-38734174 on Build GRCh38
rs192626541 Chr.2: 38734449-38734449 on Build GRCh38
rs867852737 Chr.2: 38734428-38734428 on Build GRCh38
rs575916563 Chr.2: 38734321-38734321 on Build GRCh38
rs920911958 Chr.2: 38734361-38734361 on Build GRCh38
rs1054049045 Chr.2: 38734165-38734165 on Build GRCh38
rs891180291 Chr.2: 38733997-38733997 on Build GRCh38
rs924909730 Chr.2: 38734250-38734250 on Build GRCh38
rs916109961 Chr.2: 38734265-38734265 on Build GRCh38
rs962298541 Chr.2: 38734038-38734038 on Build GRCh38
rs75847197 Chr.2: 38734386-38734386 on Build GRCh38
rs973335131 Chr.2: 38734075-38734075 on Build GRCh38
rs774358903 Chr.2: 38734035-38734035 on Build GRCh38
rs971942130 Chr.2: 38734313-38734313 on Build GRCh38
rs1004639011 Chr.2: 38734004-38734004 on Build GRCh38
rs185620462 Chr.2: 38734477-38734477 on Build GRCh38
rs917407823 Chr.2: 38734125-38734125 on Build GRCh38
rs977623680 Chr.2: 38734325-38734325 on Build GRCh38
rs1050826423 Chr.2: 38734180-38734180 on Build GRCh38
rs924899593 Chr.2: 38734159-38734159 on Build GRCh38
rs557239418 Chr.2: 38734294-38734294 on Build GRCh38
rs569139967 Chr.2: 38734152-38734152 on Build GRCh38
rs907295675 Chr.2: 38734289-38734289 on Build GRCh38
rs145849530 Chr.2: 38734017-38734017 on Build GRCh38
rs945086716 Chr.2: 38734205-38734205 on Build GRCh38
rs571354827 Chr.2: 38734121-38734121 on Build GRCh38
rs995467853 Chr.2: 38734367-38734367 on Build GRCh38
rs978051272 Chr.2: 38734134-38734134 on Build GRCh38
rs532383518 Chr.2: 38734124-38734124 on Build GRCh38
rs191652284 Chr.2: 38734127-38734127 on Build GRCh38
rs372608625 Chr.2: 38734427-38734427 on Build GRCh38
rs990393019 Chr.2: 38734356-38734356 on Build GRCh38
rs546751843 Chr.2: 38734077-38734077 on Build GRCh38
rs911661178 Chr.2: 38734404-38734404 on Build GRCh38
rs957638690 Chr.2: 38734346-38734346 on Build GRCh38
rs907243320 Chr.2: 38734106-38734106 on Build GRCh38
rs935629309 Chr.2: 38734164-38734164 on Build GRCh38
rs534435127 Chr.2: 38734417-38734418 on Build GRCh38
rs187610350 Chr.2: 38734429-38734429 on Build GRCh38
rs1138974 Chr.2: 38734267-38734267 on Build GRCh38
rs936284968 Chr.2: 38734261-38734261 on Build GRCh38
rs368722657 Chr.2: 38734421-38734421 on Build GRCh38
rs920965729 Chr.2: 38734173-38734173 on Build GRCh38
rs369352641 Chr.2: 38734383-38734384 on Build GRCh38
rs890931669 Chr.2: 38734194-38734194 on Build GRCh38
rs70954749 Chr.2: 38734384-38734385 on Build GRCh38
rs561322616 Chr.2: 38734042-38734044 on Build GRCh38
rs1037322966 Chr.2: 38734214-38734214 on Build GRCh38
rs1008016653 Chr.2: 38734420-38734420 on Build GRCh38
rs1138975 Chr.2: 38734273-38734273 on Build GRCh38
rs924815079 Chr.2: 38734342-38734342 on Build GRCh38
rs528070215 Chr.2: 38734036-38734036 on Build GRCh38
rs912308129 Chr.2: 38734198-38734198 on Build GRCh38
rs953869432 Chr.2: 38734181-38734181 on Build GRCh38
rs77428435 Chr.2: 38734226-38734226 on Build GRCh38
rs771445427 Chr.2: 38734026-38734027 on Build GRCh38
rs1024381104 Chr.2: 38734102-38734102 on Build GRCh38
rs368177329 Chr.2: 38734029-38734029 on Build GRCh38
rs74857574 Chr.2: 38734385-38734385 on Build GRCh38
rs866073477 Chr.2: 38734408-38734408 on Build GRCh38
rs72909121 Chr.2: 38734360-38734360 on Build GRCh38
rs1027528999 Chr.2: 38734117-38734117 on Build GRCh38
rs898414146 Chr.2: 38734222-38734222 on Build GRCh38
rs112911853 Chr.2: 38734426-38734426 on Build GRCh38
rs1004062841 Chr.2: 38734209-38734209 on Build GRCh38
rs1004219382 Chr.2: 38734123-38734123 on Build GRCh38

View Full Product Details