Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42570177-42570576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTA3
Entrez Gene ID
57504
Gene Name
metastasis associated 1 family member 3
Gene Aliases
-
UniGene
Hs.435413
Gene Chromosome Location
Chr.2: 42494569-42795032 on Build GRCh38
Transcripts
NM_001282755.1 NM_001282756.1 NM_020744.3 XM_005264456.2 XM_005264458.2 XM_005264459.2 XM_017004561.1 XM_017004562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1408092 c.53C>A p.S18Y Chr.2: 42570461-42570461 on Build GRCh38
5756066 c.85G>T p.E29* Chr.2: 42570493-42570493 on Build GRCh38
5018294 c.96delG p.T33fs*10 Chr.2: 42570504-42570504 on Build GRCh38
1662869 c.55A>G p.S19G Chr.2: 42570463-42570463 on Build GRCh38
5015941 c.30delT p.Y11fs*13 Chr.2: 42570438-42570438 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569358706 Chr.2: 42570337-42570337 on Build GRCh38
rs942229196 Chr.2: 42570368-42570368 on Build GRCh38
rs979724024 Chr.2: 42570335-42570335 on Build GRCh38
rs368916301 Chr.2: 42570216-42570216 on Build GRCh38
rs919406453 Chr.2: 42570386-42570386 on Build GRCh38
rs1017478994 Chr.2: 42570246-42570246 on Build GRCh38
rs756683649 Chr.2: 42570504-42570504 on Build GRCh38
rs765752505 Chr.2: 42570275-42570275 on Build GRCh38
rs746637934 Chr.2: 42570261-42570261 on Build GRCh38
rs769793345 Chr.2: 42570388-42570388 on Build GRCh38
rs752231642 Chr.2: 42570521-42570521 on Build GRCh38
rs1051848600 Chr.2: 42570418-42570418 on Build GRCh38
rs777438500 Chr.2: 42570527-42570527 on Build GRCh38
rs929408861 Chr.2: 42570406-42570406 on Build GRCh38
rs767788336 Chr.2: 42570478-42570478 on Build GRCh38
rs113073749 Chr.2: 42570268-42570268 on Build GRCh38
rs866077796 Chr.2: 42570451-42570451 on Build GRCh38
rs767032845 Chr.2: 42570505-42570505 on Build GRCh38
rs199605489 Chr.2: 42570461-42570461 on Build GRCh38
rs969746343 Chr.2: 42570321-42570321 on Build GRCh38
rs892672614 Chr.2: 42570543-42570543 on Build GRCh38
rs371566563 Chr.2: 42570553-42570553 on Build GRCh38
rs1038982458 Chr.2: 42570539-42570539 on Build GRCh38
rs762172314 Chr.2: 42570468-42570468 on Build GRCh38
rs757861206 Chr.2: 42570242-42570242 on Build GRCh38
rs868150404 Chr.2: 42570458-42570458 on Build GRCh38
rs916938330 Chr.2: 42570317-42570317 on Build GRCh38
rs992799507 Chr.2: 42570264-42570264 on Build GRCh38
rs746642270 Chr.2: 42570532-42570532 on Build GRCh38
rs539031341 Chr.2: 42570237-42570237 on Build GRCh38
rs999999001 Chr.2: 42570265-42570265 on Build GRCh38
rs925371758 Chr.2: 42570357-42570357 on Build GRCh38
rs763743663 Chr.2: 42570393-42570393 on Build GRCh38
rs777417805 Chr.2: 42570260-42570261 on Build GRCh38
rs992579537 Chr.2: 42570315-42570315 on Build GRCh38
rs750491983 Chr.2: 42570428-42570428 on Build GRCh38
rs567173867 Chr.2: 42570219-42570219 on Build GRCh38
rs763073822 Chr.2: 42570402-42570402 on Build GRCh38
rs973586374 Chr.2: 42570378-42570378 on Build GRCh38
rs770665376 Chr.2: 42570266-42570266 on Build GRCh38
rs775520669 Chr.2: 42570394-42570394 on Build GRCh38
rs926868285 Chr.2: 42570350-42570350 on Build GRCh38
rs538364740 Chr.2: 42570413-42570413 on Build GRCh38
rs879240481 Chr.2: 42570316-42570316 on Build GRCh38
rs185501388 Chr.2: 42570208-42570208 on Build GRCh38
rs553281997 Chr.2: 42570206-42570206 on Build GRCh38
rs574637541 Chr.2: 42570379-42570379 on Build GRCh38
rs113619159 Chr.2: 42570202-42570202 on Build GRCh38
rs757016428 Chr.2: 42570534-42570534 on Build GRCh38
rs762352532 Chr.2: 42570224-42570224 on Build GRCh38
rs968571898 Chr.2: 42570257-42570257 on Build GRCh38
rs980827503 Chr.2: 42570304-42570304 on Build GRCh38
rs369055097 Chr.2: 42570459-42570459 on Build GRCh38
rs757864979 Chr.2: 42570525-42570525 on Build GRCh38
rs750873690 Chr.2: 42570501-42570501 on Build GRCh38

View Full Product Details