Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43292018-43292513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THADA
Entrez Gene ID
63892
Gene Name
THADA, armadillo repeat containing
Gene Aliases
ARMC13, GITA
UniGene
Hs.369592
Gene Chromosome Location
Chr.2: 43230836-43596046 on Build GRCh38
Transcripts
NM_001083953.1 NM_022065.4 NR_073394.1 XM_006712061.3 XM_006712062.1 XM_006712063.1 XM_006712064.2 XM_006712065.1 XM_006712066.1 XM_006712067.1 XM_006712069.3 XM_017004674.1 XM_017004675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1229123 c.4896G>T p.K1632N Chr.2: 43292145-43292145 on Build GRCh38
3380162 c.4831C>A p.L1611I Chr.2: 43292210-43292210 on Build GRCh38
6509293 c.4924G>T p.A1642S Chr.2: 43292117-43292117 on Build GRCh38
6091516 c.4866G>C p.Q1622H Chr.2: 43292175-43292175 on Build GRCh38
1020989 c.4869G>A p.T1623T Chr.2: 43292172-43292172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146912753 Chr.2: 43292164-43292164 on Build GRCh38
rs768127270 Chr.2: 43292078-43292078 on Build GRCh38
rs769583799 Chr.2: 43292174-43292174 on Build GRCh38
rs549164637 Chr.2: 43292066-43292066 on Build GRCh38
rs746792700 Chr.2: 43292104-43292104 on Build GRCh38
rs780110508 Chr.2: 43292178-43292178 on Build GRCh38
rs778319782 Chr.2: 43292086-43292086 on Build GRCh38
rs544472192 Chr.2: 43292346-43292346 on Build GRCh38
rs774651058 Chr.2: 43292197-43292197 on Build GRCh38
rs960754609 Chr.2: 43292357-43292357 on Build GRCh38
rs1028360248 Chr.2: 43292315-43292316 on Build GRCh38
rs752144060 Chr.2: 43292160-43292160 on Build GRCh38
rs777206654 Chr.2: 43292217-43292217 on Build GRCh38
rs998436438 Chr.2: 43292158-43292158 on Build GRCh38
rs761142574 Chr.2: 43292207-43292207 on Build GRCh38
rs983444227 Chr.2: 43292331-43292331 on Build GRCh38
rs748414681 Chr.2: 43292201-43292201 on Build GRCh38
rs372424140 Chr.2: 43292127-43292127 on Build GRCh38
rs768798061 Chr.2: 43292185-43292185 on Build GRCh38
rs757956921 Chr.2: 43292095-43292095 on Build GRCh38
rs752346798 Chr.2: 43292092-43292092 on Build GRCh38
rs749766574 Chr.2: 43292255-43292258 on Build GRCh38
rs201100629 Chr.2: 43292130-43292130 on Build GRCh38
rs991325212 Chr.2: 43292406-43292406 on Build GRCh38
rs1048052700 Chr.2: 43292364-43292364 on Build GRCh38
rs1043642609 Chr.2: 43292481-43292481 on Build GRCh38
rs562620125 Chr.2: 43292488-43292488 on Build GRCh38
rs915360803 Chr.2: 43292041-43292041 on Build GRCh38
rs750920926 Chr.2: 43292148-43292148 on Build GRCh38
rs78613857 Chr.2: 43292300-43292300 on Build GRCh38
rs369444028 Chr.2: 43292125-43292125 on Build GRCh38
rs977255973 Chr.2: 43292065-43292065 on Build GRCh38
rs916377669 Chr.2: 43292408-43292408 on Build GRCh38
rs977222960 Chr.2: 43292222-43292222 on Build GRCh38
rs769859816 Chr.2: 43292122-43292122 on Build GRCh38
rs565286049 Chr.2: 43292385-43292385 on Build GRCh38
rs764161008 Chr.2: 43292440-43292440 on Build GRCh38
rs139334985 Chr.2: 43292189-43292189 on Build GRCh38
rs950710640 Chr.2: 43292320-43292320 on Build GRCh38
rs777348090 Chr.2: 43292167-43292167 on Build GRCh38
rs202161314 Chr.2: 43292210-43292210 on Build GRCh38
rs1036771852 Chr.2: 43292466-43292466 on Build GRCh38
rs772518035 Chr.2: 43292204-43292204 on Build GRCh38
rs1050820874 Chr.2: 43292251-43292251 on Build GRCh38
rs960167887 Chr.2: 43292261-43292261 on Build GRCh38
rs34600960 Chr.2: 43292156-43292156 on Build GRCh38
rs766959700 Chr.2: 43292159-43292159 on Build GRCh38
rs763137214 Chr.2: 43292126-43292126 on Build GRCh38
rs751160842 Chr.2: 43292245-43292245 on Build GRCh38
rs751216022 Chr.2: 43292171-43292171 on Build GRCh38
rs768000359 Chr.2: 43292142-43292142 on Build GRCh38
rs200266713 Chr.2: 43292128-43292128 on Build GRCh38
rs999238046 Chr.2: 43292053-43292053 on Build GRCh38
rs948356679 Chr.2: 43292339-43292339 on Build GRCh38
rs372626788 Chr.2: 43292083-43292083 on Build GRCh38
rs553102694 Chr.2: 43292288-43292288 on Build GRCh38
rs901337130 Chr.2: 43292475-43292475 on Build GRCh38
rs757003700 Chr.2: 43292172-43292172 on Build GRCh38
rs756877018 Chr.2: 43292252-43292252 on Build GRCh38
rs375630962 Chr.2: 43292205-43292205 on Build GRCh38
rs374782110 Chr.2: 43292077-43292077 on Build GRCh38
rs373291919 Chr.2: 43292239-43292239 on Build GRCh38
rs1018258126 Chr.2: 43292094-43292094 on Build GRCh38
rs965251609 Chr.2: 43292304-43292304 on Build GRCh38
rs780984068 Chr.2: 43292173-43292173 on Build GRCh38
rs575575151 Chr.2: 43292413-43292413 on Build GRCh38
rs781038918 Chr.2: 43292116-43292116 on Build GRCh38
rs377250218 Chr.2: 43292110-43292110 on Build GRCh38
rs59763196 Chr.2: 43292079-43292079 on Build GRCh38
rs371812749 Chr.2: 43292060-43292060 on Build GRCh38
rs909614420 Chr.2: 43292439-43292439 on Build GRCh38
rs995058484 Chr.2: 43292314-43292314 on Build GRCh38
rs749319540 Chr.2: 43292180-43292180 on Build GRCh38
rs184716483 Chr.2: 43292240-43292240 on Build GRCh38
rs140468535 Chr.2: 43292208-43292208 on Build GRCh38
rs946581160 Chr.2: 43292458-43292458 on Build GRCh38
rs915849537 Chr.2: 43292316-43292316 on Build GRCh38
rs745832871 Chr.2: 43292117-43292117 on Build GRCh38
rs775786571 Chr.2: 43292243-43292243 on Build GRCh38
rs545477547 Chr.2: 43292301-43292301 on Build GRCh38
rs201504112 Chr.2: 43292098-43292098 on Build GRCh38

View Full Product Details