Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 44320361-44320873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PREPL
Entrez Gene ID
9581
Gene Name
prolyl endopeptidase-like
Gene Aliases
-
UniGene
Hs.444349
Gene Chromosome Location
Chr.2: 44317607-44361862 on Build GRCh38
Transcripts
NM_001042385.2 NM_001042386.2 NM_001171603.1 NM_001171606.1 NM_001171613.1 NM_001171617.1 NM_006036.4 XM_011533198.1 XM_011533200.1 XM_011533202.1 XM_017005384.1 XM_017005385.1 NM_000341.3

Gene Symbol
SLC3A1
Entrez Gene ID
6519
Gene Name
solute carrier family 3 member 1
Gene Aliases
ATR1, CSNU1, D2H, NBAT, RBAT
UniGene
Hs.112916
Gene Chromosome Location
Chr.2: 44275458-44320824 on Build GRCh38
Transcripts
NM_001042385.2 NM_001042386.2 NM_001171603.1 NM_001171606.1 NM_001171613.1 NM_001171617.1 NM_006036.4 XM_011533198.1 XM_011533200.1 XM_011533202.1 XM_017005384.1 XM_017005385.1 NM_000341.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3185318 c.1956G>A p.T652T Chr.2: 44320537-44320537 on Build GRCh38
138298 c.1928G>A p.G643E Chr.2: 44320509-44320509 on Build GRCh38
3185316 c.1881C>T p.A627A Chr.2: 44320462-44320462 on Build GRCh38
6782214 c.1990G>A p.D664N Chr.2: 44320571-44320571 on Build GRCh38
5425577 c.1854G>A p.M618I Chr.2: 44320435-44320435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751717838 Chr.2: 44320667-44320667 on Build GRCh38
rs779104116 Chr.2: 44320625-44320625 on Build GRCh38
rs148449612 Chr.2: 44320581-44320581 on Build GRCh38
rs747267676 Chr.2: 44320635-44320635 on Build GRCh38
rs758647324 Chr.2: 44320690-44320690 on Build GRCh38
rs997845021 Chr.2: 44320754-44320754 on Build GRCh38
rs769571471 Chr.2: 44320545-44320545 on Build GRCh38
rs974590296 Chr.2: 44320764-44320764 on Build GRCh38
rs143740561 Chr.2: 44320427-44320427 on Build GRCh38
rs924248668 Chr.2: 44320619-44320619 on Build GRCh38
rs201758843 Chr.2: 44320682-44320682 on Build GRCh38
rs200953276 Chr.2: 44320683-44320683 on Build GRCh38
rs767011523 Chr.2: 44320662-44320662 on Build GRCh38
rs962167700 Chr.2: 44320583-44320583 on Build GRCh38
rs144293821 Chr.2: 44320760-44320760 on Build GRCh38
rs775393219 Chr.2: 44320461-44320461 on Build GRCh38
rs772132458 Chr.2: 44320632-44320632 on Build GRCh38
rs988923059 Chr.2: 44320814-44320814 on Build GRCh38
rs376083735 Chr.2: 44320488-44320488 on Build GRCh38
rs745773702 Chr.2: 44320452-44320452 on Build GRCh38
rs759732153 Chr.2: 44320416-44320416 on Build GRCh38
rs371136651 Chr.2: 44320594-44320594 on Build GRCh38
rs763292534 Chr.2: 44320462-44320462 on Build GRCh38
rs955228283 Chr.2: 44320513-44320513 on Build GRCh38
rs903552003 Chr.2: 44320707-44320707 on Build GRCh38
rs199685360 Chr.2: 44320421-44320421 on Build GRCh38
rs1030086903 Chr.2: 44320758-44320758 on Build GRCh38
rs770711385 Chr.2: 44320450-44320450 on Build GRCh38
rs752209571 Chr.2: 44320735-44320735 on Build GRCh38
rs756823144 Chr.2: 44320598-44320598 on Build GRCh38
rs746978183 Chr.2: 44320390-44320390 on Build GRCh38
rs1028203465 Chr.2: 44320721-44320721 on Build GRCh38
rs376270408 Chr.2: 44320603-44320603 on Build GRCh38
rs748211329 Chr.2: 44320393-44320393 on Build GRCh38
rs761541250 Chr.2: 44320661-44320661 on Build GRCh38
rs773635956 Chr.2: 44320633-44320633 on Build GRCh38
rs111875587 Chr.2: 44320746-44320746 on Build GRCh38
rs146303355 Chr.2: 44320463-44320463 on Build GRCh38
rs766252432 Chr.2: 44320678-44320678 on Build GRCh38
rs755700908 Chr.2: 44320748-44320748 on Build GRCh38
rs121912693 Chr.2: 44320614-44320614 on Build GRCh38
rs975911420 Chr.2: 44320586-44320586 on Build GRCh38
rs267599386 Chr.2: 44320502-44320502 on Build GRCh38
rs767759640 Chr.2: 44320471-44320471 on Build GRCh38
rs373698143 Chr.2: 44320563-44320563 on Build GRCh38
rs551571927 Chr.2: 44320715-44320715 on Build GRCh38
rs751254985 Chr.2: 44320595-44320595 on Build GRCh38
rs376256364 Chr.2: 44320417-44320417 on Build GRCh38
rs757852597 Chr.2: 44320426-44320426 on Build GRCh38
rs947752517 Chr.2: 44320684-44320684 on Build GRCh38
rs750128585 Chr.2: 44320608-44320608 on Build GRCh38
rs542758042 Chr.2: 44320623-44320623 on Build GRCh38
rs945139684 Chr.2: 44320835-44320835 on Build GRCh38
rs35046592 Chr.2: 44320569-44320569 on Build GRCh38
rs377448239 Chr.2: 44320572-44320572 on Build GRCh38
rs370262167 Chr.2: 44320554-44320554 on Build GRCh38
rs767168885 Chr.2: 44320577-44320577 on Build GRCh38
rs558927581 Chr.2: 44320485-44320485 on Build GRCh38
rs780297364 Chr.2: 44320552-44320552 on Build GRCh38
rs781253461 Chr.2: 44320451-44320451 on Build GRCh38
rs546219789 Chr.2: 44320795-44320795 on Build GRCh38
rs748115627 Chr.2: 44320688-44320688 on Build GRCh38
rs567296835 Chr.2: 44320792-44320792 on Build GRCh38
rs761263537 Chr.2: 44320573-44320573 on Build GRCh38
rs777564297 Chr.2: 44320443-44320443 on Build GRCh38
rs1044871710 Chr.2: 44320706-44320706 on Build GRCh38
rs761211666 Chr.2: 44320401-44320401 on Build GRCh38
rs771277513 Chr.2: 44320640-44320640 on Build GRCh38
rs973226196 Chr.2: 44320403-44320403 on Build GRCh38
rs777149216 Chr.2: 44320600-44320601 on Build GRCh38
rs750087384 Chr.2: 44320665-44320665 on Build GRCh38
rs780821343 Chr.2: 44320537-44320537 on Build GRCh38
rs938957412 Chr.2: 44320808-44320808 on Build GRCh38
rs770801300 Chr.2: 44320391-44320391 on Build GRCh38
rs749314531 Chr.2: 44320553-44320553 on Build GRCh38
rs750986488 Chr.2: 44320472-44320472 on Build GRCh38
rs751203733 Chr.2: 44320535-44320535 on Build GRCh38
rs138907837 Chr.2: 44320455-44320455 on Build GRCh38
rs121912695 Chr.2: 44320536-44320536 on Build GRCh38
rs764488098 Chr.2: 44320580-44320581 on Build GRCh38
rs750999234 Chr.2: 44320772-44320772 on Build GRCh38
rs563709928 Chr.2: 44320555-44320555 on Build GRCh38
rs149749634 Chr.2: 44320534-44320534 on Build GRCh38
rs886056073 Chr.2: 44320435-44320440 on Build GRCh38
rs8886 Chr.2: 44320770-44320770 on Build GRCh38
rs372828295 Chr.2: 44320475-44320475 on Build GRCh38
rs896210706 Chr.2: 44320491-44320491 on Build GRCh38
rs955280022 Chr.2: 44320723-44320723 on Build GRCh38
rs376897765 Chr.2: 44320618-44320618 on Build GRCh38
rs886056074 Chr.2: 44320739-44320739 on Build GRCh38
rs121912696 Chr.2: 44320424-44320424 on Build GRCh38
rs748433525 Chr.2: 44320630-44320630 on Build GRCh38
rs763629706 Chr.2: 44320645-44320645 on Build GRCh38
rs766179607 Chr.2: 44320529-44320529 on Build GRCh38
rs200817012 Chr.2: 44320396-44320396 on Build GRCh38
rs754574367 Chr.2: 44320430-44320430 on Build GRCh38
rs755921812 Chr.2: 44320673-44320673 on Build GRCh38
rs200592320 Chr.2: 44320407-44320407 on Build GRCh38
rs934090455 Chr.2: 44320410-44320410 on Build GRCh38
rs777174361 Chr.2: 44320643-44320643 on Build GRCh38
rs199949010 Chr.2: 44320560-44320560 on Build GRCh38
rs772957544 Chr.2: 44320387-44320387 on Build GRCh38
rs762511951 Chr.2: 44320593-44320593 on Build GRCh38
rs369641479 Chr.2: 44320420-44320420 on Build GRCh38
rs928863060 Chr.2: 44320798-44320798 on Build GRCh38
rs774861444 Chr.2: 44320464-44320464 on Build GRCh38
rs1009209206 Chr.2: 44320449-44320449 on Build GRCh38
rs370106397 Chr.2: 44320679-44320679 on Build GRCh38
rs756083951 Chr.2: 44320613-44320613 on Build GRCh38
rs368784606 Chr.2: 44320588-44320588 on Build GRCh38
rs550953255 Chr.2: 44320771-44320771 on Build GRCh38
rs775832166 Chr.2: 44320418-44320418 on Build GRCh38
rs745852156 Chr.2: 44320543-44320543 on Build GRCh38
rs1044014948 Chr.2: 44320445-44320445 on Build GRCh38
rs698761 Chr.2: 44320435-44320435 on Build GRCh38
rs150557210 Chr.2: 44320470-44320470 on Build GRCh38
rs761832215 Chr.2: 44320668-44320671 on Build GRCh38
rs758003140 Chr.2: 44320439-44320439 on Build GRCh38
rs913420984 Chr.2: 44320817-44320817 on Build GRCh38
rs773806097 Chr.2: 44320657-44320657 on Build GRCh38

View Full Product Details