Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55549737-55550273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R3B
Entrez Gene ID
57223
Gene Name
protein phosphatase 4 regulatory subunit 3B
Gene Aliases
FLFL2, PP4R3B, PSY2, SMEK2, smk1
UniGene
Hs.516182
Gene Chromosome Location
Chr.2: 55547292-55617757 on Build GRCh38
Transcripts
NM_001122964.2 NM_001282850.1 NM_020463.3 XM_005264442.2 XM_005264444.2 XM_005264445.2 XM_005264446.2 XM_005264447.2 XM_017004531.1 XM_017004532.1 XM_017004533.1 XM_017004534.1 XM_017004535.1 XM_017004536.1 XM_017004537.1 XM_017004538.1 XM_017004539.1 XM_017004540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1266275 c.2276G>C p.R759T Chr.2: 55549930-55549930 on Build GRCh38
6785382 c.2250A>T p.E750D Chr.2: 55549956-55549956 on Build GRCh38
1408703 c.2262G>A p.S754S Chr.2: 55549944-55549944 on Build GRCh38
6297362 c.2544C>A p.G848G Chr.2: 55549917-55549917 on Build GRCh38
1408705 c.2261C>T p.S754L Chr.2: 55549945-55549945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770947322 Chr.2: 55549979-55549979 on Build GRCh38
rs564100138 Chr.2: 55549786-55549786 on Build GRCh38
rs769458936 Chr.2: 55550006-55550006 on Build GRCh38
rs894955646 Chr.2: 55550172-55550172 on Build GRCh38
rs1003786878 Chr.2: 55549802-55549802 on Build GRCh38
rs1029297395 Chr.2: 55550205-55550205 on Build GRCh38
rs1010668592 Chr.2: 55549852-55549852 on Build GRCh38
rs750842062 Chr.2: 55549925-55549925 on Build GRCh38
rs1047764334 Chr.2: 55550076-55550076 on Build GRCh38
rs906476364 Chr.2: 55549781-55549781 on Build GRCh38
rs533075484 Chr.2: 55549844-55549844 on Build GRCh38
rs762340063 Chr.2: 55549923-55549923 on Build GRCh38
rs943558958 Chr.2: 55550024-55550024 on Build GRCh38
rs750697639 Chr.2: 55549947-55549952 on Build GRCh38
rs139911944 Chr.2: 55550070-55550070 on Build GRCh38
rs563944037 Chr.2: 55549942-55549944 on Build GRCh38
rs546472115 Chr.2: 55549862-55549862 on Build GRCh38
rs775847314 Chr.2: 55549878-55549878 on Build GRCh38
rs955022418 Chr.2: 55549992-55549992 on Build GRCh38
rs540166160 Chr.2: 55550189-55550189 on Build GRCh38
rs1003426638 Chr.2: 55550132-55550132 on Build GRCh38
rs921344379 Chr.2: 55550073-55550073 on Build GRCh38
rs753967761 Chr.2: 55550156-55550158 on Build GRCh38
rs763183036 Chr.2: 55549879-55549879 on Build GRCh38
rs1055453254 Chr.2: 55550145-55550145 on Build GRCh38
rs553701060 Chr.2: 55550202-55550202 on Build GRCh38
rs761768895 Chr.2: 55550044-55550044 on Build GRCh38
rs148775305 Chr.2: 55549924-55549924 on Build GRCh38
rs959473981 Chr.2: 55549814-55549814 on Build GRCh38
rs745737853 Chr.2: 55550002-55550002 on Build GRCh38
rs1022070189 Chr.2: 55549826-55549826 on Build GRCh38
rs935267954 Chr.2: 55550128-55550128 on Build GRCh38
rs764848400 Chr.2: 55549914-55549914 on Build GRCh38
rs780958281 Chr.2: 55549988-55549988 on Build GRCh38
rs886747254 Chr.2: 55550221-55550221 on Build GRCh38
rs374278198 Chr.2: 55549961-55549961 on Build GRCh38
rs770090922 Chr.2: 55549875-55549875 on Build GRCh38
rs377174063 Chr.2: 55550198-55550198 on Build GRCh38
rs757913068 Chr.2: 55549949-55549949 on Build GRCh38
rs750994824 Chr.2: 55549790-55549790 on Build GRCh38
rs147722277 Chr.2: 55550083-55550084 on Build GRCh38
rs1010755652 Chr.2: 55550185-55550185 on Build GRCh38
rs570921357 Chr.2: 55550118-55550118 on Build GRCh38
rs377399177 Chr.2: 55550020-55550020 on Build GRCh38
rs35375285 Chr.2: 55549859-55549860 on Build GRCh38
rs528461869 Chr.2: 55550025-55550025 on Build GRCh38
rs1052752215 Chr.2: 55550165-55550165 on Build GRCh38
rs762771133 Chr.2: 55550096-55550096 on Build GRCh38
rs181722685 Chr.2: 55550080-55550080 on Build GRCh38
rs370777934 Chr.2: 55549912-55549912 on Build GRCh38
rs756431092 Chr.2: 55549937-55549937 on Build GRCh38
rs765351948 Chr.2: 55549947-55549949 on Build GRCh38
rs915086882 Chr.2: 55550179-55550179 on Build GRCh38
rs752292325 Chr.2: 55549944-55549944 on Build GRCh38
rs549612556 Chr.2: 55550175-55550175 on Build GRCh38
rs777227527 Chr.2: 55549951-55549951 on Build GRCh38
rs759226664 Chr.2: 55549873-55549873 on Build GRCh38
rs986748908 Chr.2: 55550100-55550100 on Build GRCh38
rs761990028 Chr.2: 55549944-55549949 on Build GRCh38
rs929293316 Chr.2: 55550054-55550054 on Build GRCh38
rs551175813 Chr.2: 55550113-55550113 on Build GRCh38
rs763228900 Chr.2: 55549966-55549968 on Build GRCh38
rs906437514 Chr.2: 55550119-55550119 on Build GRCh38
rs190444596 Chr.2: 55550208-55550208 on Build GRCh38
rs1052256078 Chr.2: 55549811-55549811 on Build GRCh38
rs1017117701 Chr.2: 55550026-55550026 on Build GRCh38
rs778595417 Chr.2: 55549938-55549938 on Build GRCh38
rs1003235626 Chr.2: 55550212-55550212 on Build GRCh38
rs774522293 Chr.2: 55550030-55550030 on Build GRCh38
rs186120220 Chr.2: 55550188-55550188 on Build GRCh38
rs775867706 Chr.2: 55550010-55550010 on Build GRCh38
rs1010618344 Chr.2: 55549823-55549823 on Build GRCh38
rs746946394 Chr.2: 55549970-55549970 on Build GRCh38
rs769182277 Chr.2: 55549900-55549901 on Build GRCh38
rs151116027 Chr.2: 55550045-55550045 on Build GRCh38
rs1056162227 Chr.2: 55550213-55550213 on Build GRCh38
rs1033551079 Chr.2: 55549812-55549812 on Build GRCh38
rs752114941 Chr.2: 55549920-55549920 on Build GRCh38
rs372630317 Chr.2: 55550098-55550098 on Build GRCh38
rs760927925 Chr.2: 55550063-55550063 on Build GRCh38

View Full Product Details