Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 56184761-56185260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85A
Entrez Gene ID
114800
Gene Name
coiled-coil domain containing 85A
Gene Aliases
-
UniGene
Hs.117136
Gene Chromosome Location
Chr.2: 56184123-56386174 on Build GRCh38
Transcripts
NM_001080433.1 XM_005264125.1 NR_125368.1

Gene Symbol
LOC100129434
Entrez Gene ID
100129434
Gene Name
uncharacterized LOC100129434
Gene Aliases
-
UniGene
Hs.580330
Gene Chromosome Location
Chr.2: 56173534-56185770 on Build GRCh38
Transcripts
NM_001080433.1 XM_005264125.1 NR_125368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4591250 c.226G>T p.E76* Chr.2: 56184850-56184850 on Build GRCh38
6371098 c.144C>A p.S48R Chr.2: 56184768-56184768 on Build GRCh38
4141174 c.164G>C p.S55T Chr.2: 56184788-56184788 on Build GRCh38
3695553 c.169C>T p.R57W Chr.2: 56184793-56184793 on Build GRCh38
6347792 c.169C>A p.R57R Chr.2: 56184793-56184793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005557401 Chr.2: 56185026-56185026 on Build GRCh38
rs549095871 Chr.2: 56184990-56184990 on Build GRCh38
rs746654537 Chr.2: 56185129-56185130 on Build GRCh38
rs940082047 Chr.2: 56184790-56184790 on Build GRCh38
rs759463129 Chr.2: 56184823-56184823 on Build GRCh38
rs894491216 Chr.2: 56185239-56185239 on Build GRCh38
rs911100178 Chr.2: 56185135-56185135 on Build GRCh38
rs77267018 Chr.2: 56185193-56185193 on Build GRCh38
rs760061619 Chr.2: 56185216-56185216 on Build GRCh38
rs888441115 Chr.2: 56185010-56185010 on Build GRCh38
rs752453675 Chr.2: 56184849-56184849 on Build GRCh38
rs1025772599 Chr.2: 56184995-56184995 on Build GRCh38
rs764349835 Chr.2: 56184870-56184870 on Build GRCh38
rs917638258 Chr.2: 56184843-56184843 on Build GRCh38
rs1034974523 Chr.2: 56185029-56185029 on Build GRCh38
rs888540052 Chr.2: 56185196-56185196 on Build GRCh38
rs1049733288 Chr.2: 56185108-56185108 on Build GRCh38
rs1005938714 Chr.2: 56185200-56185200 on Build GRCh38
rs1034342601 Chr.2: 56185220-56185220 on Build GRCh38
rs1050465530 Chr.2: 56184865-56184865 on Build GRCh38
rs571467757 Chr.2: 56185124-56185124 on Build GRCh38
rs72914757 Chr.2: 56185173-56185173 on Build GRCh38
rs967559043 Chr.2: 56185093-56185093 on Build GRCh38
rs1024410117 Chr.2: 56185091-56185091 on Build GRCh38
rs565024722 Chr.2: 56184893-56184893 on Build GRCh38
rs937440880 Chr.2: 56185038-56185038 on Build GRCh38
rs923702791 Chr.2: 56185118-56185118 on Build GRCh38
rs757348777 Chr.2: 56184892-56184892 on Build GRCh38
rs920004621 Chr.2: 56185088-56185088 on Build GRCh38
rs750912243 Chr.2: 56184908-56184908 on Build GRCh38
rs569574588 Chr.2: 56185009-56185009 on Build GRCh38
rs889715163 Chr.2: 56184926-56184926 on Build GRCh38
rs959973083 Chr.2: 56184993-56184993 on Build GRCh38
rs769181223 Chr.2: 56185047-56185047 on Build GRCh38
rs11437651 Chr.2: 56185132-56185133 on Build GRCh38
rs147892797 Chr.2: 56184951-56184951 on Build GRCh38
rs542646030 Chr.2: 56185189-56185189 on Build GRCh38
rs1041004216 Chr.2: 56185172-56185172 on Build GRCh38
rs993031478 Chr.2: 56184799-56184799 on Build GRCh38
rs891375548 Chr.2: 56184880-56184880 on Build GRCh38
rs533662714 Chr.2: 56185153-56185153 on Build GRCh38
rs145093438 Chr.2: 56185128-56185129 on Build GRCh38
rs1008836508 Chr.2: 56184897-56184897 on Build GRCh38
rs959000386 Chr.2: 56185034-56185034 on Build GRCh38
rs531527213 Chr.2: 56184910-56184910 on Build GRCh38
rs375758033 Chr.2: 56184971-56184971 on Build GRCh38
rs989181269 Chr.2: 56185125-56185125 on Build GRCh38
rs538146349 Chr.2: 56185036-56185036 on Build GRCh38
rs545280108 Chr.2: 56185234-56185234 on Build GRCh38
rs1001629963 Chr.2: 56185146-56185146 on Build GRCh38
rs960250192 Chr.2: 56185147-56185147 on Build GRCh38
rs983704963 Chr.2: 56185021-56185021 on Build GRCh38
rs188314040 Chr.2: 56185198-56185198 on Build GRCh38
rs760317264 Chr.2: 56184784-56184784 on Build GRCh38
rs573638831 Chr.2: 56185177-56185177 on Build GRCh38
rs369855583 Chr.2: 56184848-56184848 on Build GRCh38
rs959091898 Chr.2: 56184785-56184785 on Build GRCh38
rs749718844 Chr.2: 56184943-56184943 on Build GRCh38
rs891400312 Chr.2: 56185115-56185118 on Build GRCh38
rs963216774 Chr.2: 56185192-56185192 on Build GRCh38
rs776440818 Chr.2: 56184807-56184807 on Build GRCh38
rs751768577 Chr.2: 56184879-56184879 on Build GRCh38
rs1011305500 Chr.2: 56184917-56184917 on Build GRCh38
rs780482219 Chr.2: 56184862-56184862 on Build GRCh38
rs993516614 Chr.2: 56185052-56185052 on Build GRCh38
rs557933866 Chr.2: 56185044-56185044 on Build GRCh38
rs765905929 Chr.2: 56184793-56184793 on Build GRCh38
rs1038354501 Chr.2: 56184801-56184801 on Build GRCh38
rs942617584 Chr.2: 56185137-56185137 on Build GRCh38
rs977805719 Chr.2: 56185104-56185104 on Build GRCh38
rs13430597 Chr.2: 56184858-56184858 on Build GRCh38
rs942503306 Chr.2: 56184935-56184935 on Build GRCh38
rs567287089 Chr.2: 56184826-56184826 on Build GRCh38
rs376801761 Chr.2: 56184918-56184918 on Build GRCh38

View Full Product Details