Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 58083749-58084271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VRK2
Entrez Gene ID
7444
Gene Name
vaccinia related kinase 2
Gene Aliases
-
UniGene
Hs.715298
Gene Chromosome Location
Chr.2: 57907651-58163991 on Build GRCh38
Transcripts
NM_001130480.2 NM_001130481.2 NM_001130482.2 NM_001130483.2 NM_001288836.1 NM_001288837.1 NM_001288838.1 NM_001288839.1 NM_006296.6 XM_005264540.4 XM_006712090.3 XM_006712091.3 XM_006712092.3 XM_006712093.3 XM_011533092.2 XM_017004858.1 XM_017004859.1 XM_017004860.1 XM_017004861.1 XM_017004862.1 XM_017004863.1 XM_017004864.1 XM_017004865.1 XM_017004866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6158316 c.137-2A>T p.? Chr.2: 58084087-58084087 on Build GRCh38
5166421 c.166G>A p.A56T Chr.2: 58084118-58084118 on Build GRCh38
3972287 c.177A>G p.V59V Chr.2: 58084129-58084129 on Build GRCh38
6467341 c.137-3C>G p.? Chr.2: 58084086-58084086 on Build GRCh38
6467342 c.137-3C>G p.? Chr.2: 58084086-58084086 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897551425 Chr.2: 58083837-58083837 on Build GRCh38
rs769177298 Chr.2: 58084039-58084039 on Build GRCh38
rs765271886 Chr.2: 58084173-58084173 on Build GRCh38
rs894448624 Chr.2: 58083912-58083912 on Build GRCh38
rs767165623 Chr.2: 58084062-58084062 on Build GRCh38
rs781297733 Chr.2: 58084130-58084130 on Build GRCh38
rs764010150 Chr.2: 58084139-58084142 on Build GRCh38
rs910199362 Chr.2: 58083785-58083785 on Build GRCh38
rs768999749 Chr.2: 58084154-58084154 on Build GRCh38
rs535808651 Chr.2: 58084137-58084137 on Build GRCh38
rs752676871 Chr.2: 58084176-58084176 on Build GRCh38
rs910448760 Chr.2: 58084032-58084032 on Build GRCh38
rs778852734 Chr.2: 58084118-58084118 on Build GRCh38
rs143894742 Chr.2: 58084234-58084234 on Build GRCh38
rs75118116 Chr.2: 58084152-58084152 on Build GRCh38
rs775129011 Chr.2: 58084073-58084073 on Build GRCh38
rs576667886 Chr.2: 58084231-58084231 on Build GRCh38
rs948534462 Chr.2: 58083929-58083929 on Build GRCh38
rs555181710 Chr.2: 58083815-58083815 on Build GRCh38
rs368145455 Chr.2: 58084204-58084204 on Build GRCh38
rs984985752 Chr.2: 58083829-58083829 on Build GRCh38
rs967296603 Chr.2: 58084191-58084191 on Build GRCh38
rs1017826682 Chr.2: 58083846-58083846 on Build GRCh38
rs200928672 Chr.2: 58084125-58084125 on Build GRCh38
rs760347517 Chr.2: 58084160-58084160 on Build GRCh38
rs778206641 Chr.2: 58084119-58084119 on Build GRCh38
rs546025315 Chr.2: 58084168-58084168 on Build GRCh38
rs945509241 Chr.2: 58083885-58083888 on Build GRCh38
rs371516428 Chr.2: 58084112-58084112 on Build GRCh38
rs757136629 Chr.2: 58084097-58084097 on Build GRCh38
rs1006210324 Chr.2: 58083902-58083902 on Build GRCh38
rs547898123 Chr.2: 58083924-58083924 on Build GRCh38
rs967399978 Chr.2: 58083940-58083940 on Build GRCh38
rs957341876 Chr.2: 58083996-58083996 on Build GRCh38
rs760164463 Chr.2: 58084072-58084072 on Build GRCh38
rs566526800 Chr.2: 58083928-58083928 on Build GRCh38
rs925365903 Chr.2: 58084232-58084232 on Build GRCh38
rs757329042 Chr.2: 58084128-58084128 on Build GRCh38
rs772916324 Chr.2: 58084069-58084069 on Build GRCh38
rs944385959 Chr.2: 58084055-58084055 on Build GRCh38
rs1045605387 Chr.2: 58084011-58084011 on Build GRCh38
rs772791503 Chr.2: 58084156-58084156 on Build GRCh38
rs192345672 Chr.2: 58083920-58083920 on Build GRCh38
rs777478501 Chr.2: 58083927-58083927 on Build GRCh38
rs944498215 Chr.2: 58083803-58083803 on Build GRCh38
rs575133454 Chr.2: 58083967-58083967 on Build GRCh38
rs202068002 Chr.2: 58084151-58084151 on Build GRCh38
rs149009548 Chr.2: 58083953-58083953 on Build GRCh38
rs964810009 Chr.2: 58083847-58083847 on Build GRCh38
rs906936507 Chr.2: 58084019-58084019 on Build GRCh38
rs543926036 Chr.2: 58084072-58084073 on Build GRCh38
rs570354982 Chr.2: 58084043-58084043 on Build GRCh38
rs770181830 Chr.2: 58084041-58084041 on Build GRCh38
rs753704819 Chr.2: 58084180-58084180 on Build GRCh38
rs759461587 Chr.2: 58083787-58083787 on Build GRCh38
rs765893477 Chr.2: 58084077-58084077 on Build GRCh38
rs373910355 Chr.2: 58084177-58084177 on Build GRCh38
rs1034383559 Chr.2: 58083907-58083907 on Build GRCh38
rs556068139 Chr.2: 58084074-58084074 on Build GRCh38
rs1056467965 Chr.2: 58084189-58084189 on Build GRCh38
rs978478029 Chr.2: 58083900-58083900 on Build GRCh38
rs369968720 Chr.2: 58084171-58084171 on Build GRCh38
rs558229220 Chr.2: 58083901-58083901 on Build GRCh38
rs374084410 Chr.2: 58084127-58084127 on Build GRCh38
rs371297361 Chr.2: 58084212-58084212 on Build GRCh38
rs148092376 Chr.2: 58084193-58084193 on Build GRCh38
rs888744051 Chr.2: 58083874-58083874 on Build GRCh38
rs930137347 Chr.2: 58083848-58083848 on Build GRCh38
rs765218773 Chr.2: 58084107-58084107 on Build GRCh38
rs375028915 Chr.2: 58084026-58084026 on Build GRCh38
rs1047223441 Chr.2: 58083858-58083858 on Build GRCh38
rs770304280 Chr.2: 58084149-58084149 on Build GRCh38
rs562375844 Chr.2: 58083831-58083831 on Build GRCh38
rs188045115 Chr.2: 58084053-58084053 on Build GRCh38
rs775485167 Chr.2: 58083990-58083990 on Build GRCh38
rs1026009948 Chr.2: 58083801-58083801 on Build GRCh38
rs555993721 Chr.2: 58084092-58084092 on Build GRCh38
rs755900386 Chr.2: 58083843-58083843 on Build GRCh38
rs371099662 Chr.2: 58084025-58084025 on Build GRCh38
rs368403988 Chr.2: 58084035-58084035 on Build GRCh38
rs925281988 Chr.2: 58083904-58083904 on Build GRCh38
rs183120932 Chr.2: 58083961-58083961 on Build GRCh38
rs34130684 Chr.2: 58084100-58084100 on Build GRCh38
rs993564269 Chr.2: 58083786-58083786 on Build GRCh38

View Full Product Details