Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61177772-61178270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
UniGene
Hs.655602
Gene Chromosome Location
Chr.2: 61177418-61191210 on Build GRCh38
Transcripts
NM_001321300.1 NM_001321301.1 NM_001321302.1 NM_001321303.1 NM_001321304.1 NM_001321305.1 NM_152392.4 XM_011532543.2 XM_017003378.1

SNP Detail

SNP ID Chromosome Location
rs1017964393 Chr.2: 61177911-61177911 on Build GRCh38
rs183498903 Chr.2: 61178000-61178000 on Build GRCh38
rs886266661 Chr.2: 61177937-61177937 on Build GRCh38
rs974824703 Chr.2: 61178044-61178044 on Build GRCh38
rs1038366729 Chr.2: 61178067-61178068 on Build GRCh38
rs563299686 Chr.2: 61178249-61178249 on Build GRCh38
rs867086416 Chr.2: 61178140-61178140 on Build GRCh38
rs750573607 Chr.2: 61178147-61178147 on Build GRCh38
rs2600667 Chr.2: 61177796-61177796 on Build GRCh38
rs190977372 Chr.2: 61177838-61177838 on Build GRCh38
rs989434029 Chr.2: 61178036-61178036 on Build GRCh38
rs1011926684 Chr.2: 61177827-61177827 on Build GRCh38
rs989090461 Chr.2: 61178195-61178195 on Build GRCh38
rs1011895369 Chr.2: 61177981-61177981 on Build GRCh38
rs770076578 Chr.2: 61177899-61177899 on Build GRCh38
rs947909130 Chr.2: 61178205-61178205 on Build GRCh38
rs940489716 Chr.2: 61178074-61178074 on Build GRCh38
rs752111829 Chr.2: 61178229-61178229 on Build GRCh38
rs34863304 Chr.2: 61178114-61178115 on Build GRCh38
rs906196923 Chr.2: 61178100-61178100 on Build GRCh38
rs765595345 Chr.2: 61178139-61178139 on Build GRCh38
rs936258693 Chr.2: 61178031-61178031 on Build GRCh38
rs552566809 Chr.2: 61178176-61178176 on Build GRCh38
rs907792664 Chr.2: 61178073-61178073 on Build GRCh38
rs780226433 Chr.2: 61178175-61178175 on Build GRCh38
rs1045053219 Chr.2: 61177807-61177807 on Build GRCh38
rs1024083126 Chr.2: 61178136-61178136 on Build GRCh38
rs1037821722 Chr.2: 61177960-61177960 on Build GRCh38
rs934185535 Chr.2: 61178217-61178217 on Build GRCh38
rs921926269 Chr.2: 61178049-61178049 on Build GRCh38
rs889850059 Chr.2: 61178062-61178062 on Build GRCh38
rs911342432 Chr.2: 61178225-61178225 on Build GRCh38
rs1021637364 Chr.2: 61178019-61178019 on Build GRCh38
rs147852965 Chr.2: 61177983-61177983 on Build GRCh38
rs540662070 Chr.2: 61178154-61178154 on Build GRCh38
rs773109691 Chr.2: 61177995-61177995 on Build GRCh38
rs767554149 Chr.2: 61177905-61177905 on Build GRCh38
rs3732175 Chr.2: 61178150-61178150 on Build GRCh38
rs942809964 Chr.2: 61178242-61178242 on Build GRCh38
rs373382199 Chr.2: 61177803-61177803 on Build GRCh38
rs934080774 Chr.2: 61177875-61177875 on Build GRCh38
rs565027001 Chr.2: 61177941-61177941 on Build GRCh38
rs573282716 Chr.2: 61178151-61178151 on Build GRCh38
rs898459315 Chr.2: 61178079-61178079 on Build GRCh38
rs1003214029 Chr.2: 61178105-61178105 on Build GRCh38
rs933224308 Chr.2: 61177926-61177926 on Build GRCh38
rs913422119 Chr.2: 61178041-61178041 on Build GRCh38
rs373862830 Chr.2: 61178085-61178085 on Build GRCh38
rs927707543 Chr.2: 61178227-61178227 on Build GRCh38
rs558038780 Chr.2: 61178080-61178080 on Build GRCh38
rs968794712 Chr.2: 61178027-61178027 on Build GRCh38
rs566211907 Chr.2: 61178005-61178005 on Build GRCh38
rs763130093 Chr.2: 61178236-61178236 on Build GRCh38
rs921830426 Chr.2: 61177916-61177916 on Build GRCh38
rs577291682 Chr.2: 61178177-61178177 on Build GRCh38
rs1023192582 Chr.2: 61177829-61177829 on Build GRCh38
rs568456314 Chr.2: 61177979-61177979 on Build GRCh38
rs914666505 Chr.2: 61177891-61177891 on Build GRCh38
rs746032 Chr.2: 61177873-61177873 on Build GRCh38
rs987501892 Chr.2: 61178169-61178169 on Build GRCh38
rs1044871501 Chr.2: 61178093-61178093 on Build GRCh38
rs986928606 Chr.2: 61178218-61178218 on Build GRCh38
rs141403407 Chr.2: 61178090-61178090 on Build GRCh38
rs961594903 Chr.2: 61177812-61177812 on Build GRCh38
rs545092304 Chr.2: 61178193-61178193 on Build GRCh38
rs892061936 Chr.2: 61178126-61178126 on Build GRCh38
rs1051153670 Chr.2: 61178055-61178055 on Build GRCh38
rs1003528664 Chr.2: 61177991-61177991 on Build GRCh38
rs1051719620 Chr.2: 61177927-61177927 on Build GRCh38
rs933361646 Chr.2: 61178058-61178058 on Build GRCh38
rs926236592 Chr.2: 61178028-61178028 on Build GRCh38
rs879546963 Chr.2: 61178059-61178059 on Build GRCh38
rs913474512 Chr.2: 61178206-61178206 on Build GRCh38
rs1036319891 Chr.2: 61178114-61178114 on Build GRCh38
rs906228009 Chr.2: 61177984-61177984 on Build GRCh38
rs143108621 Chr.2: 61177883-61177883 on Build GRCh38
rs995113955 Chr.2: 61177932-61177932 on Build GRCh38
rs970315771 Chr.2: 61177834-61177834 on Build GRCh38
rs1006879897 Chr.2: 61177900-61177900 on Build GRCh38
rs559052574 Chr.2: 61177811-61177811 on Build GRCh38
rs942737138 Chr.2: 61178064-61178064 on Build GRCh38
rs954837064 Chr.2: 61178165-61178165 on Build GRCh38
rs544844790 Chr.2: 61177847-61177847 on Build GRCh38
rs539937642 Chr.2: 61178029-61178029 on Build GRCh38
rs950489613 Chr.2: 61177947-61177947 on Build GRCh38
rs1010800434 Chr.2: 61178127-61178127 on Build GRCh38
rs1044799061 Chr.2: 61178215-61178215 on Build GRCh38
rs764889238 Chr.2: 61177825-61177826 on Build GRCh38
rs933740502 Chr.2: 61177986-61177986 on Build GRCh38

View Full Product Details