Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61192812-61193312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP34
Entrez Gene ID
9736
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
-
UniGene
Hs.644708
Gene Chromosome Location
Chr.2: 61187455-61470714 on Build GRCh38
Transcripts
NM_014709.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3962964 c.9550C>A p.L3184M Chr.2: 61192939-61192939 on Build GRCh38
6818593 c.9521C>T p.A3174V Chr.2: 61192968-61192968 on Build GRCh38
1632069 c.9524A>G p.Y3175C Chr.2: 61192965-61192965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762405901 Chr.2: 61192958-61192958 on Build GRCh38
rs376775021 Chr.2: 61193213-61193217 on Build GRCh38
rs757131591 Chr.2: 61192980-61192980 on Build GRCh38
rs11897723 Chr.2: 61193005-61193005 on Build GRCh38
rs762320995 Chr.2: 61192914-61192914 on Build GRCh38
rs908074360 Chr.2: 61193199-61193199 on Build GRCh38
rs1054481464 Chr.2: 61193236-61193236 on Build GRCh38
rs552764261 Chr.2: 61192953-61192953 on Build GRCh38
rs996983609 Chr.2: 61193256-61193256 on Build GRCh38
rs777633966 Chr.2: 61192873-61192873 on Build GRCh38
rs781524564 Chr.2: 61192878-61192878 on Build GRCh38
rs1010856524 Chr.2: 61193275-61193275 on Build GRCh38
rs948499810 Chr.2: 61193033-61193033 on Build GRCh38
rs1011460470 Chr.2: 61193136-61193136 on Build GRCh38
rs955405169 Chr.2: 61193284-61193284 on Build GRCh38
rs764058928 Chr.2: 61192965-61192965 on Build GRCh38
rs746063447 Chr.2: 61192888-61192888 on Build GRCh38
rs538535128 Chr.2: 61193081-61193081 on Build GRCh38
rs372151309 Chr.2: 61192913-61192913 on Build GRCh38
rs759113316 Chr.2: 61192945-61192945 on Build GRCh38
rs780843618 Chr.2: 61192984-61192984 on Build GRCh38
rs148101580 Chr.2: 61193131-61193131 on Build GRCh38
rs1048441154 Chr.2: 61193153-61193153 on Build GRCh38
rs1059543 Chr.2: 61193104-61193104 on Build GRCh38
rs536117453 Chr.2: 61193211-61193215 on Build GRCh38
rs928318043 Chr.2: 61193050-61193050 on Build GRCh38
rs905959638 Chr.2: 61193219-61193221 on Build GRCh38
rs575049009 Chr.2: 61193185-61193185 on Build GRCh38
rs772314047 Chr.2: 61192934-61192934 on Build GRCh38
rs920243360 Chr.2: 61193125-61193125 on Build GRCh38
rs751710053 Chr.2: 61192907-61192907 on Build GRCh38
rs865933856 Chr.2: 61192954-61192954 on Build GRCh38
rs1043906914 Chr.2: 61193204-61193204 on Build GRCh38
rs974399485 Chr.2: 61193112-61193113 on Build GRCh38
rs939570216 Chr.2: 61193110-61193110 on Build GRCh38
rs749419530 Chr.2: 61193013-61193013 on Build GRCh38
rs982698800 Chr.2: 61193000-61193000 on Build GRCh38
rs756970459 Chr.2: 61192876-61192876 on Build GRCh38
rs200281894 Chr.2: 61192890-61192890 on Build GRCh38
rs776413619 Chr.2: 61192944-61192944 on Build GRCh38
rs1030069958 Chr.2: 61193261-61193261 on Build GRCh38
rs1001594355 Chr.2: 61193220-61193220 on Build GRCh38
rs756314631 Chr.2: 61192994-61192994 on Build GRCh38
rs911641455 Chr.2: 61193083-61193083 on Build GRCh38
rs768837911 Chr.2: 61193025-61193025 on Build GRCh38
rs754701976 Chr.2: 61192844-61192844 on Build GRCh38
rs973952157 Chr.2: 61193024-61193024 on Build GRCh38
rs1045806600 Chr.2: 61193041-61193041 on Build GRCh38
rs941222785 Chr.2: 61193007-61193007 on Build GRCh38
rs530389964 Chr.2: 61192834-61192834 on Build GRCh38
rs780295717 Chr.2: 61192891-61192891 on Build GRCh38
rs1052708536 Chr.2: 61193122-61193122 on Build GRCh38
rs899878104 Chr.2: 61193237-61193237 on Build GRCh38
rs543905394 Chr.2: 61193274-61193274 on Build GRCh38
rs749333056 Chr.2: 61192910-61192910 on Build GRCh38
rs770866357 Chr.2: 61192865-61192866 on Build GRCh38
rs751475697 Chr.2: 61192978-61192978 on Build GRCh38
rs10195957 Chr.2: 61193273-61193273 on Build GRCh38
rs764882819 Chr.2: 61192946-61192946 on Build GRCh38
rs779624177 Chr.2: 61193030-61193030 on Build GRCh38

View Full Product Details