Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68180520-68181060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP3R1
Entrez Gene ID
5534
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
Gene Aliases
CALNB1, CNB, CNB1
UniGene
Hs.280604
Gene Chromosome Location
Chr.2: 68178857-68252519 on Build GRCh38
Transcripts
NM_000945.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5820939 c.499G>A p.V167M Chr.2: 68180977-68180977 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748964144 Chr.2: 68180802-68180803 on Build GRCh38
rs553786924 Chr.2: 68180643-68180643 on Build GRCh38
rs947740832 Chr.2: 68180548-68180548 on Build GRCh38
rs1004619662 Chr.2: 68180592-68180592 on Build GRCh38
rs3730336 Chr.2: 68180761-68180761 on Build GRCh38
rs371265804 Chr.2: 68180783-68180783 on Build GRCh38
rs770282462 Chr.2: 68180897-68180897 on Build GRCh38
rs191957824 Chr.2: 68180773-68180773 on Build GRCh38
rs754945144 Chr.2: 68180915-68180915 on Build GRCh38
rs1015047800 Chr.2: 68180942-68180942 on Build GRCh38
rs1056760421 Chr.2: 68180896-68180896 on Build GRCh38
rs875 Chr.2: 68180794-68180794 on Build GRCh38
rs775168479 Chr.2: 68180918-68180918 on Build GRCh38
rs1046166881 Chr.2: 68180575-68180575 on Build GRCh38
rs935195151 Chr.2: 68180788-68180788 on Build GRCh38
rs866273916 Chr.2: 68180662-68180662 on Build GRCh38
rs553214918 Chr.2: 68180907-68180907 on Build GRCh38
rs574390985 Chr.2: 68180595-68180595 on Build GRCh38
rs773512158 Chr.2: 68180688-68180692 on Build GRCh38
rs762565592 Chr.2: 68180927-68180927 on Build GRCh38
rs916529940 Chr.2: 68180888-68180888 on Build GRCh38
rs767847814 Chr.2: 68180990-68180990 on Build GRCh38
rs544783950 Chr.2: 68180597-68180597 on Build GRCh38
rs984035591 Chr.2: 68180867-68180867 on Build GRCh38
rs369993901 Chr.2: 68181026-68181026 on Build GRCh38
rs898922297 Chr.2: 68180983-68180983 on Build GRCh38
rs751824649 Chr.2: 68180939-68180939 on Build GRCh38
rs557152932 Chr.2: 68180579-68180583 on Build GRCh38
rs1029290976 Chr.2: 68180742-68180742 on Build GRCh38
rs776266611 Chr.2: 68180900-68180900 on Build GRCh38
rs1019625922 Chr.2: 68180743-68180743 on Build GRCh38
rs915350038 Chr.2: 68180802-68180802 on Build GRCh38
rs183274593 Chr.2: 68180579-68180579 on Build GRCh38
rs767424295 Chr.2: 68180942-68180943 on Build GRCh38
rs374227598 Chr.2: 68181031-68181031 on Build GRCh38
rs756321770 Chr.2: 68181021-68181021 on Build GRCh38
rs758631487 Chr.2: 68180576-68180580 on Build GRCh38
rs1035182036 Chr.2: 68180558-68180558 on Build GRCh38
rs946671137 Chr.2: 68180821-68180821 on Build GRCh38
rs764476662 Chr.2: 68180933-68180933 on Build GRCh38
rs187546083 Chr.2: 68180583-68180583 on Build GRCh38
rs981793424 Chr.2: 68180906-68180906 on Build GRCh38
rs750938363 Chr.2: 68181014-68181014 on Build GRCh38
rs866164378 Chr.2: 68181003-68181003 on Build GRCh38
rs973094528 Chr.2: 68180732-68180732 on Build GRCh38
rs1021874635 Chr.2: 68180615-68180615 on Build GRCh38
rs939563741 Chr.2: 68180864-68180864 on Build GRCh38
rs1045081449 Chr.2: 68180833-68180833 on Build GRCh38
rs965862551 Chr.2: 68180744-68180744 on Build GRCh38
rs368622412 Chr.2: 68180915-68180915 on Build GRCh38
rs905229661 Chr.2: 68180849-68180849 on Build GRCh38
rs935085030 Chr.2: 68180926-68180926 on Build GRCh38
rs972551487 Chr.2: 68180758-68180758 on Build GRCh38
rs3770643 Chr.2: 68180545-68180545 on Build GRCh38
rs954029852 Chr.2: 68180648-68180651 on Build GRCh38
rs149881293 Chr.2: 68180602-68180602 on Build GRCh38
rs891959087 Chr.2: 68181027-68181027 on Build GRCh38
rs372874240 Chr.2: 68180953-68180953 on Build GRCh38

View Full Product Details