Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68977203-68977748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKN1
Entrez Gene ID
56287
Gene Name
gastrokine 1
Gene Aliases
AMP18, BRICD1, CA11, FOV, foveolin
UniGene
Hs.69319
Gene Chromosome Location
Chr.2: 68974573-68980980 on Build GRCh38
Transcripts
NM_019617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3758316 c.57T>C p.I19I Chr.2: 68977497-68977497 on Build GRCh38
3758317 c.99A>T p.L33L Chr.2: 68977539-68977539 on Build GRCh38
270835 c.130A>G p.N44D Chr.2: 68977658-68977658 on Build GRCh38
3055916 c.209G>T p.G70V Chr.2: 68977737-68977737 on Build GRCh38
6671329 c.69A>G p.G23G Chr.2: 68977509-68977509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563455167 Chr.2: 68977476-68977476 on Build GRCh38
rs1011594710 Chr.2: 68977503-68977503 on Build GRCh38
rs35624281 Chr.2: 68977497-68977497 on Build GRCh38
rs115182400 Chr.2: 68977446-68977446 on Build GRCh38
rs774901401 Chr.2: 68977506-68977506 on Build GRCh38
rs200879154 Chr.2: 68977686-68977686 on Build GRCh38
rs751480213 Chr.2: 68977575-68977575 on Build GRCh38
rs750113317 Chr.2: 68977593-68977593 on Build GRCh38
rs753526359 Chr.2: 68977722-68977722 on Build GRCh38
rs760833410 Chr.2: 68977549-68977549 on Build GRCh38
rs553034259 Chr.2: 68977234-68977234 on Build GRCh38
rs749911001 Chr.2: 68977708-68977708 on Build GRCh38
rs761747954 Chr.2: 68977692-68977692 on Build GRCh38
rs777614890 Chr.2: 68977439-68977439 on Build GRCh38
rs752654004 Chr.2: 68977280-68977280 on Build GRCh38
rs894272199 Chr.2: 68977432-68977432 on Build GRCh38
rs749618789 Chr.2: 68977605-68977605 on Build GRCh38
rs771095645 Chr.2: 68977645-68977645 on Build GRCh38
rs748218048 Chr.2: 68977510-68977510 on Build GRCh38
rs746842496 Chr.2: 68977467-68977469 on Build GRCh38
rs147955757 Chr.2: 68977536-68977536 on Build GRCh38
rs772189819 Chr.2: 68977514-68977514 on Build GRCh38
rs921393707 Chr.2: 68977276-68977276 on Build GRCh38
rs750987557 Chr.2: 68977464-68977464 on Build GRCh38
rs989876571 Chr.2: 68977567-68977567 on Build GRCh38
rs188005906 Chr.2: 68977552-68977552 on Build GRCh38
rs9808077 Chr.2: 68977539-68977539 on Build GRCh38
rs141054391 Chr.2: 68977675-68977675 on Build GRCh38
rs1024584900 Chr.2: 68977457-68977457 on Build GRCh38
rs769002727 Chr.2: 68977504-68977504 on Build GRCh38
rs572784517 Chr.2: 68977323-68977323 on Build GRCh38
rs762694170 Chr.2: 68977649-68977649 on Build GRCh38
rs200894594 Chr.2: 68977690-68977690 on Build GRCh38
rs144238565 Chr.2: 68977515-68977515 on Build GRCh38
rs1012999733 Chr.2: 68977240-68977240 on Build GRCh38
rs924429596 Chr.2: 68977500-68977500 on Build GRCh38
rs897501059 Chr.2: 68977374-68977374 on Build GRCh38
rs940293900 Chr.2: 68977351-68977355 on Build GRCh38
rs1016766535 Chr.2: 68977395-68977395 on Build GRCh38
rs760425589 Chr.2: 68977711-68977711 on Build GRCh38
rs541105960 Chr.2: 68977331-68977331 on Build GRCh38
rs572253415 Chr.2: 68977265-68977265 on Build GRCh38
rs374143649 Chr.2: 68977554-68977554 on Build GRCh38
rs977377013 Chr.2: 68977242-68977242 on Build GRCh38
rs765025103 Chr.2: 68977550-68977550 on Build GRCh38
rs780507925 Chr.2: 68977496-68977496 on Build GRCh38
rs767139304 Chr.2: 68977697-68977697 on Build GRCh38
rs551791350 Chr.2: 68977595-68977595 on Build GRCh38
rs375676817 Chr.2: 68977597-68977597 on Build GRCh38
rs779356901 Chr.2: 68977613-68977613 on Build GRCh38
rs528465833 Chr.2: 68977425-68977425 on Build GRCh38
rs906918200 Chr.2: 68977678-68977678 on Build GRCh38
rs755340037 Chr.2: 68977611-68977611 on Build GRCh38
rs754641714 Chr.2: 68977649-68977651 on Build GRCh38
rs776814123 Chr.2: 68977646-68977646 on Build GRCh38
rs368056527 Chr.2: 68977600-68977600 on Build GRCh38
rs143197704 Chr.2: 68977364-68977364 on Build GRCh38
rs1031673408 Chr.2: 68977243-68977243 on Build GRCh38
rs957444127 Chr.2: 68977247-68977247 on Build GRCh38
rs1006228823 Chr.2: 68977335-68977335 on Build GRCh38
rs972981834 Chr.2: 68977592-68977592 on Build GRCh38
rs193227168 Chr.2: 68977324-68977324 on Build GRCh38
rs780871457 Chr.2: 68977664-68977665 on Build GRCh38
rs772058667 Chr.2: 68977629-68977629 on Build GRCh38
rs372237156 Chr.2: 68977624-68977624 on Build GRCh38
rs370812060 Chr.2: 68977468-68977468 on Build GRCh38
rs964720348 Chr.2: 68977428-68977428 on Build GRCh38
rs987402269 Chr.2: 68977294-68977294 on Build GRCh38
rs998763170 Chr.2: 68977479-68977479 on Build GRCh38
rs144357553 Chr.2: 68977499-68977499 on Build GRCh38
rs1049234885 Chr.2: 68977650-68977650 on Build GRCh38
rs546719142 Chr.2: 68977465-68977465 on Build GRCh38
rs200487427 Chr.2: 68977485-68977485 on Build GRCh38
rs747266601 Chr.2: 68977641-68977641 on Build GRCh38
rs767118741 Chr.2: 68977583-68977583 on Build GRCh38
rs185471500 Chr.2: 68977487-68977487 on Build GRCh38
rs766176560 Chr.2: 68977714-68977714 on Build GRCh38
rs990202973 Chr.2: 68977248-68977248 on Build GRCh38
rs565532872 Chr.2: 68977638-68977638 on Build GRCh38
rs756780374 Chr.2: 68977581-68977581 on Build GRCh38
rs758268905 Chr.2: 68977461-68977461 on Build GRCh38
rs867319188 Chr.2: 68977715-68977715 on Build GRCh38
rs986984110 Chr.2: 68977572-68977572 on Build GRCh38
rs955254444 Chr.2: 68977475-68977475 on Build GRCh38
rs558809429 Chr.2: 68977393-68977393 on Build GRCh38

View Full Product Details