Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69812573-69813081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA4
Entrez Gene ID
307
Gene Name
annexin A4
Gene Aliases
ANX4, HEL-S-274, P32.5, PAP-II, PIG28, PP4-X, ZAP36
UniGene
Hs.422986
Gene Chromosome Location
Chr.2: 69643805-69826477 on Build GRCh38
Transcripts
NM_001153.4 NM_001320698.1 NM_001320700.1 NM_001320702.1 XM_017003943.1 XM_017003944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3991414 c.497A>T p.N166I Chr.2: 69812672-69812672 on Build GRCh38
3056200 c.495A>C p.G165G Chr.2: 69812670-69812670 on Build GRCh38
6496693 c.530C>T p.A177V Chr.2: 69812705-69812705 on Build GRCh38
3056202 c.516C>T p.L172L Chr.2: 69812691-69812691 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141776547 Chr.2: 69812691-69812691 on Build GRCh38
rs956998188 Chr.2: 69812806-69812806 on Build GRCh38
rs561975904 Chr.2: 69812967-69812967 on Build GRCh38
rs530998882 Chr.2: 69812975-69812975 on Build GRCh38
rs760217430 Chr.2: 69812636-69812636 on Build GRCh38
rs770565360 Chr.2: 69812758-69812758 on Build GRCh38
rs768371846 Chr.2: 69812619-69812619 on Build GRCh38
rs763519273 Chr.2: 69812744-69812787 on Build GRCh38
rs376642042 Chr.2: 69812754-69812754 on Build GRCh38
rs957104904 Chr.2: 69813041-69813041 on Build GRCh38
rs979899142 Chr.2: 69812646-69812646 on Build GRCh38
rs746085005 Chr.2: 69812696-69812696 on Build GRCh38
rs13432169 Chr.2: 69813012-69813012 on Build GRCh38
rs942466160 Chr.2: 69812838-69812838 on Build GRCh38
rs1016955976 Chr.2: 69812948-69812948 on Build GRCh38
rs560371116 Chr.2: 69812899-69812899 on Build GRCh38
rs771931798 Chr.2: 69812753-69812753 on Build GRCh38
rs761640021 Chr.2: 69812625-69812625 on Build GRCh38
rs999003402 Chr.2: 69812802-69812802 on Build GRCh38
rs773114334 Chr.2: 69812615-69812615 on Build GRCh38
rs771173092 Chr.2: 69812602-69812602 on Build GRCh38
rs998843221 Chr.2: 69812981-69812981 on Build GRCh38
rs781342958 Chr.2: 69812605-69812605 on Build GRCh38
rs909984679 Chr.2: 69812946-69812946 on Build GRCh38
rs184226986 Chr.2: 69812717-69812717 on Build GRCh38
rs978400269 Chr.2: 69812819-69812819 on Build GRCh38
rs1002719632 Chr.2: 69813006-69813006 on Build GRCh38
rs929161271 Chr.2: 69812908-69812908 on Build GRCh38
rs764622740 Chr.2: 69812682-69812682 on Build GRCh38
rs41285959 Chr.2: 69812902-69812902 on Build GRCh38
rs925671154 Chr.2: 69812836-69812836 on Build GRCh38
rs991134299 Chr.2: 69812826-69812826 on Build GRCh38
rs749093252 Chr.2: 69812609-69812609 on Build GRCh38
rs372395797 Chr.2: 69812620-69812620 on Build GRCh38
rs758801174 Chr.2: 69812674-69812674 on Build GRCh38
rs540125167 Chr.2: 69812860-69812860 on Build GRCh38
rs371943367 Chr.2: 69812782-69812782 on Build GRCh38
rs545803782 Chr.2: 69812932-69812932 on Build GRCh38
rs529464630 Chr.2: 69812700-69812700 on Build GRCh38
rs754858216 Chr.2: 69812716-69812716 on Build GRCh38
rs752332365 Chr.2: 69812982-69812982 on Build GRCh38
rs371871857 Chr.2: 69812650-69812650 on Build GRCh38
rs554426923 Chr.2: 69812776-69812776 on Build GRCh38
rs745874715 Chr.2: 69812644-69812644 on Build GRCh38
rs759076218 Chr.2: 69812804-69812804 on Build GRCh38
rs753299182 Chr.2: 69812665-69812665 on Build GRCh38
rs1030228455 Chr.2: 69812787-69812787 on Build GRCh38
rs13011127 Chr.2: 69813015-69813015 on Build GRCh38
rs941311647 Chr.2: 69812955-69812955 on Build GRCh38
rs1039716045 Chr.2: 69812963-69812963 on Build GRCh38
rs748153801 Chr.2: 69812725-69812725 on Build GRCh38
rs766147001 Chr.2: 69812642-69812642 on Build GRCh38
rs575330346 Chr.2: 69812692-69812692 on Build GRCh38
rs1047753551 Chr.2: 69812877-69812877 on Build GRCh38
rs981421078 Chr.2: 69812874-69812874 on Build GRCh38
rs573251317 Chr.2: 69812795-69812795 on Build GRCh38
rs903316580 Chr.2: 69812997-69812997 on Build GRCh38
rs1012643147 Chr.2: 69813058-69813058 on Build GRCh38
rs1005556813 Chr.2: 69812720-69812720 on Build GRCh38
rs574690737 Chr.2: 69812831-69812831 on Build GRCh38
rs755593637 Chr.2: 69812632-69812632 on Build GRCh38
rs1047174699 Chr.2: 69812698-69812698 on Build GRCh38
rs765507345 Chr.2: 69812654-69812654 on Build GRCh38
rs967613474 Chr.2: 69812798-69812798 on Build GRCh38
rs191883284 Chr.2: 69812643-69812643 on Build GRCh38
rs771656774 Chr.2: 69812632-69812632 on Build GRCh38
rs915605233 Chr.2: 69812840-69812840 on Build GRCh38
rs1047548773 Chr.2: 69812910-69812910 on Build GRCh38
rs887396116 Chr.2: 69812885-69812885 on Build GRCh38

View Full Product Details