Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69818508-69819016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA4
Entrez Gene ID
307
Gene Name
annexin A4
Gene Aliases
ANX4, HEL-S-274, P32.5, PAP-II, PIG28, PP4-X, ZAP36
UniGene
Hs.422986
Gene Chromosome Location
Chr.2: 69643805-69826477 on Build GRCh38
Transcripts
NM_001153.4 NM_001320698.1 NM_001320700.1 NM_001320702.1 XM_017003943.1 XM_017003944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7334132 c.724+10T>C p.? Chr.2: 69818704-69818704 on Build GRCh38
6240558 c.647G>T p.R216M Chr.2: 69818617-69818617 on Build GRCh38
3963007 c.667G>A p.E223K Chr.2: 69818637-69818637 on Build GRCh38
477595 c.634G>A p.D212N Chr.2: 69818604-69818604 on Build GRCh38
3783436 c.632T>G p.F211C Chr.2: 69818602-69818602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184254253 Chr.2: 69818544-69818544 on Build GRCh38
rs41291183 Chr.2: 69818938-69818938 on Build GRCh38
rs78131371 Chr.2: 69818905-69818905 on Build GRCh38
rs560695293 Chr.2: 69818829-69818829 on Build GRCh38
rs755637268 Chr.2: 69818644-69818644 on Build GRCh38
rs1035863034 Chr.2: 69818791-69818791 on Build GRCh38
rs1018396129 Chr.2: 69818878-69818878 on Build GRCh38
rs781279032 Chr.2: 69818629-69818629 on Build GRCh38
rs192893273 Chr.2: 69818770-69818770 on Build GRCh38
rs908855444 Chr.2: 69818927-69818927 on Build GRCh38
rs902592723 Chr.2: 69818543-69818543 on Build GRCh38
rs925828741 Chr.2: 69818895-69818895 on Build GRCh38
rs766472468 Chr.2: 69818582-69818582 on Build GRCh38
rs1012269391 Chr.2: 69818914-69818914 on Build GRCh38
rs748956799 Chr.2: 69818560-69818560 on Build GRCh38
rs370840392 Chr.2: 69818578-69818578 on Build GRCh38
rs544213695 Chr.2: 69818817-69818817 on Build GRCh38
rs751424173 Chr.2: 69818609-69818609 on Build GRCh38
rs1022064710 Chr.2: 69818815-69818815 on Build GRCh38
rs906773781 Chr.2: 69818928-69818928 on Build GRCh38
rs759773473 Chr.2: 69818592-69818592 on Build GRCh38
rs34461855 Chr.2: 69818655-69818655 on Build GRCh38
rs779639470 Chr.2: 69818557-69818557 on Build GRCh38
rs929224008 Chr.2: 69818921-69818921 on Build GRCh38
rs776803426 Chr.2: 69818741-69818741 on Build GRCh38
rs906017756 Chr.2: 69818752-69818752 on Build GRCh38
rs536375938 Chr.2: 69818603-69818603 on Build GRCh38
rs199638836 Chr.2: 69818554-69818554 on Build GRCh38
rs112292997 Chr.2: 69818850-69818850 on Build GRCh38
rs748929436 Chr.2: 69818656-69818656 on Build GRCh38
rs76176779 Chr.2: 69818633-69818633 on Build GRCh38
rs895100252 Chr.2: 69818912-69818912 on Build GRCh38
rs765516047 Chr.2: 69818605-69818605 on Build GRCh38
rs144905340 Chr.2: 69818635-69818635 on Build GRCh38
rs116265883 Chr.2: 69818669-69818669 on Build GRCh38
rs774663657 Chr.2: 69818568-69818568 on Build GRCh38
rs546750007 Chr.2: 69818862-69818862 on Build GRCh38
rs537310538 Chr.2: 69818964-69818964 on Build GRCh38
rs772425427 Chr.2: 69818713-69818713 on Build GRCh38
rs773652255 Chr.2: 69818714-69818714 on Build GRCh38
rs762123595 Chr.2: 69818570-69818570 on Build GRCh38
rs983917972 Chr.2: 69818769-69818769 on Build GRCh38
rs754060345 Chr.2: 69818584-69818584 on Build GRCh38
rs772586601 Chr.2: 69818572-69818572 on Build GRCh38
rs1010165303 Chr.2: 69818812-69818812 on Build GRCh38
rs936750063 Chr.2: 69818900-69818900 on Build GRCh38
rs1047059199 Chr.2: 69818923-69818923 on Build GRCh38
rs769054864 Chr.2: 69818565-69818565 on Build GRCh38
rs575411357 Chr.2: 69818771-69818771 on Build GRCh38
rs760735854 Chr.2: 69818580-69818580 on Build GRCh38
rs779615707 Chr.2: 69818687-69818693 on Build GRCh38
rs369666928 Chr.2: 69818715-69818715 on Build GRCh38
rs977050300 Chr.2: 69818806-69818806 on Build GRCh38
rs1041513146 Chr.2: 69818880-69818880 on Build GRCh38
rs757061542 Chr.2: 69818610-69818610 on Build GRCh38
rs551958029 Chr.2: 69818936-69818936 on Build GRCh38
rs778485271 Chr.2: 69818703-69818703 on Build GRCh38
rs761226369 Chr.2: 69818725-69818725 on Build GRCh38
rs952862862 Chr.2: 69818969-69818969 on Build GRCh38
rs770957010 Chr.2: 69818731-69818731 on Build GRCh38
rs17403799 Chr.2: 69818933-69818933 on Build GRCh38

View Full Product Details