Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69938044-69938568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXD1
Entrez Gene ID
4084
Gene Name
MAX dimerization protein 1
Gene Aliases
BHLHC58, MAD, MAD1
UniGene
Hs.468908
Gene Chromosome Location
Chr.2: 69915041-69942945 on Build GRCh38
Transcripts
NM_001202513.1 NM_001202514.1 NM_002357.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
443138 c.571A>C p.S191R Chr.2: 69938189-69938189 on Build GRCh38
3056246 c.552C>T p.S184S Chr.2: 69938170-69938170 on Build GRCh38
121251 c.489C>T p.D163D Chr.2: 69938107-69938107 on Build GRCh38
6452122 c.553G>A p.D185N Chr.2: 69938171-69938171 on Build GRCh38
7080740 c.499G>C p.E167Q Chr.2: 69938117-69938117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748444588 Chr.2: 69938222-69938222 on Build GRCh38
rs150336449 Chr.2: 69938184-69938184 on Build GRCh38
rs376296102 Chr.2: 69938300-69938300 on Build GRCh38
rs958065528 Chr.2: 69938375-69938375 on Build GRCh38
rs999725507 Chr.2: 69938367-69938367 on Build GRCh38
rs778069948 Chr.2: 69938324-69938324 on Build GRCh38
rs570034711 Chr.2: 69938320-69938320 on Build GRCh38
rs1016510973 Chr.2: 69938139-69938139 on Build GRCh38
rs750008868 Chr.2: 69938180-69938180 on Build GRCh38
rs772363417 Chr.2: 69938108-69938108 on Build GRCh38
rs773339337 Chr.2: 69938304-69938304 on Build GRCh38
rs1024585510 Chr.2: 69938156-69938156 on Build GRCh38
rs542477699 Chr.2: 69938284-69938284 on Build GRCh38
rs774081091 Chr.2: 69938277-69938277 on Build GRCh38
rs17037259 Chr.2: 69938327-69938327 on Build GRCh38
rs200915568 Chr.2: 69938244-69938244 on Build GRCh38
rs149943253 Chr.2: 69938281-69938281 on Build GRCh38
rs113008231 Chr.2: 69938283-69938283 on Build GRCh38
rs148753961 Chr.2: 69938170-69938170 on Build GRCh38
rs762136621 Chr.2: 69938074-69938076 on Build GRCh38
rs1052035768 Chr.2: 69938352-69938352 on Build GRCh38
rs773231407 Chr.2: 69938073-69938073 on Build GRCh38
rs756109003 Chr.2: 69938518-69938518 on Build GRCh38
rs766523302 Chr.2: 69938083-69938083 on Build GRCh38
rs776858613 Chr.2: 69938232-69938232 on Build GRCh38
rs762644972 Chr.2: 69938096-69938096 on Build GRCh38
rs34760279 Chr.2: 69938312-69938313 on Build GRCh38
rs752606332 Chr.2: 69938090-69938090 on Build GRCh38
rs765830543 Chr.2: 69938292-69938292 on Build GRCh38
rs1027434149 Chr.2: 69938349-69938349 on Build GRCh38
rs566054342 Chr.2: 69938081-69938081 on Build GRCh38
rs765516764 Chr.2: 69938268-69938268 on Build GRCh38
rs932711092 Chr.2: 69938530-69938530 on Build GRCh38
rs761995312 Chr.2: 69938070-69938070 on Build GRCh38
rs201472069 Chr.2: 69938204-69938204 on Build GRCh38
rs759762880 Chr.2: 69938163-69938163 on Build GRCh38
rs779300408 Chr.2: 69938125-69938125 on Build GRCh38
rs376761809 Chr.2: 69938113-69938113 on Build GRCh38
rs1010657240 Chr.2: 69938393-69938393 on Build GRCh38
rs369156630 Chr.2: 69938167-69938167 on Build GRCh38
rs141185829 Chr.2: 69938149-69938149 on Build GRCh38
rs772174539 Chr.2: 69938071-69938071 on Build GRCh38
rs757212371 Chr.2: 69938107-69938107 on Build GRCh38
rs758742658 Chr.2: 69938313-69938313 on Build GRCh38
rs990093032 Chr.2: 69938516-69938516 on Build GRCh38
rs376954560 Chr.2: 69938187-69938187 on Build GRCh38
rs763817124 Chr.2: 69938164-69938164 on Build GRCh38
rs770011251 Chr.2: 69938265-69938266 on Build GRCh38
rs374812564 Chr.2: 69938376-69938376 on Build GRCh38
rs535119144 Chr.2: 69938105-69938105 on Build GRCh38
rs1030742421 Chr.2: 69938374-69938374 on Build GRCh38
rs894210380 Chr.2: 69938358-69938358 on Build GRCh38
rs767349181 Chr.2: 69938360-69938360 on Build GRCh38
rs947903501 Chr.2: 69938074-69938074 on Build GRCh38
rs748628861 Chr.2: 69938142-69938142 on Build GRCh38
rs776683903 Chr.2: 69938154-69938154 on Build GRCh38
rs144479868 Chr.2: 69938229-69938229 on Build GRCh38
rs1036439092 Chr.2: 69938321-69938321 on Build GRCh38
rs747143734 Chr.2: 69938248-69938248 on Build GRCh38
rs927907849 Chr.2: 69938485-69938485 on Build GRCh38
rs771201928 Chr.2: 69938151-69938151 on Build GRCh38
rs185788603 Chr.2: 69938114-69938114 on Build GRCh38
rs145908169 Chr.2: 69938124-69938124 on Build GRCh38
rs773284763 Chr.2: 69938146-69938146 on Build GRCh38
rs755660364 Chr.2: 69938183-69938183 on Build GRCh38
rs972060152 Chr.2: 69938389-69938389 on Build GRCh38
rs191421946 Chr.2: 69938257-69938257 on Build GRCh38
rs764130167 Chr.2: 69938104-69938104 on Build GRCh38
rs970797316 Chr.2: 69938408-69938408 on Build GRCh38
rs774701412 Chr.2: 69938068-69938068 on Build GRCh38
rs184062453 Chr.2: 69938455-69938455 on Build GRCh38
rs200626699 Chr.2: 69938299-69938299 on Build GRCh38
rs772514901 Chr.2: 69938143-69938143 on Build GRCh38
rs957424889 Chr.2: 69938503-69938503 on Build GRCh38
rs996402787 Chr.2: 69938344-69938344 on Build GRCh38
rs916498325 Chr.2: 69938192-69938192 on Build GRCh38
rs750345622 Chr.2: 69938482-69938482 on Build GRCh38
rs760650819 Chr.2: 69938291-69938291 on Build GRCh38
rs754613344 Chr.2: 69938203-69938203 on Build GRCh38
rs749882250 Chr.2: 69938110-69938110 on Build GRCh38
rs12997754 Chr.2: 69938269-69938269 on Build GRCh38
rs1023685412 Chr.2: 69938411-69938411 on Build GRCh38
rs765818913 Chr.2: 69938523-69938523 on Build GRCh38
rs1032332403 Chr.2: 69938431-69938431 on Build GRCh38
rs760901042 Chr.2: 69938077-69938077 on Build GRCh38
rs372477349 Chr.2: 69938179-69938179 on Build GRCh38
rs981969648 Chr.2: 69938477-69938477 on Build GRCh38

View Full Product Details