Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70958899-70959398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1B1
Entrez Gene ID
525
Gene Name
ATPase H+ transporting V1 subunit B1
Gene Aliases
ATP6B1, RTA1B, VATB, VMA2, VPP3
UniGene
Hs.64173
Gene Chromosome Location
Chr.2: 70935868-70965431 on Build GRCh38
Transcripts
NM_001692.3 XM_011532907.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4971896 c.400G>A p.D134N Chr.2: 70959050-70959050 on Build GRCh38
6608104 c.426G>T p.E142D Chr.2: 70959076-70959076 on Build GRCh38
3364810 c.385T>C p.S129P Chr.2: 70959035-70959035 on Build GRCh38
1184118 c.371G>A p.R124Q Chr.2: 70959021-70959021 on Build GRCh38
6317884 c.389G>T p.G130V Chr.2: 70959039-70959039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782305129 Chr.2: 70959134-70959134 on Build GRCh38
rs112998211 Chr.2: 70959317-70959317 on Build GRCh38
rs372842500 Chr.2: 70959072-70959072 on Build GRCh38
rs782194159 Chr.2: 70958955-70958955 on Build GRCh38
rs782199733 Chr.2: 70959036-70959036 on Build GRCh38
rs141969350 Chr.2: 70959073-70959073 on Build GRCh38
rs782613510 Chr.2: 70959108-70959108 on Build GRCh38
rs727505222 Chr.2: 70959020-70959020 on Build GRCh38
rs782389236 Chr.2: 70959124-70959124 on Build GRCh38
rs1016535730 Chr.2: 70959350-70959350 on Build GRCh38
rs368567693 Chr.2: 70959069-70959069 on Build GRCh38
rs549905293 Chr.2: 70959360-70959360 on Build GRCh38
rs782599375 Chr.2: 70958985-70958985 on Build GRCh38
rs1023023596 Chr.2: 70958991-70958991 on Build GRCh38
rs920968031 Chr.2: 70959373-70959373 on Build GRCh38
rs572294780 Chr.2: 70959058-70959058 on Build GRCh38
rs782801066 Chr.2: 70958970-70958970 on Build GRCh38
rs111764508 Chr.2: 70959170-70959170 on Build GRCh38
rs556238776 Chr.2: 70958967-70958967 on Build GRCh38
rs113754494 Chr.2: 70959284-70959284 on Build GRCh38
rs782130714 Chr.2: 70959048-70959048 on Build GRCh38
rs11886712 Chr.2: 70959243-70959243 on Build GRCh38
rs569922123 Chr.2: 70959205-70959206 on Build GRCh38
rs375885326 Chr.2: 70959188-70959188 on Build GRCh38
rs973744649 Chr.2: 70959366-70959366 on Build GRCh38
rs17006573 Chr.2: 70959252-70959252 on Build GRCh38
rs782235774 Chr.2: 70959109-70959109 on Build GRCh38
rs782229919 Chr.2: 70959269-70959269 on Build GRCh38
rs375516585 Chr.2: 70959086-70959086 on Build GRCh38
rs200038589 Chr.2: 70959012-70959012 on Build GRCh38
rs782448284 Chr.2: 70959104-70959104 on Build GRCh38
rs782769838 Chr.2: 70959082-70959082 on Build GRCh38
rs782039692 Chr.2: 70959064-70959064 on Build GRCh38
rs950943729 Chr.2: 70959316-70959316 on Build GRCh38
rs782681801 Chr.2: 70959087-70959087 on Build GRCh38
rs970107225 Chr.2: 70959251-70959251 on Build GRCh38
rs182227049 Chr.2: 70959216-70959216 on Build GRCh38
rs545157208 Chr.2: 70959006-70959006 on Build GRCh38
rs782630765 Chr.2: 70959346-70959346 on Build GRCh38
rs999163843 Chr.2: 70959140-70959140 on Build GRCh38
rs547811274 Chr.2: 70959326-70959326 on Build GRCh38
rs1053054284 Chr.2: 70959157-70959157 on Build GRCh38
rs373522105 Chr.2: 70959008-70959008 on Build GRCh38
rs782663753 Chr.2: 70959125-70959125 on Build GRCh38
rs1022971173 Chr.2: 70959248-70959248 on Build GRCh38
rs781791406 Chr.2: 70959102-70959102 on Build GRCh38
rs782032773 Chr.2: 70959063-70959063 on Build GRCh38
rs781920553 Chr.2: 70959027-70959027 on Build GRCh38
rs782224062 Chr.2: 70959005-70959005 on Build GRCh38
rs782317134 Chr.2: 70959138-70959138 on Build GRCh38
rs781975535 Chr.2: 70959038-70959038 on Build GRCh38
rs113233667 Chr.2: 70958981-70958981 on Build GRCh38
rs781880491 Chr.2: 70959085-70959085 on Build GRCh38
rs782281671 Chr.2: 70959015-70959015 on Build GRCh38
rs921020254 Chr.2: 70959091-70959091 on Build GRCh38
rs200464609 Chr.2: 70958982-70958982 on Build GRCh38
rs932440896 Chr.2: 70959122-70959122 on Build GRCh38

View Full Product Details