Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73202570-73203059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOTO
Entrez Gene ID
344022
Gene Name
notochord homeobox
Gene Aliases
-
UniGene
Hs.694384
Gene Chromosome Location
Chr.2: 73202258-73211212 on Build GRCh38
Transcripts
NM_001134462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391844 c.204G>T p.P68P Chr.2: 73202870-73202870 on Build GRCh38
6542715 c.250C>T p.L84L Chr.2: 73202916-73202916 on Build GRCh38
6528342 c.109A>C p.N37H Chr.2: 73202775-73202775 on Build GRCh38
3839970 c.20G>A p.R7Q Chr.2: 73202686-73202686 on Build GRCh38
6584223 c.118C>T p.R40C Chr.2: 73202784-73202784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987743189 Chr.2: 73202989-73202989 on Build GRCh38
rs745707352 Chr.2: 73202967-73202967 on Build GRCh38
rs1005362968 Chr.2: 73202867-73202867 on Build GRCh38
rs983573052 Chr.2: 73202624-73202624 on Build GRCh38
rs746933684 Chr.2: 73203024-73203024 on Build GRCh38
rs199642782 Chr.2: 73202748-73202748 on Build GRCh38
rs771062317 Chr.2: 73202901-73202901 on Build GRCh38
rs1033710427 Chr.2: 73202830-73202830 on Build GRCh38
rs768718226 Chr.2: 73202846-73202846 on Build GRCh38
rs930549536 Chr.2: 73202928-73202928 on Build GRCh38
rs111611158 Chr.2: 73202752-73202752 on Build GRCh38
rs1025408663 Chr.2: 73202843-73202843 on Build GRCh38
rs775245634 Chr.2: 73203005-73203005 on Build GRCh38
rs561320422 Chr.2: 73202793-73202793 on Build GRCh38
rs538761658 Chr.2: 73202659-73202659 on Build GRCh38
rs1020247373 Chr.2: 73202845-73202845 on Build GRCh38
rs1016383359 Chr.2: 73202868-73202868 on Build GRCh38
rs900709709 Chr.2: 73202808-73202808 on Build GRCh38
rs749500588 Chr.2: 73202834-73202834 on Build GRCh38
rs920401272 Chr.2: 73202886-73202886 on Build GRCh38
rs996141169 Chr.2: 73202877-73202877 on Build GRCh38
rs749049754 Chr.2: 73202888-73202888 on Build GRCh38
rs922220414 Chr.2: 73202695-73202695 on Build GRCh38
rs957660217 Chr.2: 73202832-73202832 on Build GRCh38
rs933563497 Chr.2: 73202719-73202719 on Build GRCh38
rs935053907 Chr.2: 73202743-73202743 on Build GRCh38
rs572157704 Chr.2: 73202739-73202739 on Build GRCh38
rs937856402 Chr.2: 73202688-73202688 on Build GRCh38
rs964875664 Chr.2: 73202592-73202592 on Build GRCh38
rs914954914 Chr.2: 73203030-73203030 on Build GRCh38
rs1864492 Chr.2: 73202680-73202680 on Build GRCh38
rs987604479 Chr.2: 73202721-73202721 on Build GRCh38
rs565299081 Chr.2: 73202853-73202853 on Build GRCh38
rs1047645078 Chr.2: 73202949-73202949 on Build GRCh38
rs963707124 Chr.2: 73202869-73202869 on Build GRCh38
rs532397202 Chr.2: 73202875-73202875 on Build GRCh38
rs950830920 Chr.2: 73202622-73202622 on Build GRCh38
rs959495641 Chr.2: 73202882-73202882 on Build GRCh38
rs947813412 Chr.2: 73203038-73203038 on Build GRCh38
rs543717403 Chr.2: 73202840-73202840 on Build GRCh38
rs981530876 Chr.2: 73202866-73202866 on Build GRCh38
rs924088847 Chr.2: 73202990-73202990 on Build GRCh38
rs1016660586 Chr.2: 73202626-73202626 on Build GRCh38
rs913210672 Chr.2: 73202654-73202654 on Build GRCh38
rs927821341 Chr.2: 73202681-73202681 on Build GRCh38
rs114247644 Chr.2: 73202956-73202956 on Build GRCh38
rs990682980 Chr.2: 73202885-73202885 on Build GRCh38
rs906422276 Chr.2: 73202827-73202827 on Build GRCh38
rs780197622 Chr.2: 73202820-73202820 on Build GRCh38
rs753410022 Chr.2: 73202829-73202829 on Build GRCh38
rs988784361 Chr.2: 73202649-73202649 on Build GRCh38
rs1025436852 Chr.2: 73202620-73202620 on Build GRCh38
rs1029006770 Chr.2: 73202880-73202880 on Build GRCh38
rs757762792 Chr.2: 73202684-73202684 on Build GRCh38
rs576678553 Chr.2: 73202796-73202796 on Build GRCh38
rs1040551984 Chr.2: 73203011-73203011 on Build GRCh38
rs930519837 Chr.2: 73202723-73202723 on Build GRCh38
rs762525005 Chr.2: 73203021-73203021 on Build GRCh38
rs947761374 Chr.2: 73202784-73202784 on Build GRCh38
rs1040117756 Chr.2: 73202802-73202802 on Build GRCh38
rs893451284 Chr.2: 73202782-73202782 on Build GRCh38
rs60009779 Chr.2: 73202614-73202614 on Build GRCh38
rs781531278 Chr.2: 73202713-73202713 on Build GRCh38
rs1008660242 Chr.2: 73202758-73202758 on Build GRCh38
rs148083845 Chr.2: 73202993-73202993 on Build GRCh38

View Full Product Details