Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73247680-73248190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT7
Entrez Gene ID
10574
Gene Name
chaperonin containing TCP1 subunit 7
Gene Aliases
CCTETA, CCTH, NIP7-1, TCP1ETA
UniGene
Hs.368149
Gene Chromosome Location
Chr.2: 73233431-73253022 on Build GRCh38
Transcripts
NM_001009570.2 NM_001166284.1 NM_001166285.1 NM_006429.3 NR_029402.1 NR_029403.1 XM_011532478.2 XM_011532479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3839975 c.667G>C p.A223P Chr.2: 73247810-73247810 on Build GRCh38
5862820 c.704C>T p.P235L Chr.2: 73247847-73247847 on Build GRCh38
1022766 c.732C>T p.L244L Chr.2: 73247875-73247875 on Build GRCh38
4620167 c.153A>G p.I51M Chr.2: 73247908-73247908 on Build GRCh38
4670310 c.744T>C p.A248A Chr.2: 73247887-73247887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540215961 Chr.2: 73248118-73248118 on Build GRCh38
rs776717969 Chr.2: 73247954-73247954 on Build GRCh38
rs377213070 Chr.2: 73247720-73247720 on Build GRCh38
rs750434166 Chr.2: 73247735-73247735 on Build GRCh38
rs747585718 Chr.2: 73247793-73247793 on Build GRCh38
rs1011407575 Chr.2: 73247815-73247815 on Build GRCh38
rs752751728 Chr.2: 73247721-73247721 on Build GRCh38
rs529016465 Chr.2: 73248128-73248128 on Build GRCh38
rs770104696 Chr.2: 73247945-73247945 on Build GRCh38
rs758434119 Chr.2: 73247723-73247723 on Build GRCh38
rs753701453 Chr.2: 73247747-73247747 on Build GRCh38
rs1019269841 Chr.2: 73247788-73247788 on Build GRCh38
rs2231430 Chr.2: 73248060-73248060 on Build GRCh38
rs1021519324 Chr.2: 73248149-73248149 on Build GRCh38
rs372428396 Chr.2: 73247809-73247809 on Build GRCh38
rs181929902 Chr.2: 73247754-73247754 on Build GRCh38
rs745580382 Chr.2: 73248012-73248012 on Build GRCh38
rs371752895 Chr.2: 73247715-73247715 on Build GRCh38
rs1022666946 Chr.2: 73248001-73248001 on Build GRCh38
rs111324560 Chr.2: 73247759-73247759 on Build GRCh38
rs61736516 Chr.2: 73247896-73247896 on Build GRCh38
rs539762585 Chr.2: 73247741-73247741 on Build GRCh38
rs917904686 Chr.2: 73247973-73247973 on Build GRCh38
rs775593325 Chr.2: 73247850-73247850 on Build GRCh38
rs753648198 Chr.2: 73247893-73247893 on Build GRCh38
rs776580468 Chr.2: 73248013-73248013 on Build GRCh38
rs940294866 Chr.2: 73247986-73247986 on Build GRCh38
rs904432717 Chr.2: 73248166-73248166 on Build GRCh38
rs767440185 Chr.2: 73247728-73247728 on Build GRCh38
rs545782028 Chr.2: 73248057-73248057 on Build GRCh38
rs776247614 Chr.2: 73247961-73247961 on Build GRCh38
rs746191234 Chr.2: 73247934-73247934 on Build GRCh38
rs2231428 Chr.2: 73247920-73247920 on Build GRCh38
rs561792685 Chr.2: 73248124-73248124 on Build GRCh38
rs777325378 Chr.2: 73247919-73247919 on Build GRCh38
rs778316309 Chr.2: 73247783-73247783 on Build GRCh38
rs374990103 Chr.2: 73247916-73247916 on Build GRCh38
rs920095371 Chr.2: 73248019-73248019 on Build GRCh38
rs1050344793 Chr.2: 73248036-73248036 on Build GRCh38
rs952529370 Chr.2: 73248010-73248010 on Build GRCh38
rs776920888 Chr.2: 73247833-73247833 on Build GRCh38
rs572915617 Chr.2: 73248100-73248100 on Build GRCh38
rs755174095 Chr.2: 73248139-73248140 on Build GRCh38
rs1010162884 Chr.2: 73248126-73248126 on Build GRCh38
rs1049879110 Chr.2: 73248072-73248072 on Build GRCh38
rs574150597 Chr.2: 73248138-73248138 on Build GRCh38
rs766425960 Chr.2: 73247875-73247875 on Build GRCh38
rs781684672 Chr.2: 73247931-73247931 on Build GRCh38
rs768703417 Chr.2: 73247747-73247748 on Build GRCh38
rs771357322 Chr.2: 73247924-73247924 on Build GRCh38
rs760750893 Chr.2: 73247869-73247869 on Build GRCh38
rs747646519 Chr.2: 73247778-73247778 on Build GRCh38
rs556892501 Chr.2: 73248031-73248031 on Build GRCh38
rs759895510 Chr.2: 73247836-73247836 on Build GRCh38
rs2231427 Chr.2: 73247918-73247918 on Build GRCh38
rs201769090 Chr.2: 73247908-73247908 on Build GRCh38
rs931531635 Chr.2: 73248030-73248030 on Build GRCh38
rs771535488 Chr.2: 73247781-73247781 on Build GRCh38
rs1008377953 Chr.2: 73248047-73248047 on Build GRCh38
rs369803693 Chr.2: 73247853-73247853 on Build GRCh38
rs755940674 Chr.2: 73247738-73247738 on Build GRCh38
rs764120112 Chr.2: 73247857-73247857 on Build GRCh38
rs775749940 Chr.2: 73247957-73247957 on Build GRCh38
rs2231429 Chr.2: 73248041-73248041 on Build GRCh38
rs763389501 Chr.2: 73248135-73248136 on Build GRCh38
rs2231426 Chr.2: 73247844-73247844 on Build GRCh38

View Full Product Details