Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73640970-73641475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAT8
Entrez Gene ID
9027
Gene Name
N-acetyltransferase 8 (putative)
Gene Aliases
ATase2, CCNAT, CML1, GLA, Hcml1, TSC501, TSC510
UniGene
Hs.14637
Gene Chromosome Location
Chr.2: 73640723-73642410 on Build GRCh38
Transcripts
NM_003960.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316171 c.523C>T p.L175L Chr.2: 73641106-73641106 on Build GRCh38
1409432 c.507C>T p.G169G Chr.2: 73641122-73641122 on Build GRCh38
1216654 c.496C>T p.R166W Chr.2: 73641133-73641133 on Build GRCh38
71780 c.362T>A p.M121K Chr.2: 73641267-73641267 on Build GRCh38
4914870 c.479C>A p.T160N Chr.2: 73641150-73641150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760928472 Chr.2: 73641190-73641190 on Build GRCh38
rs780589282 Chr.2: 73641122-73641122 on Build GRCh38
rs1048276711 Chr.2: 73641281-73641281 on Build GRCh38
rs900823959 Chr.2: 73641164-73641164 on Build GRCh38
rs371788953 Chr.2: 73641396-73641396 on Build GRCh38
rs556298591 Chr.2: 73641366-73641366 on Build GRCh38
rs376530047 Chr.2: 73641407-73641407 on Build GRCh38
rs780980721 Chr.2: 73641218-73641218 on Build GRCh38
rs368778053 Chr.2: 73641180-73641180 on Build GRCh38
rs376906675 Chr.2: 73641136-73641136 on Build GRCh38
rs748048542 Chr.2: 73641358-73641358 on Build GRCh38
rs779891010 Chr.2: 73641127-73641127 on Build GRCh38
rs373666252 Chr.2: 73641155-73641155 on Build GRCh38
rs769040744 Chr.2: 73641132-73641132 on Build GRCh38
rs761252433 Chr.2: 73641437-73641437 on Build GRCh38
rs35112876 Chr.2: 73640997-73640998 on Build GRCh38
rs773871949 Chr.2: 73641425-73641425 on Build GRCh38
rs72635973 Chr.2: 73641239-73641239 on Build GRCh38
rs752191497 Chr.2: 73641378-73641378 on Build GRCh38
rs774580085 Chr.2: 73641074-73641074 on Build GRCh38
rs146559874 Chr.2: 73641042-73641042 on Build GRCh38
rs1062867 Chr.2: 73641008-73641008 on Build GRCh38
rs146954600 Chr.2: 73641181-73641181 on Build GRCh38
rs1062863 Chr.2: 73641093-73641093 on Build GRCh38
rs764655777 Chr.2: 73641374-73641374 on Build GRCh38
rs13538 Chr.2: 73641201-73641201 on Build GRCh38
rs967217971 Chr.2: 73641230-73641230 on Build GRCh38
rs557262972 Chr.2: 73641435-73641435 on Build GRCh38
rs750679455 Chr.2: 73641394-73641394 on Build GRCh38
rs142218708 Chr.2: 73641278-73641278 on Build GRCh38
rs747235153 Chr.2: 73641261-73641261 on Build GRCh38
rs199809643 Chr.2: 73641370-73641370 on Build GRCh38
rs772449539 Chr.2: 73641182-73641182 on Build GRCh38
rs1062864 Chr.2: 73641072-73641072 on Build GRCh38
rs767447107 Chr.2: 73641019-73641019 on Build GRCh38
rs746702182 Chr.2: 73641100-73641100 on Build GRCh38
rs746634543 Chr.2: 73641033-73641033 on Build GRCh38
rs72635972 Chr.2: 73641411-73641411 on Build GRCh38
rs754529892 Chr.2: 73641315-73641315 on Build GRCh38
rs764421364 Chr.2: 73641070-73641070 on Build GRCh38
rs368252131 Chr.2: 73641257-73641257 on Build GRCh38
rs760707489 Chr.2: 73641141-73641141 on Build GRCh38
rs767640560 Chr.2: 73641092-73641092 on Build GRCh38
rs142272948 Chr.2: 73641450-73641450 on Build GRCh38
rs756238072 Chr.2: 73641389-73641389 on Build GRCh38
rs750070844 Chr.2: 73641171-73641171 on Build GRCh38
rs758261249 Chr.2: 73641025-73641025 on Build GRCh38
rs772240653 Chr.2: 73641135-73641135 on Build GRCh38
rs140011851 Chr.2: 73641252-73641252 on Build GRCh38
rs868408088 Chr.2: 73641288-73641288 on Build GRCh38
rs778156959 Chr.2: 73641259-73641259 on Build GRCh38
rs546259119 Chr.2: 73641235-73641235 on Build GRCh38
rs761564516 Chr.2: 73641220-73641220 on Build GRCh38
rs139574802 Chr.2: 73641026-73641026 on Build GRCh38
rs752092584 Chr.2: 73641385-73641385 on Build GRCh38
rs749748874 Chr.2: 73641123-73641123 on Build GRCh38
rs540594336 Chr.2: 73641266-73641266 on Build GRCh38
rs1062866 Chr.2: 73641024-73641024 on Build GRCh38
rs778010113 Chr.2: 73641321-73641321 on Build GRCh38
rs202210289 Chr.2: 73641222-73641222 on Build GRCh38
rs375182915 Chr.2: 73641223-73641223 on Build GRCh38
rs373428358 Chr.2: 73641040-73641040 on Build GRCh38
rs780127577 Chr.2: 73641404-73641404 on Build GRCh38
rs779326199 Chr.2: 73641251-73641251 on Build GRCh38
rs990779928 Chr.2: 73641271-73641271 on Build GRCh38
rs982536993 Chr.2: 73641083-73641083 on Build GRCh38
rs760502523 Chr.2: 73641365-73641365 on Build GRCh38
rs762993306 Chr.2: 73641199-73641199 on Build GRCh38
rs62000430 Chr.2: 73641267-73641267 on Build GRCh38
rs751751910 Chr.2: 73641011-73641011 on Build GRCh38
rs780887369 Chr.2: 73641109-73641109 on Build GRCh38
rs747431825 Chr.2: 73641325-73641325 on Build GRCh38
rs150075600 Chr.2: 73641347-73641347 on Build GRCh38
rs765888768 Chr.2: 73641233-73641233 on Build GRCh38
rs766086939 Chr.2: 73641295-73641295 on Build GRCh38
rs751488221 Chr.2: 73641206-73641206 on Build GRCh38
rs749800160 Chr.2: 73641173-73641173 on Build GRCh38
rs13424561 Chr.2: 73641319-73641319 on Build GRCh38
rs867033767 Chr.2: 73641102-73641102 on Build GRCh38
rs776337375 Chr.2: 73641439-73641439 on Build GRCh38
rs755697414 Chr.2: 73641228-73641228 on Build GRCh38
rs757075931 Chr.2: 73641163-73641163 on Build GRCh38
rs147340037 Chr.2: 73641007-73641007 on Build GRCh38
rs778016679 Chr.2: 73641411-73641412 on Build GRCh38
rs756205742 Chr.2: 73641096-73641096 on Build GRCh38
rs749134012 Chr.2: 73641320-73641320 on Build GRCh38
rs1062862 Chr.2: 73641097-73641097 on Build GRCh38
rs767222331 Chr.2: 73641275-73641275 on Build GRCh38
rs530621604 Chr.2: 73641098-73641098 on Build GRCh38
rs1062865 Chr.2: 73641058-73641058 on Build GRCh38
rs138439625 Chr.2: 73641403-73641403 on Build GRCh38
rs764369982 Chr.2: 73641009-73641009 on Build GRCh38
rs757407548 Chr.2: 73641015-73641015 on Build GRCh38
rs567763204 Chr.2: 73641381-73641381 on Build GRCh38
rs766541577 Chr.2: 73641150-73641150 on Build GRCh38
rs755172388 Chr.2: 73640994-73640995 on Build GRCh38
rs376732540 Chr.2: 73641059-73641059 on Build GRCh38
rs117579981 Chr.2: 73641353-73641353 on Build GRCh38
rs775899620 Chr.2: 73641345-73641345 on Build GRCh38
rs78630710 Chr.2: 73641272-73641272 on Build GRCh38
rs1062868 Chr.2: 73641003-73641003 on Build GRCh38
rs140607912 Chr.2: 73641361-73641361 on Build GRCh38
rs781261155 Chr.2: 73641025-73641027 on Build GRCh38
rs757723492 Chr.2: 73641386-73641386 on Build GRCh38
rs141315849 Chr.2: 73641294-73641294 on Build GRCh38
rs774633020 Chr.2: 73641133-73641133 on Build GRCh38
rs181152984 Chr.2: 73641099-73641099 on Build GRCh38
rs753172319 Chr.2: 73641317-73641317 on Build GRCh38
rs370219222 Chr.2: 73641453-73641453 on Build GRCh38
rs149442492 Chr.2: 73641380-73641380 on Build GRCh38
rs745439157 Chr.2: 73641115-73641115 on Build GRCh38
rs77056043 Chr.2: 73641158-73641158 on Build GRCh38
rs753000921 Chr.2: 73641352-73641352 on Build GRCh38
rs753845906 Chr.2: 73641020-73641020 on Build GRCh38
rs771215779 Chr.2: 73641265-73641265 on Build GRCh38
rs755644484 Chr.2: 73641172-73641172 on Build GRCh38
rs746899245 Chr.2: 73641436-73641436 on Build GRCh38
rs759184691 Chr.2: 73641373-73641373 on Build GRCh38
rs140837195 Chr.2: 73641152-73641152 on Build GRCh38
rs754804477 Chr.2: 73641023-73641023 on Build GRCh38
rs773129030 Chr.2: 73641363-73641363 on Build GRCh38
rs771164222 Chr.2: 73641193-73641193 on Build GRCh38
rs759119718 Chr.2: 73641449-73641449 on Build GRCh38
rs939795722 Chr.2: 73641305-73641305 on Build GRCh38
rs753457718 Chr.2: 73641306-73641306 on Build GRCh38
rs781618174 Chr.2: 73641346-73641346 on Build GRCh38
rs770493468 Chr.2: 73641041-73641041 on Build GRCh38
rs879117325 Chr.2: 73641342-73641342 on Build GRCh38
rs148676494 Chr.2: 73641005-73641005 on Build GRCh38
rs757668615 Chr.2: 73641326-73641326 on Build GRCh38
rs761901521 Chr.2: 73641079-73641079 on Build GRCh38
rs772201591 Chr.2: 73641413-73641413 on Build GRCh38
rs566417657 Chr.2: 73641057-73641057 on Build GRCh38
rs568372752 Chr.2: 73641166-73641166 on Build GRCh38
rs756790345 Chr.2: 73641108-73641108 on Build GRCh38
rs755794477 Chr.2: 73641119-73641119 on Build GRCh38
rs776780577 Chr.2: 73641196-73641196 on Build GRCh38
rs372166833 Chr.2: 73641277-73641277 on Build GRCh38
rs201682372 Chr.2: 73641387-73641387 on Build GRCh38
rs558590127 Chr.2: 73641183-73641183 on Build GRCh38
rs769796959 Chr.2: 73641348-73641348 on Build GRCh38
rs764754413 Chr.2: 73641316-73641316 on Build GRCh38

View Full Product Details