Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73958009-73958509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGUOK
Entrez Gene ID
1716
Gene Name
deoxyguanosine kinase
Gene Aliases
MTDPS3, dGK
UniGene
Hs.469022
Gene Chromosome Location
Chr.2: 73926826-73958961 on Build GRCh38
Transcripts
NM_001318859.1 NM_001318860.1 NM_001318861.1 NM_001318862.1 NM_001318863.1 NM_080916.2 NM_080918.2 NR_134893.1 NR_134894.1 NR_134895.1 NR_134896.1 NR_134897.1 NR_134898.1 XM_011532647.2 NR_104029.1 NR_104030.1

Gene Symbol
DGUOK-AS1
Entrez Gene ID
100874048
Gene Name
DGUOK antisense RNA 1
Gene Aliases
-
UniGene
Hs.600674
Gene Chromosome Location
Chr.2: 73957967-73981439 on Build GRCh38
Transcripts
NM_001318859.1 NM_001318860.1 NM_001318861.1 NM_001318862.1 NM_001318863.1 NM_080916.2 NM_080918.2 NR_134893.1 NR_134894.1 NR_134895.1 NR_134896.1 NR_134897.1 NR_134898.1 XM_011532647.2 NR_104029.1 NR_104030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1665435 c.770delC p.S257fs*4 Chr.2: 73958208-73958208 on Build GRCh38
4506519 c.722C>T p.A241V Chr.2: 73958160-73958160 on Build GRCh38
1725998 c.790G>A p.E264K Chr.2: 73958228-73958228 on Build GRCh38
4917671 c.738A>G p.P246P Chr.2: 73958176-73958176 on Build GRCh38
1250029 c.751G>A p.D251N Chr.2: 73958189-73958189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760070555 Chr.2: 73958194-73958195 on Build GRCh38
rs761355233 Chr.2: 73958273-73958273 on Build GRCh38
rs765735643 Chr.2: 73958097-73958097 on Build GRCh38
rs961372776 Chr.2: 73958409-73958409 on Build GRCh38
rs191311412 Chr.2: 73958128-73958128 on Build GRCh38
rs754292466 Chr.2: 73958212-73958212 on Build GRCh38
rs762338433 Chr.2: 73958256-73958257 on Build GRCh38
rs745732447 Chr.2: 73958230-73958230 on Build GRCh38
rs752134346 Chr.2: 73958142-73958142 on Build GRCh38
rs104893633 Chr.2: 73958201-73958201 on Build GRCh38
rs927532344 Chr.2: 73958290-73958290 on Build GRCh38
rs1023154905 Chr.2: 73958300-73958300 on Build GRCh38
rs939667784 Chr.2: 73958108-73958108 on Build GRCh38
rs547332715 Chr.2: 73958049-73958049 on Build GRCh38
rs200169027 Chr.2: 73958196-73958196 on Build GRCh38
rs1005359578 Chr.2: 73958372-73958372 on Build GRCh38
rs768045710 Chr.2: 73958161-73958161 on Build GRCh38
rs766784797 Chr.2: 73958294-73958294 on Build GRCh38
rs535331312 Chr.2: 73958393-73958393 on Build GRCh38
rs775121770 Chr.2: 73958134-73958134 on Build GRCh38
rs369947103 Chr.2: 73958098-73958098 on Build GRCh38
rs79925403 Chr.2: 73958116-73958116 on Build GRCh38
rs768231624 Chr.2: 73958259-73958259 on Build GRCh38
rs553365354 Chr.2: 73958400-73958400 on Build GRCh38
rs771606997 Chr.2: 73958180-73958181 on Build GRCh38
rs181203937 Chr.2: 73958462-73958462 on Build GRCh38
rs996038299 Chr.2: 73958059-73958059 on Build GRCh38
rs537118431 Chr.2: 73958171-73958171 on Build GRCh38
rs893616877 Chr.2: 73958155-73958155 on Build GRCh38
rs574993160 Chr.2: 73958402-73958402 on Build GRCh38
rs775276142 Chr.2: 73958195-73958197 on Build GRCh38
rs565869588 Chr.2: 73958077-73958077 on Build GRCh38
rs759202913 Chr.2: 73958105-73958105 on Build GRCh38
rs763441864 Chr.2: 73958136-73958136 on Build GRCh38
rs767974581 Chr.2: 73958275-73958275 on Build GRCh38
rs886037614 Chr.2: 73958204-73958205 on Build GRCh38
rs143125586 Chr.2: 73958429-73958429 on Build GRCh38
rs751032877 Chr.2: 73958480-73958480 on Build GRCh38
rs201930936 Chr.2: 73958144-73958144 on Build GRCh38
rs984786420 Chr.2: 73958381-73958381 on Build GRCh38
rs762549076 Chr.2: 73958253-73958253 on Build GRCh38
rs763706988 Chr.2: 73958199-73958200 on Build GRCh38
rs1049531836 Chr.2: 73958145-73958145 on Build GRCh38
rs769518888 Chr.2: 73958238-73958238 on Build GRCh38
rs868377068 Chr.2: 73958210-73958210 on Build GRCh38
rs537322389 Chr.2: 73958334-73958334 on Build GRCh38
rs914877481 Chr.2: 73958264-73958264 on Build GRCh38
rs370071744 Chr.2: 73958143-73958143 on Build GRCh38
rs776245624 Chr.2: 73958123-73958123 on Build GRCh38
rs996153143 Chr.2: 73958453-73958453 on Build GRCh38
rs780483507 Chr.2: 73958175-73958175 on Build GRCh38
rs886037846 Chr.2: 73958235-73958235 on Build GRCh38
rs548320231 Chr.2: 73958135-73958135 on Build GRCh38
rs375267143 Chr.2: 73958138-73958138 on Build GRCh38
rs554005472 Chr.2: 73958124-73958124 on Build GRCh38
rs370641355 Chr.2: 73958289-73958289 on Build GRCh38
rs770791760 Chr.2: 73958109-73958109 on Build GRCh38
rs376428469 Chr.2: 73958286-73958286 on Build GRCh38
rs770826218 Chr.2: 73958212-73958212 on Build GRCh38
rs938800036 Chr.2: 73958326-73958326 on Build GRCh38
rs370883181 Chr.2: 73958396-73958396 on Build GRCh38
rs749464475 Chr.2: 73958187-73958187 on Build GRCh38
rs746993196 Chr.2: 73958199-73958199 on Build GRCh38
rs947760013 Chr.2: 73958274-73958274 on Build GRCh38
rs963434745 Chr.2: 73958047-73958047 on Build GRCh38
rs764655238 Chr.2: 73958440-73958440 on Build GRCh38
rs763609930 Chr.2: 73958392-73958392 on Build GRCh38
rs750950974 Chr.2: 73958288-73958288 on Build GRCh38
rs773782732 Chr.2: 73958265-73958265 on Build GRCh38
rs754144597 Chr.2: 73958296-73958296 on Build GRCh38
rs756540646 Chr.2: 73958174-73958174 on Build GRCh38
rs989266462 Chr.2: 73958234-73958234 on Build GRCh38
rs886650083 Chr.2: 73958335-73958335 on Build GRCh38
rs932422221 Chr.2: 73958469-73958469 on Build GRCh38
rs775125640 Chr.2: 73958252-73958252 on Build GRCh38
rs111244955 Chr.2: 73958340-73958340 on Build GRCh38
rs755114361 Chr.2: 73958195-73958195 on Build GRCh38
rs976379340 Chr.2: 73958418-73958418 on Build GRCh38
rs781156960 Chr.2: 73958213-73958213 on Build GRCh38

View Full Product Details