Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74460085-74460576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INO80B-WBP1
Entrez Gene ID
100532735
Gene Name
INO80B-WBP1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.410786
Gene Chromosome Location
Chr.2: 74455023-74460891 on Build GRCh38
Transcripts
NR_037849.1 NM_012477.3

Gene Symbol
WBP1
Entrez Gene ID
23559
Gene Name
WW domain binding protein 1
Gene Aliases
WBP-1
UniGene
Hs.516114
Gene Chromosome Location
Chr.2: 74458400-74460891 on Build GRCh38
Transcripts
NR_037849.1 NM_012477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292779 c.667G>A p.G223S Chr.2: 74460538-74460538 on Build GRCh38
6039766 c.412_413insC p.Y141fs*14 Chr.2: 74460283-74460284 on Build GRCh38
2999545 c.608G>T p.C203F Chr.2: 74460479-74460479 on Build GRCh38
2999544 c.598delC p.S202fs*7 Chr.2: 74460469-74460469 on Build GRCh38
3682692 c.417C>G p.P139P Chr.2: 74460288-74460288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925480280 Chr.2: 74460135-74460135 on Build GRCh38
rs61735750 Chr.2: 74460424-74460424 on Build GRCh38
rs749060395 Chr.2: 74460416-74460416 on Build GRCh38
rs761750781 Chr.2: 74460283-74460283 on Build GRCh38
rs776590688 Chr.2: 74460282-74460283 on Build GRCh38
rs547369832 Chr.2: 74460476-74460476 on Build GRCh38
rs746408559 Chr.2: 74460505-74460505 on Build GRCh38
rs369595876 Chr.2: 74460176-74460176 on Build GRCh38
rs143463665 Chr.2: 74460508-74460508 on Build GRCh38
rs747539014 Chr.2: 74460422-74460422 on Build GRCh38
rs151202039 Chr.2: 74460523-74460523 on Build GRCh38
rs564721270 Chr.2: 74460469-74460469 on Build GRCh38
rs774553850 Chr.2: 74460482-74460482 on Build GRCh38
rs747689185 Chr.2: 74460342-74460342 on Build GRCh38
rs774043546 Chr.2: 74460338-74460338 on Build GRCh38
rs372766305 Chr.2: 74460179-74460179 on Build GRCh38
rs773779353 Chr.2: 74460348-74460348 on Build GRCh38
rs1043655203 Chr.2: 74460112-74460112 on Build GRCh38
rs752802907 Chr.2: 74460294-74460294 on Build GRCh38
rs754833382 Chr.2: 74460466-74460466 on Build GRCh38
rs1045101272 Chr.2: 74460213-74460213 on Build GRCh38
rs377611855 Chr.2: 74460553-74460553 on Build GRCh38
rs765501935 Chr.2: 74460440-74460440 on Build GRCh38
rs139399620 Chr.2: 74460474-74460474 on Build GRCh38
rs767646471 Chr.2: 74460485-74460485 on Build GRCh38
rs538409962 Chr.2: 74460284-74460284 on Build GRCh38
rs151042212 Chr.2: 74460288-74460288 on Build GRCh38
rs767794247 Chr.2: 74460211-74460211 on Build GRCh38
rs947864016 Chr.2: 74460354-74460354 on Build GRCh38
rs764267221 Chr.2: 74460442-74460442 on Build GRCh38
rs980744012 Chr.2: 74460378-74460378 on Build GRCh38
rs762339861 Chr.2: 74460210-74460210 on Build GRCh38
rs953554283 Chr.2: 74460262-74460262 on Build GRCh38
rs903794468 Chr.2: 74460118-74460118 on Build GRCh38
rs547055147 Chr.2: 74460415-74460416 on Build GRCh38
rs752467047 Chr.2: 74460269-74460269 on Build GRCh38
rs765325949 Chr.2: 74460290-74460290 on Build GRCh38
rs746492371 Chr.2: 74460473-74460473 on Build GRCh38
rs758491863 Chr.2: 74460375-74460375 on Build GRCh38
rs774654440 Chr.2: 74460538-74460560 on Build GRCh38
rs748968829 Chr.2: 74460320-74460320 on Build GRCh38
rs764757065 Chr.2: 74460205-74460205 on Build GRCh38
rs141608282 Chr.2: 74460472-74460472 on Build GRCh38
rs766762910 Chr.2: 74460357-74460357 on Build GRCh38
rs778900005 Chr.2: 74460258-74460258 on Build GRCh38
rs756065273 Chr.2: 74460412-74460412 on Build GRCh38
rs369013249 Chr.2: 74460268-74460268 on Build GRCh38
rs773064295 Chr.2: 74460423-74460423 on Build GRCh38
rs768977648 Chr.2: 74460524-74460524 on Build GRCh38
rs780041744 Chr.2: 74460414-74460414 on Build GRCh38
rs147509892 Chr.2: 74460285-74460285 on Build GRCh38
rs775680937 Chr.2: 74460475-74460475 on Build GRCh38
rs549491734 Chr.2: 74460458-74460458 on Build GRCh38
rs768295951 Chr.2: 74460237-74460244 on Build GRCh38
rs146501258 Chr.2: 74460289-74460289 on Build GRCh38
rs377511681 Chr.2: 74460359-74460359 on Build GRCh38
rs139343669 Chr.2: 74460490-74460490 on Build GRCh38
rs1016449449 Chr.2: 74460461-74460461 on Build GRCh38
rs768726887 Chr.2: 74460481-74460481 on Build GRCh38
rs574091294 Chr.2: 74460282-74460282 on Build GRCh38
rs988327403 Chr.2: 74460350-74460350 on Build GRCh38
rs61735753 Chr.2: 74460454-74460454 on Build GRCh38
rs762355291 Chr.2: 74460469-74460469 on Build GRCh38
rs534766337 Chr.2: 74460311-74460311 on Build GRCh38
rs72907206 Chr.2: 74460222-74460222 on Build GRCh38
rs773375398 Chr.2: 74460543-74460543 on Build GRCh38
rs201149235 Chr.2: 74460450-74460450 on Build GRCh38
rs759893560 Chr.2: 74460439-74460439 on Build GRCh38
rs8469 Chr.2: 74460423-74460423 on Build GRCh38
rs775180412 Chr.2: 74460193-74460193 on Build GRCh38
rs751568217 Chr.2: 74460449-74460449 on Build GRCh38
rs201448542 Chr.2: 74460310-74460310 on Build GRCh38
rs768400715 Chr.2: 74460331-74460331 on Build GRCh38
rs142859048 Chr.2: 74460287-74460287 on Build GRCh38
rs770205483 Chr.2: 74460252-74460252 on Build GRCh38
rs935277494 Chr.2: 74460380-74460380 on Build GRCh38
rs979627676 Chr.2: 74460134-74460134 on Build GRCh38
rs750381366 Chr.2: 74460409-74460409 on Build GRCh38
rs780617840 Chr.2: 74460504-74460504 on Build GRCh38
rs201308898 Chr.2: 74460420-74460420 on Build GRCh38
rs778610691 Chr.2: 74460417-74460417 on Build GRCh38
rs761018448 Chr.2: 74460353-74460353 on Build GRCh38
rs770857195 Chr.2: 74460270-74460270 on Build GRCh38
rs762549769 Chr.2: 74460196-74460196 on Build GRCh38
rs1029143614 Chr.2: 74460254-74460254 on Build GRCh38
rs376014615 Chr.2: 74460267-74460267 on Build GRCh38
rs765417916 Chr.2: 74460368-74460368 on Build GRCh38
rs113321228 Chr.2: 74460131-74460132 on Build GRCh38
rs373763194 Chr.2: 74460507-74460507 on Build GRCh38
rs144923360 Chr.2: 74460538-74460538 on Build GRCh38
rs965201058 Chr.2: 74460468-74460468 on Build GRCh38
rs757186927 Chr.2: 74460299-74460299 on Build GRCh38
rs753973126 Chr.2: 74460253-74460253 on Build GRCh38
rs756327007 Chr.2: 74460223-74460223 on Build GRCh38
rs201203980 Chr.2: 74460274-74460274 on Build GRCh38
rs779930964 Chr.2: 74460314-74460314 on Build GRCh38
rs377518393 Chr.2: 74460488-74460488 on Build GRCh38
rs925167261 Chr.2: 74460372-74460372 on Build GRCh38
rs369879489 Chr.2: 74460501-74460501 on Build GRCh38
rs764023727 Chr.2: 74460385-74460385 on Build GRCh38
rs775795287 Chr.2: 74460441-74460441 on Build GRCh38
rs765926615 Chr.2: 74460525-74460526 on Build GRCh38
rs143298451 Chr.2: 74460487-74460487 on Build GRCh38
rs143637433 Chr.2: 74460502-74460502 on Build GRCh38
rs374807274 Chr.2: 74460209-74460209 on Build GRCh38
rs144263559 Chr.2: 74460491-74460491 on Build GRCh38
rs542611467 Chr.2: 74460421-74460421 on Build GRCh38
rs752786025 Chr.2: 74460370-74460370 on Build GRCh38
rs999441535 Chr.2: 74460143-74460143 on Build GRCh38
rs772509427 Chr.2: 74460344-74460344 on Build GRCh38
rs769470262 Chr.2: 74460184-74460184 on Build GRCh38
rs768311124 Chr.2: 74460286-74460286 on Build GRCh38
rs777670693 Chr.2: 74460173-74460173 on Build GRCh38
rs199658744 Chr.2: 74460133-74460134 on Build GRCh38
rs376698954 Chr.2: 74460539-74460539 on Build GRCh38

View Full Product Details