Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79158531-79159026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG3A
Entrez Gene ID
5068
Gene Name
regenerating family member 3 alpha
Gene Aliases
HIP, HIP/PAP, INGAP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3
UniGene
Hs.567312
Gene Chromosome Location
Chr.2: 79157006-79159754 on Build GRCh38
Transcripts
NM_002580.2 NM_138937.2 NM_138938.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3583546 c.85C>T p.P29S Chr.2: 79158761-79158761 on Build GRCh38
722336 c.88C>G p.Q30E Chr.2: 79158758-79158758 on Build GRCh38
4521617 c.111G>A p.R37R Chr.2: 79158735-79158735 on Build GRCh38
722338 c.119G>T p.C40F Chr.2: 79158727-79158727 on Build GRCh38
4562947 c.94G>A p.E32K Chr.2: 79158752-79158752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879246680 Chr.2: 79158740-79158740 on Build GRCh38
rs190379872 Chr.2: 79158733-79158733 on Build GRCh38
rs2272109 Chr.2: 79158932-79158932 on Build GRCh38
rs757687452 Chr.2: 79158782-79158782 on Build GRCh38
rs558734956 Chr.2: 79158761-79158761 on Build GRCh38
rs976750083 Chr.2: 79158947-79158947 on Build GRCh38
rs556491033 Chr.2: 79158971-79158971 on Build GRCh38
rs563327404 Chr.2: 79158868-79158868 on Build GRCh38
rs757491659 Chr.2: 79158754-79158754 on Build GRCh38
rs187429978 Chr.2: 79158559-79158559 on Build GRCh38
rs574526028 Chr.2: 79158841-79158841 on Build GRCh38
rs781336129 Chr.2: 79158609-79158609 on Build GRCh38
rs949559505 Chr.2: 79158831-79158831 on Build GRCh38
rs887568017 Chr.2: 79158824-79158824 on Build GRCh38
rs983618707 Chr.2: 79158844-79158844 on Build GRCh38
rs138199548 Chr.2: 79158736-79158736 on Build GRCh38
rs968133123 Chr.2: 79158885-79158885 on Build GRCh38
rs543576857 Chr.2: 79158590-79158590 on Build GRCh38
rs780873955 Chr.2: 79158840-79158840 on Build GRCh38
rs1023666937 Chr.2: 79158884-79158884 on Build GRCh38
rs368373588 Chr.2: 79158584-79158584 on Build GRCh38
rs1014628903 Chr.2: 79158830-79158830 on Build GRCh38
rs1050651709 Chr.2: 79158997-79158997 on Build GRCh38
rs2272110 Chr.2: 79158787-79158787 on Build GRCh38
rs10169162 Chr.2: 79158651-79158651 on Build GRCh38
rs151120041 Chr.2: 79158714-79158714 on Build GRCh38
rs746226853 Chr.2: 79158810-79158810 on Build GRCh38
rs775928429 Chr.2: 79158764-79158764 on Build GRCh38
rs781494401 Chr.2: 79158756-79158756 on Build GRCh38
rs781248357 Chr.2: 79158653-79158653 on Build GRCh38
rs760956845 Chr.2: 79158624-79158624 on Build GRCh38
rs757414193 Chr.2: 79158808-79158808 on Build GRCh38
rs749713335 Chr.2: 79158813-79158813 on Build GRCh38
rs979381741 Chr.2: 79158907-79158907 on Build GRCh38
rs150805284 Chr.2: 79158650-79158650 on Build GRCh38
rs931716353 Chr.2: 79158801-79158801 on Build GRCh38
rs981286434 Chr.2: 79158678-79158678 on Build GRCh38
rs748177286 Chr.2: 79158683-79158683 on Build GRCh38
rs757161884 Chr.2: 79158607-79158607 on Build GRCh38
rs80188986 Chr.2: 79158632-79158632 on Build GRCh38
rs758481517 Chr.2: 79158794-79158794 on Build GRCh38
rs758470629 Chr.2: 79158739-79158739 on Build GRCh38
rs77686105 Chr.2: 79158661-79158661 on Build GRCh38
rs747623887 Chr.2: 79158897-79158897 on Build GRCh38
rs113959793 Chr.2: 79159004-79159004 on Build GRCh38
rs201139260 Chr.2: 79158697-79158697 on Build GRCh38
rs139201953 Chr.2: 79158730-79158730 on Build GRCh38
rs575505948 Chr.2: 79158866-79158866 on Build GRCh38
rs777098727 Chr.2: 79158711-79158711 on Build GRCh38
rs897729571 Chr.2: 79158854-79158854 on Build GRCh38
rs560963480 Chr.2: 79158560-79158560 on Build GRCh38
rs778024360 Chr.2: 79158742-79158742 on Build GRCh38
rs770292081 Chr.2: 79158716-79158716 on Build GRCh38
rs766411434 Chr.2: 79158789-79158789 on Build GRCh38
rs779485360 Chr.2: 79158786-79158786 on Build GRCh38
rs756511614 Chr.2: 79158811-79158811 on Build GRCh38
rs528759526 Chr.2: 79158999-79158999 on Build GRCh38
rs781453219 Chr.2: 79158809-79158809 on Build GRCh38
rs754816439 Chr.2: 79158807-79158808 on Build GRCh38
rs200135428 Chr.2: 79158765-79158765 on Build GRCh38
rs34534850 Chr.2: 79158931-79158932 on Build GRCh38
rs747395684 Chr.2: 79158698-79158698 on Build GRCh38
rs200236722 Chr.2: 79158647-79158647 on Build GRCh38
rs1003215097 Chr.2: 79158863-79158863 on Build GRCh38
rs764774499 Chr.2: 79158637-79158637 on Build GRCh38
rs76653007 Chr.2: 79158660-79158660 on Build GRCh38
rs747782839 Chr.2: 79158931-79158931 on Build GRCh38
rs773556279 Chr.2: 79158622-79158622 on Build GRCh38
rs577021460 Chr.2: 79158869-79158869 on Build GRCh38
rs768103588 Chr.2: 79158777-79158777 on Build GRCh38
rs112949187 Chr.2: 79158998-79158998 on Build GRCh38
rs751689325 Chr.2: 79158800-79158800 on Build GRCh38
rs201118015 Chr.2: 79158618-79158618 on Build GRCh38
rs961991836 Chr.2: 79158663-79158663 on Build GRCh38
rs202062830 Chr.2: 79158791-79158791 on Build GRCh38
rs923425076 Chr.2: 79158952-79158952 on Build GRCh38
rs778930871 Chr.2: 79158785-79158786 on Build GRCh38
rs868440270 Chr.2: 79158958-79158958 on Build GRCh38
rs752224662 Chr.2: 79158648-79158648 on Build GRCh38
rs538408283 Chr.2: 79158737-79158737 on Build GRCh38
rs879047568 Chr.2: 79158773-79158773 on Build GRCh38
rs771324295 Chr.2: 79158710-79158710 on Build GRCh38
rs772275781 Chr.2: 79158695-79158695 on Build GRCh38
rs866686686 Chr.2: 79158688-79158688 on Build GRCh38
rs552809220 Chr.2: 79158610-79158610 on Build GRCh38
rs999641000 Chr.2: 79158628-79158628 on Build GRCh38
rs554252929 Chr.2: 79158826-79158826 on Build GRCh38
rs568466219 Chr.2: 79158635-79158635 on Build GRCh38
rs879170374 Chr.2: 79158784-79158784 on Build GRCh38
rs747243184 Chr.2: 79158753-79158753 on Build GRCh38
rs779093650 Chr.2: 79158667-79158667 on Build GRCh38
rs764029952 Chr.2: 79158795-79158795 on Build GRCh38
rs747905959 Chr.2: 79158815-79158816 on Build GRCh38
rs374191223 Chr.2: 79158760-79158760 on Build GRCh38
rs370618564 Chr.2: 79158600-79158600 on Build GRCh38
rs960792179 Chr.2: 79158703-79158703 on Build GRCh38
rs762429049 Chr.2: 79158724-79158724 on Build GRCh38
rs924182506 Chr.2: 79158768-79158768 on Build GRCh38
rs542541454 Chr.2: 79159000-79159000 on Build GRCh38
rs202236751 Chr.2: 79158602-79158602 on Build GRCh38
rs774925348 Chr.2: 79158719-79158719 on Build GRCh38
rs141740162 Chr.2: 79158731-79158731 on Build GRCh38
rs370821567 Chr.2: 79158619-79158619 on Build GRCh38
rs185222126 Chr.2: 79158659-79158659 on Build GRCh38
rs750566108 Chr.2: 79158786-79158786 on Build GRCh38
rs868450097 Chr.2: 79158802-79158802 on Build GRCh38
rs781005096 Chr.2: 79158808-79158809 on Build GRCh38
rs548260649 Chr.2: 79158630-79158630 on Build GRCh38
rs773670636 Chr.2: 79158819-79158819 on Build GRCh38
rs891066152 Chr.2: 79158965-79158965 on Build GRCh38
rs183324465 Chr.2: 79158983-79158983 on Build GRCh38
rs768317231 Chr.2: 79158812-79158812 on Build GRCh38
rs189432841 Chr.2: 79158568-79158568 on Build GRCh38

View Full Product Details