Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79159653-79160151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG3A
Entrez Gene ID
5068
Gene Name
regenerating family member 3 alpha
Gene Aliases
HIP, HIP/PAP, INGAP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3
UniGene
Hs.567312
Gene Chromosome Location
Chr.2: 79157006-79159754 on Build GRCh38
Transcripts
NM_002580.2 NM_138937.2 NM_138938.2

SNP Detail

SNP ID Chromosome Location
rs542116841 Chr.2: 79159949-79159949 on Build GRCh38
rs186716101 Chr.2: 79159758-79159758 on Build GRCh38
rs889347947 Chr.2: 79160006-79160006 on Build GRCh38
rs1009497047 Chr.2: 79159810-79159810 on Build GRCh38
rs957368691 Chr.2: 79159861-79159861 on Build GRCh38
rs925859972 Chr.2: 79159815-79159815 on Build GRCh38
rs550933837 Chr.2: 79160122-79160122 on Build GRCh38
rs1024768064 Chr.2: 79159861-79159871 on Build GRCh38
rs916496035 Chr.2: 79159902-79159902 on Build GRCh38
rs948387389 Chr.2: 79159730-79159730 on Build GRCh38
rs1040935608 Chr.2: 79160026-79160026 on Build GRCh38
rs534613509 Chr.2: 79160118-79160118 on Build GRCh38
rs989377850 Chr.2: 79159863-79159863 on Build GRCh38
rs898865973 Chr.2: 79159728-79159728 on Build GRCh38
rs369594444 Chr.2: 79160028-79160028 on Build GRCh38
rs867983695 Chr.2: 79159749-79159749 on Build GRCh38
rs969699652 Chr.2: 79160114-79160114 on Build GRCh38
rs72816655 Chr.2: 79159748-79159748 on Build GRCh38
rs765501945 Chr.2: 79159870-79159870 on Build GRCh38
rs866687499 Chr.2: 79159841-79159841 on Build GRCh38
rs1050675494 Chr.2: 79159995-79159995 on Build GRCh38
rs747729316 Chr.2: 79160046-79160046 on Build GRCh38
rs7607786 Chr.2: 79159864-79159864 on Build GRCh38
rs557661938 Chr.2: 79159851-79159851 on Build GRCh38
rs1051852079 Chr.2: 79159993-79159993 on Build GRCh38
rs1033063583 Chr.2: 79160064-79160064 on Build GRCh38
rs998854933 Chr.2: 79160041-79160041 on Build GRCh38
rs1013352758 Chr.2: 79160082-79160082 on Build GRCh38
rs147161306 Chr.2: 79160084-79160084 on Build GRCh38
rs750654127 Chr.2: 79159968-79159968 on Build GRCh38
rs934604033 Chr.2: 79159820-79159820 on Build GRCh38
rs940415956 Chr.2: 79159950-79159950 on Build GRCh38
rs982336291 Chr.2: 79159923-79159923 on Build GRCh38
rs530807030 Chr.2: 79160098-79160098 on Build GRCh38
rs1019133695 Chr.2: 79159818-79159818 on Build GRCh38
rs573563405 Chr.2: 79159879-79159879 on Build GRCh38
rs1036154378 Chr.2: 79159955-79159955 on Build GRCh38
rs774959563 Chr.2: 79159965-79159965 on Build GRCh38
rs930314576 Chr.2: 79159991-79159991 on Build GRCh38
rs901021441 Chr.2: 79159943-79159943 on Build GRCh38
rs7584199 Chr.2: 79160099-79160099 on Build GRCh38
rs780820921 Chr.2: 79159901-79159901 on Build GRCh38
rs957337733 Chr.2: 79160065-79160065 on Build GRCh38
rs35743580 Chr.2: 79159973-79159974 on Build GRCh38
rs761849619 Chr.2: 79159852-79159852 on Build GRCh38
rs750966585 Chr.2: 79159739-79159739 on Build GRCh38
rs377176512 Chr.2: 79159984-79159984 on Build GRCh38
rs889236099 Chr.2: 79160022-79160022 on Build GRCh38
rs889257642 Chr.2: 79159773-79159773 on Build GRCh38
rs1009181305 Chr.2: 79160011-79160011 on Build GRCh38
rs899390375 Chr.2: 79160058-79160058 on Build GRCh38
rs969227333 Chr.2: 79159906-79159906 on Build GRCh38
rs750761301 Chr.2: 79160010-79160012 on Build GRCh38

View Full Product Details