Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79651469-79651977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA2
Entrez Gene ID
1496
Gene Name
catenin alpha 2
Gene Aliases
CAP-R, CAPR, CT114, CTNR
UniGene
Hs.167368
Gene Chromosome Location
Chr.2: 79512934-80648868 on Build GRCh38
Transcripts
NM_001164883.1 NM_001282597.2 NM_001282598.1 NM_004389.3 XM_011532555.2 XM_011532556.2 XM_017003403.1 XM_017003404.1 XM_017003405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5694217 c.196G>T p.V66F Chr.2: 79651650-79651650 on Build GRCh38
4432635 c.113C>A p.A38E Chr.2: 79651567-79651567 on Build GRCh38
1659988 c.47G>T p.S16I Chr.2: 79651603-79651603 on Build GRCh38
4481337 c.24C>T p.I8I Chr.2: 79651580-79651580 on Build GRCh38
6492114 c.127A>T p.I43F Chr.2: 79651581-79651581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368286309 Chr.2: 79651912-79651912 on Build GRCh38
rs895515807 Chr.2: 79651918-79651918 on Build GRCh38
rs554380114 Chr.2: 79651497-79651497 on Build GRCh38
rs913925603 Chr.2: 79651843-79651843 on Build GRCh38
rs774723337 Chr.2: 79651702-79651702 on Build GRCh38
rs925572513 Chr.2: 79651615-79651615 on Build GRCh38
rs190886290 Chr.2: 79651576-79651576 on Build GRCh38
rs545518666 Chr.2: 79651667-79651667 on Build GRCh38
rs1013890053 Chr.2: 79651923-79651923 on Build GRCh38
rs748065831 Chr.2: 79651554-79651554 on Build GRCh38
rs367729864 Chr.2: 79651521-79651521 on Build GRCh38
rs968479110 Chr.2: 79651585-79651585 on Build GRCh38
rs758027122 Chr.2: 79651778-79651778 on Build GRCh38
rs745955594 Chr.2: 79651690-79651690 on Build GRCh38
rs889222717 Chr.2: 79651853-79651853 on Build GRCh38
rs1016571060 Chr.2: 79651789-79651789 on Build GRCh38
rs754382528 Chr.2: 79651746-79651746 on Build GRCh38
rs780303654 Chr.2: 79651693-79651693 on Build GRCh38
rs958531617 Chr.2: 79651845-79651845 on Build GRCh38
rs749910352 Chr.2: 79651543-79651543 on Build GRCh38
rs755707809 Chr.2: 79651544-79651544 on Build GRCh38
rs779742594 Chr.2: 79651545-79651545 on Build GRCh38
rs899069205 Chr.2: 79651909-79651909 on Build GRCh38
rs1026381838 Chr.2: 79651913-79651913 on Build GRCh38
rs375583816 Chr.2: 79651605-79651605 on Build GRCh38
rs778727036 Chr.2: 79651646-79651646 on Build GRCh38
rs182341209 Chr.2: 79651704-79651704 on Build GRCh38
rs959789352 Chr.2: 79651549-79651549 on Build GRCh38
rs748923719 Chr.2: 79651528-79651528 on Build GRCh38
rs942111491 Chr.2: 79651758-79651758 on Build GRCh38
rs983442779 Chr.2: 79651891-79651891 on Build GRCh38
rs771933901 Chr.2: 79651555-79651555 on Build GRCh38
rs781265941 Chr.2: 79651556-79651556 on Build GRCh38
rs750250134 Chr.2: 79651764-79651764 on Build GRCh38
rs948289556 Chr.2: 79651580-79651580 on Build GRCh38
rs959779571 Chr.2: 79651801-79651801 on Build GRCh38
rs754805652 Chr.2: 79651546-79651546 on Build GRCh38
rs3739139 Chr.2: 79651538-79651538 on Build GRCh38
rs992562002 Chr.2: 79651802-79651802 on Build GRCh38
rs543524496 Chr.2: 79651540-79651540 on Build GRCh38
rs900632375 Chr.2: 79651771-79651771 on Build GRCh38
rs774611614 Chr.2: 79651578-79651578 on Build GRCh38
rs375701118 Chr.2: 79651691-79651691 on Build GRCh38
rs778795749 Chr.2: 79651551-79651551 on Build GRCh38
rs763467447 Chr.2: 79651534-79651534 on Build GRCh38
rs551064805 Chr.2: 79651817-79651817 on Build GRCh38
rs1037548163 Chr.2: 79651899-79651899 on Build GRCh38
rs933377024 Chr.2: 79651847-79651847 on Build GRCh38
rs567315375 Chr.2: 79651793-79651793 on Build GRCh38
rs1025086270 Chr.2: 79651803-79651803 on Build GRCh38
rs777628316 Chr.2: 79651669-79651669 on Build GRCh38
rs1051818369 Chr.2: 79651848-79651848 on Build GRCh38
rs776592033 Chr.2: 79651516-79651516 on Build GRCh38
rs530980959 Chr.2: 79651798-79651798 on Build GRCh38
rs1015915966 Chr.2: 79651501-79651501 on Build GRCh38
rs369692132 Chr.2: 79651618-79651618 on Build GRCh38
rs758295172 Chr.2: 79651668-79651668 on Build GRCh38
rs764441053 Chr.2: 79651719-79651719 on Build GRCh38
rs769005515 Chr.2: 79651701-79651701 on Build GRCh38
rs767925695 Chr.2: 79651600-79651600 on Build GRCh38
rs752438244 Chr.2: 79651649-79651649 on Build GRCh38
rs765981041 Chr.2: 79651619-79651619 on Build GRCh38
rs745947461 Chr.2: 79651564-79651564 on Build GRCh38
rs368717286 Chr.2: 79651565-79651565 on Build GRCh38
rs907987384 Chr.2: 79651748-79651748 on Build GRCh38
rs751183197 Chr.2: 79651917-79651917 on Build GRCh38
rs943143036 Chr.2: 79651877-79651877 on Build GRCh38
rs186696084 Chr.2: 79651849-79651849 on Build GRCh38
rs78677982 Chr.2: 79651796-79651796 on Build GRCh38
rs1022647454 Chr.2: 79651941-79651941 on Build GRCh38
rs762317505 Chr.2: 79651596-79651596 on Build GRCh38
rs899084275 Chr.2: 79651794-79651794 on Build GRCh38
rs372730685 Chr.2: 79651569-79651569 on Build GRCh38
rs754644562 Chr.2: 79651644-79651644 on Build GRCh38
rs546708218 Chr.2: 79651939-79651939 on Build GRCh38
rs753533514 Chr.2: 79651637-79651637 on Build GRCh38
rs980983146 Chr.2: 79651815-79651815 on Build GRCh38
rs982901451 Chr.2: 79651547-79651547 on Build GRCh38
rs888978845 Chr.2: 79651761-79651761 on Build GRCh38
rs936948761 Chr.2: 79651820-79651820 on Build GRCh38
rs1047717261 Chr.2: 79651747-79651747 on Build GRCh38
rs765905334 Chr.2: 79651809-79651809 on Build GRCh38
rs765560901 Chr.2: 79651518-79651518 on Build GRCh38
rs548782428 Chr.2: 79651772-79651772 on Build GRCh38
rs764666959 Chr.2: 79651535-79651535 on Build GRCh38
rs769095342 Chr.2: 79651577-79651577 on Build GRCh38
rs528822540 Chr.2: 79651751-79651751 on Build GRCh38
rs988543077 Chr.2: 79651822-79651822 on Build GRCh38
rs917202595 Chr.2: 79651865-79651865 on Build GRCh38
rs1034598448 Chr.2: 79651842-79651842 on Build GRCh38

View Full Product Details