Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85342552-85343049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RETSAT
Entrez Gene ID
54884
Gene Name
retinol saturase
Gene Aliases
-
UniGene
Hs.440401
Gene Chromosome Location
Chr.2: 85341955-85354698 on Build GRCh38
Transcripts
NM_017750.3

SNP Detail

SNP ID Chromosome Location
rs60340620 Chr.2: 85342829-85342829 on Build GRCh38
rs566423195 Chr.2: 85342965-85342965 on Build GRCh38
rs963166661 Chr.2: 85342638-85342638 on Build GRCh38
rs186546447 Chr.2: 85342920-85342920 on Build GRCh38
rs201945906 Chr.2: 85342817-85342818 on Build GRCh38
rs183409558 Chr.2: 85342824-85342824 on Build GRCh38
rs1042221307 Chr.2: 85342961-85342961 on Build GRCh38
rs139022129 Chr.2: 85342761-85342761 on Build GRCh38
rs765494028 Chr.2: 85342654-85342654 on Build GRCh38
rs570200494 Chr.2: 85342629-85342629 on Build GRCh38
rs1017229152 Chr.2: 85342599-85342599 on Build GRCh38
rs71392941 Chr.2: 85343023-85343036 on Build GRCh38
rs1055412809 Chr.2: 85342830-85342830 on Build GRCh38
rs868863136 Chr.2: 85342890-85342890 on Build GRCh38
rs4832167 Chr.2: 85343023-85343023 on Build GRCh38
rs191374780 Chr.2: 85343013-85343013 on Build GRCh38
rs57986502 Chr.2: 85342901-85342901 on Build GRCh38
rs537576837 Chr.2: 85342643-85342643 on Build GRCh38
rs981549838 Chr.2: 85342686-85342686 on Build GRCh38
rs558174861 Chr.2: 85342582-85342582 on Build GRCh38
rs142113554 Chr.2: 85342576-85342576 on Build GRCh38
rs758440028 Chr.2: 85342966-85342966 on Build GRCh38
rs386648089 Chr.2: 85342819-85342821 on Build GRCh38
rs530015857 Chr.2: 85343006-85343006 on Build GRCh38
rs953742747 Chr.2: 85342574-85342574 on Build GRCh38
rs113820952 Chr.2: 85342840-85342840 on Build GRCh38
rs574955498 Chr.2: 85342760-85342760 on Build GRCh38
rs918649355 Chr.2: 85342668-85342668 on Build GRCh38
rs9808566 Chr.2: 85342754-85342754 on Build GRCh38
rs574914202 Chr.2: 85342601-85342601 on Build GRCh38
rs572423656 Chr.2: 85342819-85342819 on Build GRCh38
rs58145899 Chr.2: 85342894-85342894 on Build GRCh38
rs529866159 Chr.2: 85342881-85342881 on Build GRCh38
rs145057327 Chr.2: 85342802-85342802 on Build GRCh38
rs866756386 Chr.2: 85342822-85342822 on Build GRCh38
rs59202292 Chr.2: 85342872-85342872 on Build GRCh38
rs950093892 Chr.2: 85342669-85342669 on Build GRCh38
rs531818086 Chr.2: 85342839-85342839 on Build GRCh38
rs192199548 Chr.2: 85342821-85342821 on Build GRCh38

View Full Product Details