Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85545201-85545705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGCX
Entrez Gene ID
2677
Gene Name
gamma-glutamyl carboxylase
Gene Aliases
VKCFD1
UniGene
Hs.77719
Gene Chromosome Location
Chr.2: 85544720-85561534 on Build GRCh38
Transcripts
NM_000821.6 NM_001142269.3 NM_005911.5

Gene Symbol
MAT2A
Entrez Gene ID
4144
Gene Name
methionine adenosyltransferase 2A
Gene Aliases
MATA2, MATII, SAMS2
UniGene
Hs.516157
Gene Chromosome Location
Chr.2: 85538978-85545280 on Build GRCh38
Transcripts
NM_000821.6 NM_001142269.3 NM_005911.5

SNP Detail

SNP ID Chromosome Location
rs918448194 Chr.2: 85545467-85545468 on Build GRCh38
rs781163813 Chr.2: 85545660-85545660 on Build GRCh38
rs924291709 Chr.2: 85545522-85545522 on Build GRCh38
rs962640563 Chr.2: 85545435-85545436 on Build GRCh38
rs971251325 Chr.2: 85545467-85545467 on Build GRCh38
rs766703148 Chr.2: 85545539-85545539 on Build GRCh38
rs903129496 Chr.2: 85545327-85545327 on Build GRCh38
rs764634746 Chr.2: 85545335-85545335 on Build GRCh38
rs897823595 Chr.2: 85545643-85545643 on Build GRCh38
rs936970381 Chr.2: 85545311-85545311 on Build GRCh38
rs550873302 Chr.2: 85545568-85545568 on Build GRCh38
rs1015577736 Chr.2: 85545394-85545394 on Build GRCh38
rs143021753 Chr.2: 85545345-85545345 on Build GRCh38
rs949739286 Chr.2: 85545315-85545315 on Build GRCh38
rs997659203 Chr.2: 85545606-85545606 on Build GRCh38
rs962060806 Chr.2: 85545674-85545674 on Build GRCh38
rs772325888 Chr.2: 85545282-85545283 on Build GRCh38
rs532268389 Chr.2: 85545555-85545555 on Build GRCh38
rs7605975 Chr.2: 85545425-85545425 on Build GRCh38
rs949866556 Chr.2: 85545253-85545253 on Build GRCh38
rs937050843 Chr.2: 85545290-85545290 on Build GRCh38
rs138686161 Chr.2: 85545355-85545355 on Build GRCh38
rs953366242 Chr.2: 85545416-85545416 on Build GRCh38
rs768093599 Chr.2: 85545300-85545300 on Build GRCh38
rs1054028624 Chr.2: 85545535-85545535 on Build GRCh38
rs1054274273 Chr.2: 85545344-85545344 on Build GRCh38
rs566495617 Chr.2: 85545317-85545317 on Build GRCh38
rs958894585 Chr.2: 85545478-85545478 on Build GRCh38
rs916911117 Chr.2: 85545309-85545309 on Build GRCh38
rs899666330 Chr.2: 85545590-85545590 on Build GRCh38
rs1006291411 Chr.2: 85545652-85545652 on Build GRCh38
rs554891563 Chr.2: 85545604-85545606 on Build GRCh38
rs938691537 Chr.2: 85545549-85545549 on Build GRCh38
rs564820085 Chr.2: 85545537-85545537 on Build GRCh38
rs962290668 Chr.2: 85545409-85545409 on Build GRCh38
rs991234828 Chr.2: 85545487-85545487 on Build GRCh38
rs1045475871 Chr.2: 85545283-85545283 on Build GRCh38
rs554866641 Chr.2: 85545245-85545245 on Build GRCh38
rs558737437 Chr.2: 85545333-85545333 on Build GRCh38
rs530247310 Chr.2: 85545620-85545620 on Build GRCh38
rs569081895 Chr.2: 85545611-85545611 on Build GRCh38
rs764686685 Chr.2: 85545386-85545386 on Build GRCh38
rs749940639 Chr.2: 85545406-85545406 on Build GRCh38
rs1014901797 Chr.2: 85545632-85545632 on Build GRCh38
rs889180410 Chr.2: 85545631-85545631 on Build GRCh38
rs890567280 Chr.2: 85545613-85545613 on Build GRCh38
rs1027782773 Chr.2: 85545445-85545445 on Build GRCh38
rs1028362012 Chr.2: 85545679-85545679 on Build GRCh38
rs771861163 Chr.2: 85545428-85545428 on Build GRCh38
rs561174795 Chr.2: 85545441-85545441 on Build GRCh38
rs1054790916 Chr.2: 85545559-85545559 on Build GRCh38
rs890451611 Chr.2: 85545375-85545375 on Build GRCh38
rs534210883 Chr.2: 85545326-85545326 on Build GRCh38
rs144074011 Chr.2: 85545521-85545521 on Build GRCh38
rs893957344 Chr.2: 85545458-85545480 on Build GRCh38
rs3209939 Chr.2: 85545310-85545310 on Build GRCh38
rs575528704 Chr.2: 85545396-85545396 on Build GRCh38
rs113066451 Chr.2: 85545372-85545372 on Build GRCh38
rs776514553 Chr.2: 85545528-85545537 on Build GRCh38
rs886056357 Chr.2: 85545646-85545646 on Build GRCh38
rs932523625 Chr.2: 85545608-85545608 on Build GRCh38
rs1006196660 Chr.2: 85545417-85545417 on Build GRCh38
rs62165899 Chr.2: 85545505-85545505 on Build GRCh38
rs946320818 Chr.2: 85545574-85545574 on Build GRCh38
rs1016299860 Chr.2: 85545672-85545672 on Build GRCh38
rs766005676 Chr.2: 85545489-85545489 on Build GRCh38

View Full Product Details