Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85609184-85609682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf68
Entrez Gene ID
388969
Gene Name
chromosome 2 open reading frame 68
Gene Aliases
HCRCN81
UniGene
Hs.516159
Gene Chromosome Location
Chr.2: 85605253-85612073 on Build GRCh38
Transcripts
NM_001013649.3 XM_005264305.3 NR_046347.1 XM_006711922.1

Gene Symbol
USP39
Entrez Gene ID
10713
Gene Name
ubiquitin specific peptidase 39
Gene Aliases
65K, CGI-21, HSPC332, SAD1, SNRNP65
UniGene
Hs.654745
Gene Chromosome Location
Chr.2: 85602842-85649284 on Build GRCh38
Transcripts
NM_001013649.3 XM_005264305.3 NR_046347.1 XM_006711922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6994549 c.287G>T p.G96V Chr.2: 85609526-85609526 on Build GRCh38
5949693 c.268G>A p.G90S Chr.2: 85609545-85609545 on Build GRCh38
4096045 c.341C>G p.A114G Chr.2: 85609472-85609472 on Build GRCh38
4096044 c.370G>A p.V124I Chr.2: 85609443-85609443 on Build GRCh38
5819793 c.328T>G p.L110V Chr.2: 85609485-85609485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767998112 Chr.2: 85609477-85609477 on Build GRCh38
rs748616457 Chr.2: 85609598-85609598 on Build GRCh38
rs201733655 Chr.2: 85609592-85609592 on Build GRCh38
rs1011715520 Chr.2: 85609495-85609495 on Build GRCh38
rs774088906 Chr.2: 85609425-85609425 on Build GRCh38
rs765042256 Chr.2: 85609588-85609589 on Build GRCh38
rs972656596 Chr.2: 85609407-85609407 on Build GRCh38
rs763143275 Chr.2: 85609451-85609451 on Build GRCh38
rs762843845 Chr.2: 85609617-85609618 on Build GRCh38
rs769812404 Chr.2: 85609625-85609625 on Build GRCh38
rs986621377 Chr.2: 85609295-85609295 on Build GRCh38
rs749723585 Chr.2: 85609520-85609520 on Build GRCh38
rs760612952 Chr.2: 85609443-85609443 on Build GRCh38
rs915614905 Chr.2: 85609466-85609466 on Build GRCh38
rs966205631 Chr.2: 85609371-85609371 on Build GRCh38
rs372403057 Chr.2: 85609546-85609546 on Build GRCh38
rs1022333744 Chr.2: 85609333-85609333 on Build GRCh38
rs61743294 Chr.2: 85609545-85609545 on Build GRCh38
rs911054165 Chr.2: 85609303-85609303 on Build GRCh38
rs200832393 Chr.2: 85609619-85609619 on Build GRCh38
rs775489877 Chr.2: 85609445-85609446 on Build GRCh38
rs938896088 Chr.2: 85609392-85609392 on Build GRCh38
rs544571612 Chr.2: 85609321-85609321 on Build GRCh38
rs757496159 Chr.2: 85609476-85609476 on Build GRCh38
rs1021660054 Chr.2: 85609224-85609224 on Build GRCh38
rs777453681 Chr.2: 85609620-85609620 on Build GRCh38
rs1045590765 Chr.2: 85609508-85609508 on Build GRCh38
rs199705004 Chr.2: 85609497-85609497 on Build GRCh38
rs772937056 Chr.2: 85609439-85609439 on Build GRCh38
rs532868818 Chr.2: 85609568-85609568 on Build GRCh38
rs369096715 Chr.2: 85609569-85609569 on Build GRCh38
rs986673834 Chr.2: 85609571-85609571 on Build GRCh38
rs756598972 Chr.2: 85609496-85609496 on Build GRCh38
rs750775320 Chr.2: 85609480-85609480 on Build GRCh38
rs139096231 Chr.2: 85609361-85609361 on Build GRCh38
rs750340660 Chr.2: 85609617-85609617 on Build GRCh38
rs746759997 Chr.2: 85609556-85609556 on Build GRCh38
rs778385123 Chr.2: 85609536-85609536 on Build GRCh38
rs562980191 Chr.2: 85609325-85609325 on Build GRCh38
rs770609706 Chr.2: 85609564-85609564 on Build GRCh38
rs747814463 Chr.2: 85609544-85609544 on Build GRCh38
rs762155696 Chr.2: 85609577-85609577 on Build GRCh38
rs536341650 Chr.2: 85609621-85609621 on Build GRCh38
rs371396406 Chr.2: 85609572-85609572 on Build GRCh38
rs1031351548 Chr.2: 85609273-85609273 on Build GRCh38
rs760829024 Chr.2: 85609571-85609571 on Build GRCh38
rs942487474 Chr.2: 85609317-85609318 on Build GRCh38
rs762476737 Chr.2: 85609230-85609230 on Build GRCh38
rs754586751 Chr.2: 85609528-85609528 on Build GRCh38
rs199984532 Chr.2: 85609635-85609635 on Build GRCh38
rs764050244 Chr.2: 85609455-85609455 on Build GRCh38
rs1045538241 Chr.2: 85609236-85609236 on Build GRCh38
rs928555463 Chr.2: 85609539-85609539 on Build GRCh38
rs771916910 Chr.2: 85609436-85609436 on Build GRCh38
rs919648294 Chr.2: 85609331-85609331 on Build GRCh38
rs755385923 Chr.2: 85609618-85609618 on Build GRCh38
rs553188002 Chr.2: 85609254-85609254 on Build GRCh38
rs868268307 Chr.2: 85609601-85609601 on Build GRCh38
rs768025934 Chr.2: 85609404-85609404 on Build GRCh38
rs147787865 Chr.2: 85609277-85609277 on Build GRCh38
rs973911121 Chr.2: 85609607-85609610 on Build GRCh38
rs967788756 Chr.2: 85609248-85609248 on Build GRCh38
rs748779310 Chr.2: 85609387-85609387 on Build GRCh38
rs747327191 Chr.2: 85609330-85609330 on Build GRCh38
rs200523901 Chr.2: 85609624-85609624 on Build GRCh38
rs751887839 Chr.2: 85609456-85609456 on Build GRCh38
rs902114659 Chr.2: 85609650-85609650 on Build GRCh38
rs770826758 Chr.2: 85609622-85609622 on Build GRCh38
rs542754303 Chr.2: 85609341-85609341 on Build GRCh38
rs766644750 Chr.2: 85609603-85609603 on Build GRCh38
rs574973523 Chr.2: 85609332-85609332 on Build GRCh38
rs372399329 Chr.2: 85609430-85609430 on Build GRCh38
rs766195688 Chr.2: 85609618-85609618 on Build GRCh38
rs774591898 Chr.2: 85609626-85609626 on Build GRCh38
rs186499615 Chr.2: 85609567-85609567 on Build GRCh38
rs1007855554 Chr.2: 85609360-85609360 on Build GRCh38
rs750648949 Chr.2: 85609584-85609584 on Build GRCh38
rs754429802 Chr.2: 85609613-85609613 on Build GRCh38
rs375450706 Chr.2: 85609444-85609444 on Build GRCh38
rs948671393 Chr.2: 85609210-85609210 on Build GRCh38
rs767795582 Chr.2: 85609580-85609580 on Build GRCh38
rs74324298 Chr.2: 85609623-85609623 on Build GRCh38
rs977407469 Chr.2: 85609257-85609257 on Build GRCh38
rs541264911 Chr.2: 85609265-85609265 on Build GRCh38
rs534790364 Chr.2: 85609628-85609628 on Build GRCh38

View Full Product Details