Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86143925-86144436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMMT
Entrez Gene ID
10989
Gene Name
inner membrane mitochondrial protein
Gene Aliases
HMP, MINOS2, Mic60, P87, P87/89, P89, PIG4, PIG52
UniGene
Hs.148559
Gene Chromosome Location
Chr.2: 86143932-86195770 on Build GRCh38
Transcripts
NM_001100169.1 NM_001100170.1 NM_006839.2 XM_005264110.3 XM_005264113.1 XM_005264114.1 XM_011532497.2 XM_017003194.1 XM_017003195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
722995 c.2272G>C p.E758Q Chr.2: 86144273-86144273 on Build GRCh38
3991489 c.2111A>T p.E704V Chr.2: 86144434-86144434 on Build GRCh38
4141283 c.2196A>T p.L732L Chr.2: 86144349-86144349 on Build GRCh38
5783646 c.2141A>G p.K714R Chr.2: 86144404-86144404 on Build GRCh38
245030 c.2234C>T p.A745V Chr.2: 86144311-86144311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778267232 Chr.2: 86144307-86144307 on Build GRCh38
rs751752839 Chr.2: 86144394-86144394 on Build GRCh38
rs747304136 Chr.2: 86144313-86144313 on Build GRCh38
rs561421843 Chr.2: 86144185-86144185 on Build GRCh38
rs779494787 Chr.2: 86144221-86144222 on Build GRCh38
rs1028949631 Chr.2: 86144088-86144088 on Build GRCh38
rs528241458 Chr.2: 86144118-86144118 on Build GRCh38
rs113321722 Chr.2: 86144126-86144126 on Build GRCh38
rs367754331 Chr.2: 86144400-86144400 on Build GRCh38
rs756679628 Chr.2: 86144413-86144413 on Build GRCh38
rs945597609 Chr.2: 86144009-86144009 on Build GRCh38
rs183939253 Chr.2: 86144303-86144303 on Build GRCh38
rs562897498 Chr.2: 86144069-86144069 on Build GRCh38
rs762291793 Chr.2: 86144217-86144217 on Build GRCh38
rs777454852 Chr.2: 86144333-86144336 on Build GRCh38
rs760539285 Chr.2: 86144385-86144385 on Build GRCh38
rs758536015 Chr.2: 86144306-86144306 on Build GRCh38
rs546408439 Chr.2: 86144128-86144128 on Build GRCh38
rs746334035 Chr.2: 86144339-86144339 on Build GRCh38
rs373231227 Chr.2: 86144238-86144238 on Build GRCh38
rs936743725 Chr.2: 86143982-86143982 on Build GRCh38
rs774007561 Chr.2: 86144276-86144276 on Build GRCh38
rs752968371 Chr.2: 86144304-86144304 on Build GRCh38
rs200512625 Chr.2: 86144356-86144356 on Build GRCh38
rs781632033 Chr.2: 86144403-86144403 on Build GRCh38
rs11845 Chr.2: 86144110-86144110 on Build GRCh38
rs762441369 Chr.2: 86144257-86144257 on Build GRCh38
rs776319715 Chr.2: 86144234-86144234 on Build GRCh38
rs764198090 Chr.2: 86144392-86144392 on Build GRCh38
rs907573486 Chr.2: 86144048-86144048 on Build GRCh38
rs781769634 Chr.2: 86144337-86144337 on Build GRCh38
rs892485284 Chr.2: 86144002-86144004 on Build GRCh38
rs902539144 Chr.2: 86144184-86144184 on Build GRCh38
rs138955172 Chr.2: 86144079-86144079 on Build GRCh38
rs756986840 Chr.2: 86144042-86144043 on Build GRCh38
rs761880434 Chr.2: 86144280-86144280 on Build GRCh38
rs777037074 Chr.2: 86144202-86144202 on Build GRCh38
rs749966188 Chr.2: 86144414-86144414 on Build GRCh38
rs113425155 Chr.2: 86144156-86144156 on Build GRCh38
rs760916623 Chr.2: 86144176-86144176 on Build GRCh38
rs992930867 Chr.2: 86144138-86144138 on Build GRCh38
rs746387163 Chr.2: 86144406-86144406 on Build GRCh38
rs768193439 Chr.2: 86144359-86144359 on Build GRCh38
rs771337004 Chr.2: 86144225-86144225 on Build GRCh38
rs745893774 Chr.2: 86143965-86143965 on Build GRCh38
rs201764625 Chr.2: 86144334-86144334 on Build GRCh38
rs753970633 Chr.2: 86144301-86144301 on Build GRCh38
rs973156990 Chr.2: 86144084-86144084 on Build GRCh38
rs755686154 Chr.2: 86144245-86144246 on Build GRCh38
rs146069459 Chr.2: 86143992-86143992 on Build GRCh38
rs763967469 Chr.2: 86144259-86144259 on Build GRCh38
rs970417559 Chr.2: 86144178-86144178 on Build GRCh38
rs920227118 Chr.2: 86144378-86144378 on Build GRCh38
rs925786928 Chr.2: 86144194-86144194 on Build GRCh38
rs767414578 Chr.2: 86144296-86144296 on Build GRCh38
rs961907901 Chr.2: 86144341-86144341 on Build GRCh38
rs774245699 Chr.2: 86144360-86144360 on Build GRCh38
rs765770591 Chr.2: 86144297-86144297 on Build GRCh38
rs759091751 Chr.2: 86144235-86144235 on Build GRCh38
rs753993433 Chr.2: 86144031-86144031 on Build GRCh38
rs1032318520 Chr.2: 86144203-86144203 on Build GRCh38
rs974594557 Chr.2: 86143955-86143955 on Build GRCh38
rs757725417 Chr.2: 86144399-86144399 on Build GRCh38
rs768947577 Chr.2: 86144199-86144199 on Build GRCh38
rs370590382 Chr.2: 86144343-86144343 on Build GRCh38
rs765358502 Chr.2: 86144387-86144387 on Build GRCh38
rs1047065745 Chr.2: 86144177-86144177 on Build GRCh38
rs756734991 Chr.2: 86143967-86143967 on Build GRCh38
rs200487700 Chr.2: 86144351-86144351 on Build GRCh38
rs560874003 Chr.2: 86144111-86144111 on Build GRCh38
rs997018194 Chr.2: 86144087-86144087 on Build GRCh38
rs958082215 Chr.2: 86144198-86144198 on Build GRCh38
rs1024216165 Chr.2: 86144172-86144172 on Build GRCh38
rs373987983 Chr.2: 86144370-86144370 on Build GRCh38
rs949701646 Chr.2: 86144135-86144135 on Build GRCh38
rs752714248 Chr.2: 86144389-86144389 on Build GRCh38
rs773165856 Chr.2: 86144380-86144380 on Build GRCh38
rs767253052 Chr.2: 86144377-86144377 on Build GRCh38
rs945384683 Chr.2: 86144209-86144209 on Build GRCh38
rs752514769 Chr.2: 86144246-86144247 on Build GRCh38
rs775309637 Chr.2: 86144242-86144242 on Build GRCh38
rs987789653 Chr.2: 86144016-86144016 on Build GRCh38

View Full Product Details