Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86145838-86146352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMMT
Entrez Gene ID
10989
Gene Name
inner membrane mitochondrial protein
Gene Aliases
HMP, MINOS2, Mic60, P87, P87/89, P89, PIG4, PIG52
UniGene
Hs.148559
Gene Chromosome Location
Chr.2: 86143932-86195770 on Build GRCh38
Transcripts
NM_001100169.1 NM_001100170.1 NM_006839.2 XM_005264110.3 XM_005264113.1 XM_005264114.1 XM_011532497.2 XM_017003194.1 XM_017003195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7156756 c.1634G>T p.R545I Chr.2: 86146097-86146097 on Build GRCh38
281918 c.1573C>T p.R525C Chr.2: 86146158-86146158 on Build GRCh38
6407582 c.1630G>A p.A544T Chr.2: 86146101-86146101 on Build GRCh38
6456952 c.1534-1G>C p.? Chr.2: 86146198-86146198 on Build GRCh38
2822957 c.1576C>T p.R526C Chr.2: 86146155-86146155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922974801 Chr.2: 86146053-86146053 on Build GRCh38
rs752460735 Chr.2: 86146013-86146013 on Build GRCh38
rs755344583 Chr.2: 86146210-86146210 on Build GRCh38
rs966273981 Chr.2: 86145942-86145942 on Build GRCh38
rs781074678 Chr.2: 86146033-86146033 on Build GRCh38
rs758251509 Chr.2: 86146164-86146164 on Build GRCh38
rs994575196 Chr.2: 86145863-86145863 on Build GRCh38
rs952029524 Chr.2: 86145991-86145991 on Build GRCh38
rs778633886 Chr.2: 86146210-86146211 on Build GRCh38
rs746156476 Chr.2: 86146052-86146052 on Build GRCh38
rs545822170 Chr.2: 86145915-86145915 on Build GRCh38
rs771628141 Chr.2: 86146103-86146103 on Build GRCh38
rs760470893 Chr.2: 86146111-86146111 on Build GRCh38
rs947047597 Chr.2: 86146180-86146180 on Build GRCh38
rs772454563 Chr.2: 86146220-86146220 on Build GRCh38
rs142392424 Chr.2: 86146231-86146231 on Build GRCh38
rs558404369 Chr.2: 86145951-86145951 on Build GRCh38
rs752692441 Chr.2: 86146160-86146160 on Build GRCh38
rs766100087 Chr.2: 86146136-86146136 on Build GRCh38
rs770462726 Chr.2: 86146065-86146066 on Build GRCh38
rs779092740 Chr.2: 86146215-86146215 on Build GRCh38
rs750437047 Chr.2: 86146171-86146171 on Build GRCh38
rs977457555 Chr.2: 86146023-86146023 on Build GRCh38
rs547347985 Chr.2: 86145916-86145916 on Build GRCh38
rs116681049 Chr.2: 86146002-86146002 on Build GRCh38
rs201463639 Chr.2: 86146187-86146187 on Build GRCh38
rs763850388 Chr.2: 86146255-86146255 on Build GRCh38
rs763955250 Chr.2: 86146276-86146276 on Build GRCh38
rs1034622586 Chr.2: 86145920-86145920 on Build GRCh38
rs756374553 Chr.2: 86146173-86146173 on Build GRCh38
rs1019491352 Chr.2: 86145908-86145908 on Build GRCh38
rs368283488 Chr.2: 86146157-86146157 on Build GRCh38
rs762601983 Chr.2: 86146180-86146181 on Build GRCh38
rs993701654 Chr.2: 86145974-86145974 on Build GRCh38
rs1049834846 Chr.2: 86146183-86146183 on Build GRCh38
rs1025648598 Chr.2: 86145896-86145896 on Build GRCh38
rs770524449 Chr.2: 86146234-86146234 on Build GRCh38
rs560315070 Chr.2: 86146155-86146155 on Build GRCh38
rs6724856 Chr.2: 86146311-86146311 on Build GRCh38
rs1007744316 Chr.2: 86145907-86145907 on Build GRCh38
rs941860808 Chr.2: 86146185-86146185 on Build GRCh38
rs1003313114 Chr.2: 86146251-86146251 on Build GRCh38
rs529938667 Chr.2: 86146069-86146069 on Build GRCh38
rs541929797 Chr.2: 86146084-86146084 on Build GRCh38
rs757362353 Chr.2: 86146032-86146032 on Build GRCh38
rs769995232 Chr.2: 86146058-86146058 on Build GRCh38
rs376280818 Chr.2: 86146061-86146061 on Build GRCh38
rs562539292 Chr.2: 86146016-86146016 on Build GRCh38
rs774956433 Chr.2: 86146096-86146096 on Build GRCh38
rs375926531 Chr.2: 86146109-86146109 on Build GRCh38
rs1034591254 Chr.2: 86146259-86146259 on Build GRCh38
rs372290516 Chr.2: 86146189-86146189 on Build GRCh38
rs114088948 Chr.2: 86146158-86146158 on Build GRCh38
rs993735859 Chr.2: 86146318-86146318 on Build GRCh38
rs201987512 Chr.2: 86146169-86146169 on Build GRCh38
rs981302794 Chr.2: 86146150-86146150 on Build GRCh38
rs749475004 Chr.2: 86146205-86146205 on Build GRCh38
rs746311647 Chr.2: 86146238-86146239 on Build GRCh38
rs889501987 Chr.2: 86145905-86145905 on Build GRCh38
rs755867536 Chr.2: 86146083-86146083 on Build GRCh38
rs914319240 Chr.2: 86146009-86146009 on Build GRCh38
rs371394842 Chr.2: 86146098-86146098 on Build GRCh38
rs1026313204 Chr.2: 86145985-86145985 on Build GRCh38
rs759117511 Chr.2: 86145883-86145883 on Build GRCh38
rs1049476419 Chr.2: 86145899-86145899 on Build GRCh38
rs368117007 Chr.2: 86146244-86146244 on Build GRCh38
rs776425248 Chr.2: 86146238-86146238 on Build GRCh38
rs764335275 Chr.2: 86146288-86146289 on Build GRCh38
rs1050348309 Chr.2: 86146100-86146100 on Build GRCh38
rs367650891 Chr.2: 86146078-86146078 on Build GRCh38
rs753491797 Chr.2: 86146154-86146154 on Build GRCh38
rs544347268 Chr.2: 86146005-86146005 on Build GRCh38

View Full Product Details