Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86791427-86791919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD8A
Entrez Gene ID
925
Gene Name
CD8a molecule
Gene Aliases
CD8, Leu2, MAL, p32
UniGene
Hs.85258
Gene Chromosome Location
Chr.2: 86784605-86808396 on Build GRCh38
Transcripts
NM_001145873.1 NM_001768.6 NM_171827.3 NR_027353.1

SNP Detail

SNP ID Chromosome Location
rs912274286 Chr.2: 86791808-86791808 on Build GRCh38
rs926809009 Chr.2: 86791738-86791738 on Build GRCh38
rs895447145 Chr.2: 86791800-86791800 on Build GRCh38
rs973663799 Chr.2: 86791794-86791794 on Build GRCh38
rs1039555376 Chr.2: 86791482-86791482 on Build GRCh38
rs544497400 Chr.2: 86791893-86791893 on Build GRCh38
rs186166993 Chr.2: 86791855-86791855 on Build GRCh38
rs567710168 Chr.2: 86791664-86791664 on Build GRCh38
rs936570494 Chr.2: 86791875-86791875 on Build GRCh38
rs36226880 Chr.2: 86791685-86791685 on Build GRCh38
rs542526339 Chr.2: 86791577-86791577 on Build GRCh38
rs944157451 Chr.2: 86791611-86791611 on Build GRCh38
rs774291179 Chr.2: 86791623-86791623 on Build GRCh38
rs773246042 Chr.2: 86791665-86791665 on Build GRCh38
rs1048198929 Chr.2: 86791682-86791682 on Build GRCh38
rs534968901 Chr.2: 86791683-86791683 on Build GRCh38
rs905376739 Chr.2: 86791836-86791836 on Build GRCh38
rs62146066 Chr.2: 86791455-86791455 on Build GRCh38
rs1008499569 Chr.2: 86791459-86791459 on Build GRCh38
rs377000831 Chr.2: 86791887-86791887 on Build GRCh38
rs925502290 Chr.2: 86791529-86791529 on Build GRCh38
rs956732495 Chr.2: 86791622-86791622 on Build GRCh38
rs1032496723 Chr.2: 86791852-86791852 on Build GRCh38
rs887055454 Chr.2: 86791730-86791730 on Build GRCh38
rs899668816 Chr.2: 86791857-86791857 on Build GRCh38
rs899720997 Chr.2: 86791531-86791531 on Build GRCh38
rs1000790467 Chr.2: 86791535-86791535 on Build GRCh38
rs772159785 Chr.2: 86791662-86791662 on Build GRCh38
rs557958674 Chr.2: 86791807-86791807 on Build GRCh38
rs1024224849 Chr.2: 86791473-86791473 on Build GRCh38
rs1047285385 Chr.2: 86791452-86791452 on Build GRCh38
rs1053737139 Chr.2: 86791880-86791880 on Build GRCh38
rs547381985 Chr.2: 86791699-86791699 on Build GRCh38
rs36226879 Chr.2: 86791800-86791800 on Build GRCh38
rs912558230 Chr.2: 86791595-86791595 on Build GRCh38
rs1039522367 Chr.2: 86791844-86791844 on Build GRCh38
rs981324873 Chr.2: 86791733-86791733 on Build GRCh38
rs1012666619 Chr.2: 86791463-86791463 on Build GRCh38
rs141578385 Chr.2: 86791801-86791801 on Build GRCh38
rs1056764372 Chr.2: 86791758-86791758 on Build GRCh38
rs867381803 Chr.2: 86791806-86791806 on Build GRCh38
rs182998895 Chr.2: 86791636-86791636 on Build GRCh38
rs963564861 Chr.2: 86791747-86791747 on Build GRCh38
rs34660074 Chr.2: 86791687-86791687 on Build GRCh38
rs1009648135 Chr.2: 86791648-86791648 on Build GRCh38
rs573713611 Chr.2: 86791560-86791560 on Build GRCh38
rs759702187 Chr.2: 86791735-86791735 on Build GRCh38
rs975490545 Chr.2: 86791620-86791620 on Build GRCh38
rs748878357 Chr.2: 86791493-86791493 on Build GRCh38
rs956921544 Chr.2: 86791567-86791567 on Build GRCh38
rs943814996 Chr.2: 86791835-86791835 on Build GRCh38
rs921112105 Chr.2: 86791634-86791634 on Build GRCh38

View Full Product Details