Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88173055-88173560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THNSL2
Entrez Gene ID
55258
Gene Name
threonine synthase like 2
Gene Aliases
SOFAT, THS2, TSH2
UniGene
Hs.739080
Gene Chromosome Location
Chr.2: 88170295-88186637 on Build GRCh38
Transcripts
NM_001244676.1 NM_001244678.1 NM_018271.4 XM_005264400.4 XM_005264401.4 XM_005264402.3 XM_005264403.4 XM_005264405.4 XM_006712041.3 XM_006712042.3 XM_006712043.2 XM_006712044.3 XM_011532953.2 XM_011532954.2 XM_011532955.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6366117 c.103C>A p.L35I Chr.2: 88173253-88173253 on Build GRCh38
722947 c.207A>G p.P69P Chr.2: 88173357-88173357 on Build GRCh38
6413254 c.85C>T p.L29F Chr.2: 88173235-88173235 on Build GRCh38
6366118 c.103C>A p.L35I Chr.2: 88173253-88173253 on Build GRCh38
5397554 c.205C>T p.P69S Chr.2: 88173355-88173355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370944153 Chr.2: 88173279-88173279 on Build GRCh38
rs755534002 Chr.2: 88173489-88173489 on Build GRCh38
rs780713167 Chr.2: 88173151-88173151 on Build GRCh38
rs992770613 Chr.2: 88173084-88173084 on Build GRCh38
rs752036548 Chr.2: 88173447-88173447 on Build GRCh38
rs778995468 Chr.2: 88173252-88173252 on Build GRCh38
rs570440662 Chr.2: 88173381-88173381 on Build GRCh38
rs377402157 Chr.2: 88173407-88173407 on Build GRCh38
rs151033743 Chr.2: 88173218-88173218 on Build GRCh38
rs746855873 Chr.2: 88173384-88173384 on Build GRCh38
rs897406236 Chr.2: 88173147-88173147 on Build GRCh38
rs1031478675 Chr.2: 88173535-88173535 on Build GRCh38
rs762450614 Chr.2: 88173302-88173302 on Build GRCh38
rs369820777 Chr.2: 88173215-88173215 on Build GRCh38
rs752048862 Chr.2: 88173299-88173299 on Build GRCh38
rs1030730532 Chr.2: 88173523-88173523 on Build GRCh38
rs771748148 Chr.2: 88173256-88173256 on Build GRCh38
rs746257144 Chr.2: 88173166-88173166 on Build GRCh38
rs1037322748 Chr.2: 88173141-88173141 on Build GRCh38
rs888626169 Chr.2: 88173120-88173120 on Build GRCh38
rs146704484 Chr.2: 88173281-88173281 on Build GRCh38
rs766553064 Chr.2: 88173184-88173184 on Build GRCh38
rs4129190 Chr.2: 88173272-88173272 on Build GRCh38
rs758947055 Chr.2: 88173129-88173129 on Build GRCh38
rs779884008 Chr.2: 88173355-88173355 on Build GRCh38
rs201183724 Chr.2: 88173405-88173405 on Build GRCh38
rs917361601 Chr.2: 88173091-88173091 on Build GRCh38
rs765203636 Chr.2: 88173192-88173192 on Build GRCh38
rs1007603568 Chr.2: 88173480-88173480 on Build GRCh38
rs1007326682 Chr.2: 88173236-88173236 on Build GRCh38
rs758858874 Chr.2: 88173197-88173197 on Build GRCh38
rs367785526 Chr.2: 88173174-88173174 on Build GRCh38
rs775702879 Chr.2: 88173401-88173401 on Build GRCh38
rs1038817043 Chr.2: 88173486-88173486 on Build GRCh38
rs779687764 Chr.2: 88173233-88173233 on Build GRCh38
rs374817310 Chr.2: 88173108-88173108 on Build GRCh38
rs374311669 Chr.2: 88173182-88173182 on Build GRCh38
rs139953850 Chr.2: 88173185-88173185 on Build GRCh38
rs934701214 Chr.2: 88173076-88173076 on Build GRCh38
rs147509382 Chr.2: 88173313-88173313 on Build GRCh38
rs769966085 Chr.2: 88173255-88173255 on Build GRCh38
rs762092823 Chr.2: 88173188-88173188 on Build GRCh38
rs1044852725 Chr.2: 88173105-88173105 on Build GRCh38
rs781210973 Chr.2: 88173170-88173184 on Build GRCh38
rs768674286 Chr.2: 88173244-88173244 on Build GRCh38
rs553775935 Chr.2: 88173462-88173463 on Build GRCh38
rs778619539 Chr.2: 88173365-88173365 on Build GRCh38
rs540198148 Chr.2: 88173163-88173163 on Build GRCh38
rs756897774 Chr.2: 88173150-88173150 on Build GRCh38
rs979295275 Chr.2: 88173087-88173087 on Build GRCh38
rs918873334 Chr.2: 88173442-88173442 on Build GRCh38
rs775485495 Chr.2: 88173133-88173133 on Build GRCh38
rs948821364 Chr.2: 88173097-88173097 on Build GRCh38
rs544283083 Chr.2: 88173222-88173222 on Build GRCh38
rs748196817 Chr.2: 88173179-88173179 on Build GRCh38
rs537739649 Chr.2: 88173253-88173253 on Build GRCh38
rs747458725 Chr.2: 88173340-88173340 on Build GRCh38
rs748829684 Chr.2: 88173237-88173237 on Build GRCh38
rs527305520 Chr.2: 88173329-88173329 on Build GRCh38
rs755015656 Chr.2: 88173104-88173104 on Build GRCh38
rs879037811 Chr.2: 88173251-88173251 on Build GRCh38
rs753185685 Chr.2: 88173109-88173109 on Build GRCh38
rs373551702 Chr.2: 88173227-88173227 on Build GRCh38
rs755671875 Chr.2: 88173341-88173341 on Build GRCh38
rs753330743 Chr.2: 88173362-88173362 on Build GRCh38
rs768760412 Chr.2: 88173178-88173178 on Build GRCh38
rs917268762 Chr.2: 88173300-88173300 on Build GRCh38
rs1000062188 Chr.2: 88173512-88173512 on Build GRCh38
rs752825797 Chr.2: 88173196-88173196 on Build GRCh38
rs890129354 Chr.2: 88173473-88173473 on Build GRCh38
rs747401298 Chr.2: 88173148-88173148 on Build GRCh38
rs4129189 Chr.2: 88173406-88173406 on Build GRCh38
rs142011143 Chr.2: 88173292-88173292 on Build GRCh38
rs774238288 Chr.2: 88173180-88173180 on Build GRCh38
rs201994131 Chr.2: 88173382-88173382 on Build GRCh38
rs114858196 Chr.2: 88173228-88173228 on Build GRCh38
rs545602278 Chr.2: 88173229-88173229 on Build GRCh38
rs769940151 Chr.2: 88173397-88173397 on Build GRCh38
rs775208851 Chr.2: 88173175-88173175 on Build GRCh38
rs143468353 Chr.2: 88173290-88173290 on Build GRCh38
rs748028126 Chr.2: 88173374-88173374 on Build GRCh38
rs199500112 Chr.2: 88173364-88173364 on Build GRCh38
rs948684709 Chr.2: 88173305-88173305 on Build GRCh38
rs764527693 Chr.2: 88173296-88173296 on Build GRCh38
rs1051325288 Chr.2: 88173467-88173467 on Build GRCh38
rs775512431 Chr.2: 88173285-88173285 on Build GRCh38

View Full Product Details