Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 95114864-95115388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS5
Entrez Gene ID
64969
Gene Name
mitochondrial ribosomal protein S5
Gene Aliases
MRP-S5, S5mt
UniGene
Hs.655259
Gene Chromosome Location
Chr.2: 95085391-95122027 on Build GRCh38
Transcripts
NM_001321995.1 NM_001321996.1 NM_001321997.1 NM_031902.4 NR_136152.1 NR_136153.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6715523 c.224C>T p.S75F Chr.2: 95115119-95115119 on Build GRCh38
477743 c.160A>G p.T54A Chr.2: 95115183-95115183 on Build GRCh38
1023676 c.256C>A p.P86T Chr.2: 95115087-95115087 on Build GRCh38
1632163 c.262_263insTATA p.S88fs*2 Chr.2: 95115080-95115081 on Build GRCh38
1023675 c.258A>G p.P86P Chr.2: 95115085-95115085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999037660 Chr.2: 95114954-95114954 on Build GRCh38
rs776241678 Chr.2: 95115166-95115166 on Build GRCh38
rs368818231 Chr.2: 95115136-95115136 on Build GRCh38
rs767188777 Chr.2: 95115250-95115250 on Build GRCh38
rs762932815 Chr.2: 95115298-95115298 on Build GRCh38
rs769826733 Chr.2: 95115061-95115061 on Build GRCh38
rs776630688 Chr.2: 95115196-95115196 on Build GRCh38
rs773242628 Chr.2: 95115109-95115109 on Build GRCh38
rs765716938 Chr.2: 95115117-95115117 on Build GRCh38
rs749084113 Chr.2: 95115103-95115103 on Build GRCh38
rs765866543 Chr.2: 95115360-95115360 on Build GRCh38
rs758103073 Chr.2: 95115018-95115018 on Build GRCh38
rs944374066 Chr.2: 95115295-95115295 on Build GRCh38
rs201430985 Chr.2: 95115149-95115149 on Build GRCh38
rs765066544 Chr.2: 95115116-95115117 on Build GRCh38
rs760873774 Chr.2: 95115028-95115029 on Build GRCh38
rs997529910 Chr.2: 95115365-95115365 on Build GRCh38
rs890160430 Chr.2: 95115037-95115037 on Build GRCh38
rs200274600 Chr.2: 95115114-95115114 on Build GRCh38
rs770289979 Chr.2: 95115206-95115206 on Build GRCh38
rs1053288351 Chr.2: 95115282-95115282 on Build GRCh38
rs756820228 Chr.2: 95115145-95115145 on Build GRCh38
rs556951498 Chr.2: 95115112-95115112 on Build GRCh38
rs542646463 Chr.2: 95115125-95115125 on Build GRCh38
rs537283433 Chr.2: 95115077-95115077 on Build GRCh38
rs186030997 Chr.2: 95115162-95115162 on Build GRCh38
rs904261742 Chr.2: 95114934-95114934 on Build GRCh38
rs1041288034 Chr.2: 95115306-95115306 on Build GRCh38
rs890088185 Chr.2: 95115291-95115291 on Build GRCh38
rs200715882 Chr.2: 95115203-95115203 on Build GRCh38
rs371204975 Chr.2: 95115163-95115163 on Build GRCh38
rs952807815 Chr.2: 95115134-95115134 on Build GRCh38
rs769551208 Chr.2: 95115209-95115209 on Build GRCh38
rs534422896 Chr.2: 95115028-95115028 on Build GRCh38
rs142632288 Chr.2: 95115150-95115150 on Build GRCh38
rs997037560 Chr.2: 95115100-95115100 on Build GRCh38
rs200418387 Chr.2: 95115116-95115116 on Build GRCh38
rs200128920 Chr.2: 95115121-95115121 on Build GRCh38
rs868815642 Chr.2: 95115309-95115309 on Build GRCh38
rs376122502 Chr.2: 95115113-95115113 on Build GRCh38
rs747161233 Chr.2: 95115182-95115182 on Build GRCh38
rs367791599 Chr.2: 95115236-95115236 on Build GRCh38
rs565430255 Chr.2: 95114985-95114985 on Build GRCh38
rs374428514 Chr.2: 95115132-95115132 on Build GRCh38
rs1052784575 Chr.2: 95114997-95114997 on Build GRCh38
rs934419480 Chr.2: 95115274-95115274 on Build GRCh38
rs571123211 Chr.2: 95115056-95115056 on Build GRCh38
rs756180899 Chr.2: 95115153-95115153 on Build GRCh38
rs772491765 Chr.2: 95115108-95115108 on Build GRCh38
rs771155530 Chr.2: 95115188-95115188 on Build GRCh38
rs557485910 Chr.2: 95115040-95115040 on Build GRCh38
rs148482144 Chr.2: 95115207-95115207 on Build GRCh38
rs781206834 Chr.2: 95115042-95115042 on Build GRCh38
rs199651077 Chr.2: 95115211-95115211 on Build GRCh38
rs144545594 Chr.2: 95115128-95115128 on Build GRCh38
rs142646997 Chr.2: 95115257-95115257 on Build GRCh38
rs149308126 Chr.2: 95114907-95114907 on Build GRCh38
rs372879753 Chr.2: 95115085-95115085 on Build GRCh38
rs758541811 Chr.2: 95114924-95114924 on Build GRCh38
rs774743884 Chr.2: 95115217-95115217 on Build GRCh38

View Full Product Details