Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 100393316-100393823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST10
Entrez Gene ID
9486
Gene Name
carbohydrate sulfotransferase 10
Gene Aliases
HNK-1ST, HNK1ST
UniGene
Hs.516370
Gene Chromosome Location
Chr.2: 100391860-100417668 on Build GRCh38
Transcripts
NM_004854.4 XM_011512207.1 XM_011512208.1 XM_011512210.1 XM_011512211.1 XM_011512212.1 XM_017005380.1 XM_017005381.1 XM_017005382.1 XM_017005383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4556198 c.825G>T p.R275S Chr.2: 100393635-100393635 on Build GRCh38
4415465 c.870C>T p.A290A Chr.2: 100393446-100393446 on Build GRCh38
6891230 c.885G>A p.P295P Chr.2: 100393575-100393575 on Build GRCh38
4421036 c.679T>A p.L227M Chr.2: 100393781-100393781 on Build GRCh38
3372363 c.1062G>A p.P354P Chr.2: 100393398-100393398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59148684 Chr.2: 100393386-100393386 on Build GRCh38
rs747722795 Chr.2: 100393445-100393445 on Build GRCh38
rs760096775 Chr.2: 100393382-100393382 on Build GRCh38
rs139681629 Chr.2: 100393575-100393575 on Build GRCh38
rs150991817 Chr.2: 100393353-100393353 on Build GRCh38
rs1017563783 Chr.2: 100393351-100393351 on Build GRCh38
rs780726705 Chr.2: 100393632-100393632 on Build GRCh38
rs376357045 Chr.2: 100393415-100393415 on Build GRCh38
rs375736968 Chr.2: 100393542-100393542 on Build GRCh38
rs1035002511 Chr.2: 100393680-100393680 on Build GRCh38
rs988936126 Chr.2: 100393400-100393400 on Build GRCh38
rs779396748 Chr.2: 100393662-100393662 on Build GRCh38
rs201758367 Chr.2: 100393618-100393618 on Build GRCh38
rs3748930 Chr.2: 100393620-100393620 on Build GRCh38
rs374282663 Chr.2: 100393722-100393722 on Build GRCh38
rs776053923 Chr.2: 100393461-100393461 on Build GRCh38
rs374730803 Chr.2: 100393576-100393576 on Build GRCh38
rs138468060 Chr.2: 100393759-100393759 on Build GRCh38
rs973733383 Chr.2: 100393747-100393747 on Build GRCh38
rs764927277 Chr.2: 100393739-100393739 on Build GRCh38
rs752195490 Chr.2: 100393522-100393522 on Build GRCh38
rs781340202 Chr.2: 100393671-100393671 on Build GRCh38
rs546199705 Chr.2: 100393771-100393771 on Build GRCh38
rs368455898 Chr.2: 100393748-100393748 on Build GRCh38
rs3748933 Chr.2: 100393521-100393521 on Build GRCh38
rs896648232 Chr.2: 100393535-100393535 on Build GRCh38
rs774446300 Chr.2: 100393478-100393478 on Build GRCh38
rs866437783 Chr.2: 100393417-100393420 on Build GRCh38
rs770471547 Chr.2: 100393564-100393564 on Build GRCh38
rs760578039 Chr.2: 100393591-100393591 on Build GRCh38
rs3748934 Chr.2: 100393446-100393446 on Build GRCh38
rs768356262 Chr.2: 100393355-100393355 on Build GRCh38
rs1001050979 Chr.2: 100393561-100393561 on Build GRCh38
rs776440097 Chr.2: 100393596-100393596 on Build GRCh38
rs770799123 Chr.2: 100393699-100393699 on Build GRCh38
rs778655889 Chr.2: 100393434-100393434 on Build GRCh38
rs749002484 Chr.2: 100393354-100393354 on Build GRCh38
rs763868069 Chr.2: 100393764-100393764 on Build GRCh38
rs147076164 Chr.2: 100393408-100393408 on Build GRCh38
rs118091549 Chr.2: 100393557-100393557 on Build GRCh38
rs756837256 Chr.2: 100393781-100393781 on Build GRCh38
rs759265127 Chr.2: 100393602-100393602 on Build GRCh38
rs140261702 Chr.2: 100393376-100393376 on Build GRCh38
rs937769970 Chr.2: 100393498-100393498 on Build GRCh38
rs747420216 Chr.2: 100393374-100393374 on Build GRCh38
rs201022725 Chr.2: 100393796-100393796 on Build GRCh38
rs12476171 Chr.2: 100393643-100393643 on Build GRCh38
rs570679582 Chr.2: 100393466-100393466 on Build GRCh38
rs945574036 Chr.2: 100393732-100393732 on Build GRCh38
rs756646964 Chr.2: 100393631-100393631 on Build GRCh38
rs748188413 Chr.2: 100393583-100393583 on Build GRCh38
rs3748932 Chr.2: 100393544-100393544 on Build GRCh38
rs753355704 Chr.2: 100393412-100393412 on Build GRCh38
rs200734208 Chr.2: 100393488-100393488 on Build GRCh38
rs766144204 Chr.2: 100393506-100393506 on Build GRCh38
rs750360245 Chr.2: 100393489-100393489 on Build GRCh38
rs370225437 Chr.2: 100393572-100393572 on Build GRCh38
rs201432165 Chr.2: 100393636-100393636 on Build GRCh38
rs781679368 Chr.2: 100393451-100393451 on Build GRCh38
rs751154094 Chr.2: 100393630-100393630 on Build GRCh38
rs112486773 Chr.2: 100393611-100393611 on Build GRCh38
rs748299923 Chr.2: 100393677-100393677 on Build GRCh38
rs146724275 Chr.2: 100393584-100393584 on Build GRCh38
rs768798605 Chr.2: 100393471-100393471 on Build GRCh38
rs937651746 Chr.2: 100393598-100393598 on Build GRCh38
rs763210815 Chr.2: 100393462-100393462 on Build GRCh38
rs373261971 Chr.2: 100393404-100393404 on Build GRCh38
rs149656297 Chr.2: 100393621-100393621 on Build GRCh38
rs766839235 Chr.2: 100393786-100393786 on Build GRCh38
rs768871548 Chr.2: 100393580-100393580 on Build GRCh38
rs367887748 Chr.2: 100393387-100393387 on Build GRCh38
rs3748931 Chr.2: 100393581-100393581 on Build GRCh38
rs770277532 Chr.2: 100393454-100393454 on Build GRCh38
rs772324034 Chr.2: 100393691-100393691 on Build GRCh38
rs761702531 Chr.2: 100393394-100393394 on Build GRCh38
rs777492199 Chr.2: 100393546-100393546 on Build GRCh38
rs760627598 Chr.2: 100393485-100393485 on Build GRCh38
rs1038070161 Chr.2: 100393470-100393470 on Build GRCh38
rs371549339 Chr.2: 100393797-100393797 on Build GRCh38
rs751685534 Chr.2: 100393390-100393390 on Build GRCh38
rs144032903 Chr.2: 100393592-100393592 on Build GRCh38
rs368177057 Chr.2: 100393702-100393702 on Build GRCh38
rs148920749 Chr.2: 100393407-100393407 on Build GRCh38
rs926287034 Chr.2: 100393595-100393595 on Build GRCh38
rs753676063 Chr.2: 100393507-100393507 on Build GRCh38
rs762993970 Chr.2: 100393788-100393789 on Build GRCh38
rs371045606 Chr.2: 100393423-100393423 on Build GRCh38
rs770056570 Chr.2: 100393385-100393385 on Build GRCh38
rs532803480 Chr.2: 100393706-100393706 on Build GRCh38
rs754291400 Chr.2: 100393791-100393791 on Build GRCh38
rs569364541 Chr.2: 100393377-100393377 on Build GRCh38
rs773239496 Chr.2: 100393693-100393693 on Build GRCh38
rs747117122 Chr.2: 100393694-100393694 on Build GRCh38
rs371699053 Chr.2: 100393398-100393398 on Build GRCh38
rs779521249 Chr.2: 100393342-100393342 on Build GRCh38
rs145728380 Chr.2: 100393359-100393359 on Build GRCh38
rs746168111 Chr.2: 100393452-100393452 on Build GRCh38
rs1042649406 Chr.2: 100393545-100393545 on Build GRCh38
rs951961263 Chr.2: 100393590-100393590 on Build GRCh38

View Full Product Details