Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101285989-101286488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF149
Entrez Gene ID
284996
Gene Name
ring finger protein 149
Gene Aliases
DNAPTP2
UniGene
Hs.142074
Gene Chromosome Location
Chr.2: 101271213-101308716 on Build GRCh38
Transcripts
NM_173647.3 XM_005263920.2 XM_005263921.3 XM_011510990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7449318 c.920C>A p.P307Q Chr.2: 101286121-101286121 on Build GRCh38
2980221 c.915A>G p.T305T Chr.2: 101286126-101286126 on Build GRCh38
1004567 c.878G>T p.R293I Chr.2: 101286163-101286163 on Build GRCh38
5779827 c.940A>T p.I314F Chr.2: 101286101-101286101 on Build GRCh38
6442105 c.920C>T p.P307L Chr.2: 101286121-101286121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047243312 Chr.2: 101286047-101286047 on Build GRCh38
rs769431448 Chr.2: 101286406-101286406 on Build GRCh38
rs1048010042 Chr.2: 101286451-101286451 on Build GRCh38
rs749483778 Chr.2: 101286052-101286052 on Build GRCh38
rs748203986 Chr.2: 101286056-101286056 on Build GRCh38
rs138622340 Chr.2: 101286296-101286296 on Build GRCh38
rs182220266 Chr.2: 101286055-101286055 on Build GRCh38
rs1041186650 Chr.2: 101286445-101286445 on Build GRCh38
rs925278120 Chr.2: 101286301-101286301 on Build GRCh38
rs778003080 Chr.2: 101286215-101286215 on Build GRCh38
rs768780003 Chr.2: 101286054-101286054 on Build GRCh38
rs371034904 Chr.2: 101286393-101286393 on Build GRCh38
rs748437446 Chr.2: 101286204-101286204 on Build GRCh38
rs369893283 Chr.2: 101286033-101286033 on Build GRCh38
rs113430267 Chr.2: 101286065-101286065 on Build GRCh38
rs993109322 Chr.2: 101286239-101286239 on Build GRCh38
rs770722293 Chr.2: 101286094-101286094 on Build GRCh38
rs375868649 Chr.2: 101286223-101286223 on Build GRCh38
rs917990184 Chr.2: 101286253-101286253 on Build GRCh38
rs772219297 Chr.2: 101286209-101286209 on Build GRCh38
rs900117749 Chr.2: 101286462-101286462 on Build GRCh38
rs760952554 Chr.2: 101286237-101286237 on Build GRCh38
rs149301037 Chr.2: 101286409-101286409 on Build GRCh38
rs773267391 Chr.2: 101286078-101286078 on Build GRCh38
rs762513006 Chr.2: 101286413-101286413 on Build GRCh38
rs1032739789 Chr.2: 101286192-101286192 on Build GRCh38
rs944246557 Chr.2: 101286434-101286434 on Build GRCh38
rs111716123 Chr.2: 101286278-101286278 on Build GRCh38
rs926561275 Chr.2: 101286308-101286308 on Build GRCh38
rs996833732 Chr.2: 101286190-101286190 on Build GRCh38
rs368810286 Chr.2: 101286073-101286073 on Build GRCh38
rs759426109 Chr.2: 101286101-101286101 on Build GRCh38
rs779273464 Chr.2: 101286203-101286203 on Build GRCh38
rs1020794978 Chr.2: 101286248-101286248 on Build GRCh38
rs201600274 Chr.2: 101286154-101286154 on Build GRCh38
rs376543893 Chr.2: 101286071-101286071 on Build GRCh38
rs200499158 Chr.2: 101286119-101286119 on Build GRCh38
rs764838490 Chr.2: 101286109-101286109 on Build GRCh38
rs985702651 Chr.2: 101286399-101286399 on Build GRCh38
rs372551230 Chr.2: 101286131-101286131 on Build GRCh38
rs199638534 Chr.2: 101286126-101286126 on Build GRCh38
rs746917191 Chr.2: 101286219-101286219 on Build GRCh38
rs776315156 Chr.2: 101286097-101286097 on Build GRCh38
rs1010038323 Chr.2: 101286218-101286218 on Build GRCh38
rs929735837 Chr.2: 101286420-101286420 on Build GRCh38
rs911642065 Chr.2: 101286419-101286419 on Build GRCh38
rs978088111 Chr.2: 101286297-101286297 on Build GRCh38
rs762440660 Chr.2: 101286123-101286123 on Build GRCh38
rs773623895 Chr.2: 101286141-101286141 on Build GRCh38
rs970508799 Chr.2: 101286252-101286252 on Build GRCh38
rs760540896 Chr.2: 101286081-101286081 on Build GRCh38
rs920228152 Chr.2: 101286407-101286407 on Build GRCh38
rs754127520 Chr.2: 101286172-101286172 on Build GRCh38
rs756446924 Chr.2: 101286031-101286031 on Build GRCh38
rs550550778 Chr.2: 101286259-101286259 on Build GRCh38
rs750865908 Chr.2: 101286130-101286130 on Build GRCh38
rs755407095 Chr.2: 101286188-101286188 on Build GRCh38

View Full Product Details