Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101293930-101294437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF149
Entrez Gene ID
284996
Gene Name
ring finger protein 149
Gene Aliases
DNAPTP2
UniGene
Hs.142074
Gene Chromosome Location
Chr.2: 101271213-101308716 on Build GRCh38
Transcripts
NM_173647.3 XM_005263920.2 XM_005263921.3 XM_011510990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7309600 c.712-7A>T p.? Chr.2: 101294089-101294089 on Build GRCh38
6424542 c.767A>G p.K256R Chr.2: 101294027-101294027 on Build GRCh38
5697057 c.729T>C p.T243T Chr.2: 101294065-101294065 on Build GRCh38
4083804 c.744C>T p.G248G Chr.2: 101294050-101294050 on Build GRCh38
1004569 c.744C>A p.G248G Chr.2: 101294050-101294050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775160357 Chr.2: 101293984-101293984 on Build GRCh38
rs866275650 Chr.2: 101294278-101294278 on Build GRCh38
rs554667988 Chr.2: 101294217-101294217 on Build GRCh38
rs760313147 Chr.2: 101294024-101294024 on Build GRCh38
rs770622985 Chr.2: 101294111-101294111 on Build GRCh38
rs145478309 Chr.2: 101294377-101294377 on Build GRCh38
rs778772367 Chr.2: 101294091-101294091 on Build GRCh38
rs774653913 Chr.2: 101294348-101294348 on Build GRCh38
rs984572924 Chr.2: 101293960-101293960 on Build GRCh38
rs756223346 Chr.2: 101294074-101294074 on Build GRCh38
rs746515772 Chr.2: 101294104-101294104 on Build GRCh38
rs560027141 Chr.2: 101293956-101293956 on Build GRCh38
rs1015134503 Chr.2: 101294258-101294258 on Build GRCh38
rs894013499 Chr.2: 101294359-101294359 on Build GRCh38
rs568823483 Chr.2: 101294208-101294208 on Build GRCh38
rs1056001021 Chr.2: 101294242-101294242 on Build GRCh38
rs770461628 Chr.2: 101293958-101293958 on Build GRCh38
rs369587252 Chr.2: 101294083-101294083 on Build GRCh38
rs147592390 Chr.2: 101293962-101293962 on Build GRCh38
rs745755681 Chr.2: 101293988-101293988 on Build GRCh38
rs764218636 Chr.2: 101293976-101293976 on Build GRCh38
rs566755004 Chr.2: 101293987-101293987 on Build GRCh38
rs371933125 Chr.2: 101294097-101294097 on Build GRCh38
rs947091812 Chr.2: 101294406-101294406 on Build GRCh38
rs771702696 Chr.2: 101293982-101293986 on Build GRCh38
rs773858321 Chr.2: 101294012-101294012 on Build GRCh38
rs976920120 Chr.2: 101294401-101294401 on Build GRCh38
rs144281444 Chr.2: 101294244-101294244 on Build GRCh38
rs776207284 Chr.2: 101294050-101294050 on Build GRCh38
rs137972787 Chr.2: 101294078-101294078 on Build GRCh38
rs141301816 Chr.2: 101294063-101294063 on Build GRCh38
rs902777660 Chr.2: 101294145-101294145 on Build GRCh38
rs768502800 Chr.2: 101294009-101294009 on Build GRCh38
rs781340540 Chr.2: 101293985-101293985 on Build GRCh38
rs745681216 Chr.2: 101293977-101293977 on Build GRCh38
rs188746193 Chr.2: 101294113-101294113 on Build GRCh38
rs367793540 Chr.2: 101293990-101293990 on Build GRCh38
rs112591942 Chr.2: 101293992-101293992 on Build GRCh38
rs751866999 Chr.2: 101293977-101293977 on Build GRCh38
rs754671445 Chr.2: 101294090-101294090 on Build GRCh38
rs138575347 Chr.2: 101294014-101294014 on Build GRCh38
rs375296169 Chr.2: 101294103-101294103 on Build GRCh38
rs552209646 Chr.2: 101294173-101294173 on Build GRCh38
rs560942196 Chr.2: 101294051-101294051 on Build GRCh38
rs528780271 Chr.2: 101293961-101293961 on Build GRCh38
rs553283362 Chr.2: 101294313-101294313 on Build GRCh38
rs377135713 Chr.2: 101294275-101294275 on Build GRCh38
rs772808658 Chr.2: 101294019-101294019 on Build GRCh38
rs923207075 Chr.2: 101294332-101294332 on Build GRCh38
rs763563082 Chr.2: 101294119-101294119 on Build GRCh38
rs185034388 Chr.2: 101294070-101294070 on Build GRCh38
rs771416783 Chr.2: 101294018-101294018 on Build GRCh38
rs13400007 Chr.2: 101294370-101294370 on Build GRCh38
rs78101413 Chr.2: 101294086-101294086 on Build GRCh38
rs1000146863 Chr.2: 101294170-101294170 on Build GRCh38
rs774803341 Chr.2: 101294134-101294134 on Build GRCh38
rs537958013 Chr.2: 101294211-101294211 on Build GRCh38
rs745781528 Chr.2: 101294233-101294233 on Build GRCh38
rs376238702 Chr.2: 101293980-101293980 on Build GRCh38
rs751669298 Chr.2: 101294061-101294061 on Build GRCh38
rs566804274 Chr.2: 101294069-101294069 on Build GRCh38
rs371001671 Chr.2: 101294344-101294344 on Build GRCh38
rs746474607 Chr.2: 101294104-101294105 on Build GRCh38
rs779959151 Chr.2: 101294080-101294080 on Build GRCh38
rs1007133501 Chr.2: 101294371-101294371 on Build GRCh38
rs192307127 Chr.2: 101294309-101294309 on Build GRCh38
rs761941100 Chr.2: 101294374-101294374 on Build GRCh38
rs758875925 Chr.2: 101293973-101293973 on Build GRCh38
rs965497495 Chr.2: 101294397-101294397 on Build GRCh38
rs999909869 Chr.2: 101294310-101294310 on Build GRCh38
rs770394101 Chr.2: 101294038-101294038 on Build GRCh38
rs1035276204 Chr.2: 101294186-101294186 on Build GRCh38
rs529934469 Chr.2: 101294055-101294055 on Build GRCh38
rs377721120 Chr.2: 101294122-101294122 on Build GRCh38
rs778615703 Chr.2: 101293968-101293972 on Build GRCh38
rs1012188698 Chr.2: 101294214-101294214 on Build GRCh38
rs747712846 Chr.2: 101294096-101294096 on Build GRCh38

View Full Product Details