Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102375773-102376298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18R1
Entrez Gene ID
8809
Gene Name
interleukin 18 receptor 1
Gene Aliases
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
UniGene
Hs.469521
Gene Chromosome Location
Chr.2: 102355817-102398777 on Build GRCh38
Transcripts
NM_001282399.1 NM_003855.3 XM_005264039.3 XM_005264040.3 XM_011512099.1 XM_017005181.1 XM_017005182.1 XM_017005183.1 XM_017005184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325541 c.475A>G p.K159E Chr.2: 102375913-102375913 on Build GRCh38
4498634 c.524C>T p.A175V Chr.2: 102375962-102375962 on Build GRCh38
1004664 c.526G>A p.E176K Chr.2: 102375964-102375964 on Build GRCh38
3836457 c.500A>C p.K167T Chr.2: 102375938-102375938 on Build GRCh38
4903110 c.510G>A p.T170T Chr.2: 102375948-102375948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765265118 Chr.2: 102375878-102375878 on Build GRCh38
rs769977470 Chr.2: 102376077-102376077 on Build GRCh38
rs771280406 Chr.2: 102376065-102376065 on Build GRCh38
rs780349962 Chr.2: 102375882-102375882 on Build GRCh38
rs1012411089 Chr.2: 102376116-102376116 on Build GRCh38
rs760882559 Chr.2: 102375859-102375859 on Build GRCh38
rs944017011 Chr.2: 102376014-102376014 on Build GRCh38
rs573316949 Chr.2: 102375993-102375993 on Build GRCh38
rs953854980 Chr.2: 102376216-102376216 on Build GRCh38
rs961336614 Chr.2: 102376196-102376196 on Build GRCh38
rs757161434 Chr.2: 102375983-102375983 on Build GRCh38
rs758425072 Chr.2: 102375901-102375901 on Build GRCh38
rs370953101 Chr.2: 102375816-102375816 on Build GRCh38
rs755777769 Chr.2: 102376106-102376106 on Build GRCh38
rs376667430 Chr.2: 102375909-102375909 on Build GRCh38
rs375166889 Chr.2: 102376098-102376098 on Build GRCh38
rs1044107585 Chr.2: 102376172-102376172 on Build GRCh38
rs868607137 Chr.2: 102376245-102376245 on Build GRCh38
rs1019695660 Chr.2: 102376227-102376227 on Build GRCh38
rs748583457 Chr.2: 102375934-102375934 on Build GRCh38
rs780950445 Chr.2: 102375914-102375914 on Build GRCh38
rs35292939 Chr.2: 102375994-102375994 on Build GRCh38
rs975164641 Chr.2: 102376047-102376047 on Build GRCh38
rs755723079 Chr.2: 102375996-102375996 on Build GRCh38
rs779440932 Chr.2: 102375930-102375930 on Build GRCh38
rs772545563 Chr.2: 102376032-102376032 on Build GRCh38
rs978585907 Chr.2: 102376252-102376252 on Build GRCh38
rs115326837 Chr.2: 102375947-102375947 on Build GRCh38
rs776875280 Chr.2: 102375961-102375961 on Build GRCh38
rs750247355 Chr.2: 102376105-102376105 on Build GRCh38
rs1037827094 Chr.2: 102375804-102375804 on Build GRCh38
rs540061267 Chr.2: 102376167-102376167 on Build GRCh38
rs779621024 Chr.2: 102376115-102376115 on Build GRCh38
rs755123193 Chr.2: 102375869-102375869 on Build GRCh38
rs201270046 Chr.2: 102375981-102375981 on Build GRCh38
rs200648833 Chr.2: 102375867-102375867 on Build GRCh38
rs199966642 Chr.2: 102375963-102375963 on Build GRCh38
rs776874442 Chr.2: 102376070-102376070 on Build GRCh38
rs777645316 Chr.2: 102375907-102375907 on Build GRCh38
rs759738406 Chr.2: 102376072-102376072 on Build GRCh38
rs756189335 Chr.2: 102376185-102376185 on Build GRCh38
rs879242072 Chr.2: 102375858-102375858 on Build GRCh38
rs995400490 Chr.2: 102376205-102376205 on Build GRCh38
rs772633602 Chr.2: 102375938-102375938 on Build GRCh38
rs752633018 Chr.2: 102375888-102375888 on Build GRCh38
rs570198678 Chr.2: 102376217-102376217 on Build GRCh38
rs912689550 Chr.2: 102376007-102376007 on Build GRCh38
rs559291188 Chr.2: 102375964-102375964 on Build GRCh38
rs906509924 Chr.2: 102375850-102375850 on Build GRCh38
rs761074985 Chr.2: 102375948-102375948 on Build GRCh38
rs766644366 Chr.2: 102375865-102375865 on Build GRCh38
rs201629380 Chr.2: 102375960-102375960 on Build GRCh38
rs2270297 Chr.2: 102376215-102376215 on Build GRCh38
rs748855104 Chr.2: 102376009-102376009 on Build GRCh38
rs754198537 Chr.2: 102375966-102375966 on Build GRCh38
rs555264984 Chr.2: 102375840-102375840 on Build GRCh38
rs906981400 Chr.2: 102376177-102376177 on Build GRCh38
rs769296603 Chr.2: 102375928-102375928 on Build GRCh38
rs1015913836 Chr.2: 102375824-102375824 on Build GRCh38
rs745536546 Chr.2: 102375927-102375927 on Build GRCh38
rs371796705 Chr.2: 102376035-102376035 on Build GRCh38
rs755124320 Chr.2: 102376086-102376087 on Build GRCh38
rs756959835 Chr.2: 102375910-102375910 on Build GRCh38
rs577427311 Chr.2: 102375800-102375800 on Build GRCh38
rs79488820 Chr.2: 102375845-102375845 on Build GRCh38
rs1015237002 Chr.2: 102376193-102376193 on Build GRCh38
rs779608094 Chr.2: 102376003-102376003 on Build GRCh38
rs747544780 Chr.2: 102376059-102376059 on Build GRCh38
rs762984691 Chr.2: 102375965-102375965 on Build GRCh38
rs775702318 Chr.2: 102376097-102376097 on Build GRCh38
rs781544952 Chr.2: 102376098-102376099 on Build GRCh38
rs751972582 Chr.2: 102376041-102376042 on Build GRCh38
rs201688186 Chr.2: 102376103-102376103 on Build GRCh38
rs374965790 Chr.2: 102375803-102375803 on Build GRCh38
rs899237391 Chr.2: 102375817-102375817 on Build GRCh38
rs184754953 Chr.2: 102376052-102376052 on Build GRCh38

View Full Product Details