Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102513964-102514508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A4
Entrez Gene ID
389015
Gene Name
solute carrier family 9 member A4
Gene Aliases
NHE4
UniGene
Hs.447686
Gene Chromosome Location
Chr.2: 102473282-102533974 on Build GRCh38
Transcripts
NM_001011552.3 XM_011511158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1690850 c.1656G>A p.M552I Chr.2: 102514186-102514186 on Build GRCh38
3379717 c.1668C>G p.I556M Chr.2: 102514198-102514198 on Build GRCh38
4083891 c.1681A>G p.T561A Chr.2: 102514211-102514211 on Build GRCh38
4727858 c.1628T>A p.I543N Chr.2: 102514158-102514158 on Build GRCh38
273273 c.1584C>T p.Y528Y Chr.2: 102514114-102514114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372761774 Chr.2: 102514196-102514196 on Build GRCh38
rs763865670 Chr.2: 102514047-102514047 on Build GRCh38
rs549805693 Chr.2: 102514199-102514199 on Build GRCh38
rs761375825 Chr.2: 102514134-102514134 on Build GRCh38
rs562164795 Chr.2: 102514161-102514161 on Build GRCh38
rs768254195 Chr.2: 102514349-102514349 on Build GRCh38
rs773486179 Chr.2: 102514482-102514482 on Build GRCh38
rs921209281 Chr.2: 102514365-102514365 on Build GRCh38
rs984725052 Chr.2: 102514031-102514031 on Build GRCh38
rs567249685 Chr.2: 102514142-102514142 on Build GRCh38
rs941043976 Chr.2: 102514329-102514329 on Build GRCh38
rs565247006 Chr.2: 102513989-102513989 on Build GRCh38
rs571366874 Chr.2: 102514335-102514335 on Build GRCh38
rs753261055 Chr.2: 102514249-102514249 on Build GRCh38
rs992350486 Chr.2: 102514233-102514233 on Build GRCh38
rs200327384 Chr.2: 102514135-102514135 on Build GRCh38
rs780148420 Chr.2: 102514194-102514194 on Build GRCh38
rs1002609958 Chr.2: 102514167-102514167 on Build GRCh38
rs923349582 Chr.2: 102514381-102514381 on Build GRCh38
rs763298299 Chr.2: 102514254-102514254 on Build GRCh38
rs768467080 Chr.2: 102514297-102514297 on Build GRCh38
rs761412983 Chr.2: 102514212-102514212 on Build GRCh38
rs745963544 Chr.2: 102514178-102514178 on Build GRCh38
rs537483513 Chr.2: 102514146-102514146 on Build GRCh38
rs777861316 Chr.2: 102514081-102514081 on Build GRCh38
rs745830954 Chr.2: 102514110-102514110 on Build GRCh38
rs909623579 Chr.2: 102514322-102514322 on Build GRCh38
rs762739690 Chr.2: 102514046-102514046 on Build GRCh38
rs770973156 Chr.2: 102514084-102514084 on Build GRCh38
rs141256616 Chr.2: 102514099-102514099 on Build GRCh38
rs375740273 Chr.2: 102514217-102514217 on Build GRCh38
rs899713293 Chr.2: 102514036-102514036 on Build GRCh38
rs547607930 Chr.2: 102514050-102514050 on Build GRCh38
rs188427617 Chr.2: 102514023-102514023 on Build GRCh38
rs756278845 Chr.2: 102514283-102514283 on Build GRCh38
rs201284506 Chr.2: 102514010-102514010 on Build GRCh38
rs950841567 Chr.2: 102514305-102514305 on Build GRCh38
rs933872737 Chr.2: 102514409-102514409 on Build GRCh38
rs866315288 Chr.2: 102514224-102514224 on Build GRCh38
rs143912526 Chr.2: 102514149-102514149 on Build GRCh38
rs764402507 Chr.2: 102514265-102514265 on Build GRCh38
rs267598812 Chr.2: 102514164-102514164 on Build GRCh38
rs766853899 Chr.2: 102514053-102514053 on Build GRCh38
rs199497876 Chr.2: 102514163-102514163 on Build GRCh38
rs750464838 Chr.2: 102514280-102514280 on Build GRCh38
rs769818324 Chr.2: 102514189-102514189 on Build GRCh38
rs377145509 Chr.2: 102514057-102514057 on Build GRCh38
rs772550155 Chr.2: 102514221-102514221 on Build GRCh38
rs751044793 Chr.2: 102514049-102514049 on Build GRCh38
rs199653058 Chr.2: 102514285-102514285 on Build GRCh38
rs762876824 Chr.2: 102514119-102514119 on Build GRCh38
rs769686433 Chr.2: 102514114-102514114 on Build GRCh38
rs749308521 Chr.2: 102514286-102514286 on Build GRCh38
rs145843156 Chr.2: 102514244-102514244 on Build GRCh38
rs982566995 Chr.2: 102514306-102514306 on Build GRCh38
rs200313519 Chr.2: 102514414-102514414 on Build GRCh38
rs745406600 Chr.2: 102514198-102514199 on Build GRCh38
rs774050036 Chr.2: 102514201-102514201 on Build GRCh38
rs1020057116 Chr.2: 102514168-102514168 on Build GRCh38
rs1034607886 Chr.2: 102514208-102514208 on Build GRCh38
rs933406653 Chr.2: 102514434-102514434 on Build GRCh38
rs778463385 Chr.2: 102514116-102514117 on Build GRCh38
rs758578804 Chr.2: 102514076-102514076 on Build GRCh38
rs773891191 Chr.2: 102514131-102514131 on Build GRCh38
rs1051009385 Chr.2: 102514419-102514419 on Build GRCh38
rs760181512 Chr.2: 102514228-102514228 on Build GRCh38
rs779290685 Chr.2: 102514061-102514061 on Build GRCh38
rs757370047 Chr.2: 102514276-102514276 on Build GRCh38
rs148398809 Chr.2: 102514399-102514399 on Build GRCh38
rs372330986 Chr.2: 102514054-102514054 on Build GRCh38
rs752991505 Chr.2: 102514145-102514145 on Build GRCh38
rs369035557 Chr.2: 102514064-102514064 on Build GRCh38
rs751779208 Chr.2: 102514267-102514267 on Build GRCh38
rs371685381 Chr.2: 102514336-102514336 on Build GRCh38
rs892325518 Chr.2: 102514457-102514457 on Build GRCh38
rs781505839 Chr.2: 102514277-102514277 on Build GRCh38
rs1033945650 Chr.2: 102514175-102514175 on Build GRCh38
rs751794126 Chr.2: 102514159-102514159 on Build GRCh38
rs60883579 Chr.2: 102514138-102514138 on Build GRCh38
rs200443203 Chr.2: 102514129-102514129 on Build GRCh38
rs747050488 Chr.2: 102514083-102514083 on Build GRCh38
rs570341669 Chr.2: 102514246-102514246 on Build GRCh38
rs775373011 Chr.2: 102514118-102514118 on Build GRCh38

View Full Product Details