Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102717732-102718264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD9
Entrez Gene ID
84804
Gene Name
major facilitator superfamily domain containing 9
Gene Aliases
-
UniGene
Hs.590898
Gene Chromosome Location
Chr.2: 102713867-102736888 on Build GRCh38
Transcripts
NM_001322080.1 NM_001322081.1 NM_032718.4 NR_136173.1 NR_136174.1 XM_011512005.2 XM_011512006.2 XM_011512007.2

SNP Detail

SNP ID Chromosome Location
rs192301842 Chr.2: 102718057-102718057 on Build GRCh38
rs1013228340 Chr.2: 102717765-102717765 on Build GRCh38
rs187063684 Chr.2: 102717976-102717976 on Build GRCh38
rs900684906 Chr.2: 102718175-102718175 on Build GRCh38
rs957265002 Chr.2: 102718144-102718144 on Build GRCh38
rs144326565 Chr.2: 102718244-102718244 on Build GRCh38
rs1036415713 Chr.2: 102718081-102718081 on Build GRCh38
rs544784532 Chr.2: 102717947-102717947 on Build GRCh38
rs927105596 Chr.2: 102718221-102718221 on Build GRCh38
rs746935047 Chr.2: 102718045-102718045 on Build GRCh38
rs372064666 Chr.2: 102718234-102718234 on Build GRCh38
rs771046019 Chr.2: 102718066-102718066 on Build GRCh38
rs555533742 Chr.2: 102717855-102717855 on Build GRCh38
rs568034014 Chr.2: 102718241-102718241 on Build GRCh38
rs971132670 Chr.2: 102718183-102718183 on Build GRCh38
rs576013129 Chr.2: 102717805-102717805 on Build GRCh38
rs182117118 Chr.2: 102718072-102718072 on Build GRCh38
rs988441448 Chr.2: 102718159-102718159 on Build GRCh38
rs1030722392 Chr.2: 102717869-102717869 on Build GRCh38
rs913318942 Chr.2: 102717924-102717924 on Build GRCh38
rs546577818 Chr.2: 102718105-102718105 on Build GRCh38
rs1029559050 Chr.2: 102718107-102718107 on Build GRCh38
rs975032920 Chr.2: 102717861-102717861 on Build GRCh38
rs1010203922 Chr.2: 102718233-102718233 on Build GRCh38
rs988578628 Chr.2: 102717954-102717954 on Build GRCh38
rs997608000 Chr.2: 102717813-102717813 on Build GRCh38
rs937235139 Chr.2: 102718005-102718005 on Build GRCh38
rs1026006128 Chr.2: 102717766-102717766 on Build GRCh38
rs925154750 Chr.2: 102717978-102717978 on Build GRCh38
rs954982414 Chr.2: 102718208-102718208 on Build GRCh38
rs752385172 Chr.2: 102717780-102717780 on Build GRCh38
rs901930093 Chr.2: 102718096-102718096 on Build GRCh38
rs374089170 Chr.2: 102717832-102717832 on Build GRCh38
rs889404571 Chr.2: 102718131-102718131 on Build GRCh38
rs1029236118 Chr.2: 102717826-102717826 on Build GRCh38
rs1032572360 Chr.2: 102718143-102718143 on Build GRCh38
rs56038403 Chr.2: 102718100-102718100 on Build GRCh38
rs1030577400 Chr.2: 102718201-102718201 on Build GRCh38
rs1055170620 Chr.2: 102717761-102717761 on Build GRCh38
rs1001538461 Chr.2: 102718134-102718134 on Build GRCh38
rs1005975921 Chr.2: 102718111-102718111 on Build GRCh38
rs1006341681 Chr.2: 102717804-102717804 on Build GRCh38
rs367961325 Chr.2: 102718058-102718058 on Build GRCh38
rs758188537 Chr.2: 102717985-102717985 on Build GRCh38
rs988847532 Chr.2: 102717909-102717909 on Build GRCh38
rs761971846 Chr.2: 102717763-102717763 on Build GRCh38
rs1038243860 Chr.2: 102718168-102718168 on Build GRCh38
rs866188048 Chr.2: 102717980-102717980 on Build GRCh38
rs73001597 Chr.2: 102717957-102717957 on Build GRCh38
rs777466681 Chr.2: 102718000-102718000 on Build GRCh38
rs543045176 Chr.2: 102717886-102717886 on Build GRCh38
rs996780348 Chr.2: 102718182-102718182 on Build GRCh38
rs927271027 Chr.2: 102717988-102717988 on Build GRCh38
rs981294266 Chr.2: 102718217-102718217 on Build GRCh38

View Full Product Details