Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127304720-127305220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K2
Entrez Gene ID
10746
Gene Name
mitogen-activated protein kinase kinase kinase 2
Gene Aliases
MEKK2, MEKK2B
UniGene
Hs.145605
Gene Chromosome Location
Chr.2: 127298669-127343229 on Build GRCh38
Transcripts
NM_006609.4

SNP Detail

SNP ID Chromosome Location
rs752986594 Chr.2: 127305016-127305017 on Build GRCh38
rs571814244 Chr.2: 127304955-127304955 on Build GRCh38
rs1034044799 Chr.2: 127304843-127304843 on Build GRCh38
rs1027664012 Chr.2: 127305081-127305081 on Build GRCh38
rs781508258 Chr.2: 127304853-127304853 on Build GRCh38
rs868564262 Chr.2: 127305033-127305033 on Build GRCh38
rs534686668 Chr.2: 127304817-127304817 on Build GRCh38
rs958796951 Chr.2: 127305090-127305090 on Build GRCh38
rs1041368434 Chr.2: 127304861-127304861 on Build GRCh38
rs766936253 Chr.2: 127304911-127304912 on Build GRCh38
rs935998696 Chr.2: 127305024-127305024 on Build GRCh38
rs573411082 Chr.2: 127305185-127305185 on Build GRCh38
rs566126010 Chr.2: 127304794-127304794 on Build GRCh38
rs973380804 Chr.2: 127305068-127305068 on Build GRCh38
rs397944539 Chr.2: 127305089-127305090 on Build GRCh38
rs201253175 Chr.2: 127305080-127305081 on Build GRCh38
rs971707795 Chr.2: 127304829-127304829 on Build GRCh38
rs937138888 Chr.2: 127304833-127304833 on Build GRCh38
rs977778516 Chr.2: 127305181-127305181 on Build GRCh38
rs951876299 Chr.2: 127304845-127304845 on Build GRCh38
rs1001608577 Chr.2: 127304834-127304834 on Build GRCh38
rs991260327 Chr.2: 127305124-127305124 on Build GRCh38
rs557086812 Chr.2: 127305026-127305026 on Build GRCh38
rs1010412245 Chr.2: 127304778-127304778 on Build GRCh38
rs139141625 Chr.2: 127304970-127304970 on Build GRCh38
rs1054459088 Chr.2: 127304869-127304869 on Build GRCh38
rs770050295 Chr.2: 127304876-127304876 on Build GRCh38
rs879887911 Chr.2: 127304972-127304972 on Build GRCh38
rs867048227 Chr.2: 127305126-127305126 on Build GRCh38
rs946189933 Chr.2: 127305129-127305129 on Build GRCh38
rs769651944 Chr.2: 127304908-127304908 on Build GRCh38
rs576984886 Chr.2: 127304816-127304816 on Build GRCh38
rs62157548 Chr.2: 127304867-127304867 on Build GRCh38
rs770386880 Chr.2: 127305162-127305162 on Build GRCh38
rs907646463 Chr.2: 127304858-127304858 on Build GRCh38
rs576014115 Chr.2: 127305143-127305143 on Build GRCh38
rs961811934 Chr.2: 127304863-127304863 on Build GRCh38
rs762624189 Chr.2: 127305018-127305020 on Build GRCh38
rs1015363568 Chr.2: 127304777-127304777 on Build GRCh38
rs917096811 Chr.2: 127305133-127305133 on Build GRCh38
rs931956808 Chr.2: 127304948-127304948 on Build GRCh38
rs907432747 Chr.2: 127305145-127305145 on Build GRCh38
rs923170016 Chr.2: 127304877-127304877 on Build GRCh38
rs887425494 Chr.2: 127305188-127305188 on Build GRCh38

View Full Product Details