Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131148300-131148801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHB2
Entrez Gene ID
55041
Gene Name
pleckstrin homology domain containing B2
Gene Aliases
EVT2
UniGene
Hs.469944
Gene Chromosome Location
Chr.2: 131104847-131149852 on Build GRCh38
Transcripts
NM_001100623.1 NM_001267062.1 NM_001267063.1 NM_001267064.1 NM_001267065.1 NM_001267066.1 NM_001267067.1 NM_001267068.1 NM_001309448.1 NM_001309450.1 NM_001309451.1 NM_001309452.1 NM_017958.2 XM_011511385.2 XM_011511386.2 XM_017004408.1 XM_017004409.1 XM_017004410.1 XM_017004411.1 XM_017004412.1 XM_017004413.1

SNP Detail

SNP ID Chromosome Location
rs755083002 Chr.2: 131148639-131148639 on Build GRCh38
rs1000931723 Chr.2: 131148743-131148743 on Build GRCh38
rs1012017702 Chr.2: 131148477-131148477 on Build GRCh38
rs959269389 Chr.2: 131148357-131148357 on Build GRCh38
rs990610562 Chr.2: 131148361-131148361 on Build GRCh38
rs917855660 Chr.2: 131148385-131148385 on Build GRCh38
rs781118015 Chr.2: 131148742-131148742 on Build GRCh38
rs943558458 Chr.2: 131148660-131148660 on Build GRCh38
rs1035546615 Chr.2: 131148643-131148644 on Build GRCh38
rs981421358 Chr.2: 131148513-131148513 on Build GRCh38
rs532451329 Chr.2: 131148612-131148612 on Build GRCh38
rs145318597 Chr.2: 131148363-131148363 on Build GRCh38
rs919241390 Chr.2: 131148554-131148554 on Build GRCh38
rs533030130 Chr.2: 131148544-131148548 on Build GRCh38
rs1049112084 Chr.2: 131148604-131148604 on Build GRCh38
rs16856546 Chr.2: 131148677-131148677 on Build GRCh38
rs755920400 Chr.2: 131148668-131148676 on Build GRCh38
rs541545095 Chr.2: 131148661-131148661 on Build GRCh38
rs915667852 Chr.2: 131148675-131148675 on Build GRCh38
rs372837800 Chr.2: 131148760-131148760 on Build GRCh38
rs983821844 Chr.2: 131148746-131148746 on Build GRCh38
rs137944543 Chr.2: 131148593-131148593 on Build GRCh38
rs959363683 Chr.2: 131148774-131148774 on Build GRCh38
rs902107369 Chr.2: 131148663-131148663 on Build GRCh38
rs940913954 Chr.2: 131148336-131148336 on Build GRCh38
rs76879872 Chr.2: 131148458-131148458 on Build GRCh38
rs977317913 Chr.2: 131148640-131148640 on Build GRCh38
rs1036581519 Chr.2: 131148345-131148345 on Build GRCh38
rs1032130429 Chr.2: 131148755-131148755 on Build GRCh38
rs757207598 Chr.2: 131148719-131148719 on Build GRCh38
rs765273230 Chr.2: 131148670-131148670 on Build GRCh38
rs113759360 Chr.2: 131148729-131148729 on Build GRCh38

View Full Product Details