Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 144007441-144007952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
UniGene
Hs.44780
Gene Chromosome Location
Chr.2: 143945787-144333092 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284235.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311677 c.522-1G>T p.? Chr.2: 144007536-144007536 on Build GRCh38
1399798 c.601G>A p.A201T Chr.2: 144007456-144007456 on Build GRCh38
5966133 c.528G>T p.M176I Chr.2: 144007529-144007529 on Build GRCh38
4085275 c.564C>T p.L188L Chr.2: 144007493-144007493 on Build GRCh38
3567340 c.584G>A p.G195D Chr.2: 144007473-144007473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771003854 Chr.2: 144007556-144007556 on Build GRCh38
rs761380884 Chr.2: 144007476-144007476 on Build GRCh38
rs985639017 Chr.2: 144007820-144007820 on Build GRCh38
rs368662987 Chr.2: 144007588-144007588 on Build GRCh38
rs781656112 Chr.2: 144007530-144007530 on Build GRCh38
rs112818590 Chr.2: 144007667-144007667 on Build GRCh38
rs751804010 Chr.2: 144007504-144007504 on Build GRCh38
rs775433446 Chr.2: 144007817-144007817 on Build GRCh38
rs912701163 Chr.2: 144007845-144007845 on Build GRCh38
rs116416428 Chr.2: 144007493-144007493 on Build GRCh38
rs768776381 Chr.2: 144007571-144007571 on Build GRCh38
rs569447683 Chr.2: 144007807-144007807 on Build GRCh38
rs537168547 Chr.2: 144007810-144007810 on Build GRCh38
rs749549218 Chr.2: 144007554-144007554 on Build GRCh38
rs3806481 Chr.2: 144007562-144007562 on Build GRCh38
rs931236573 Chr.2: 144007908-144007908 on Build GRCh38
rs372473070 Chr.2: 144007531-144007531 on Build GRCh38
rs770508572 Chr.2: 144007502-144007503 on Build GRCh38
rs780478505 Chr.2: 144007494-144007494 on Build GRCh38
rs147157212 Chr.2: 144007469-144007469 on Build GRCh38
rs142127850 Chr.2: 144007488-144007488 on Build GRCh38
rs534311269 Chr.2: 144007926-144007926 on Build GRCh38
rs1012725653 Chr.2: 144007702-144007702 on Build GRCh38
rs114317549 Chr.2: 144007900-144007900 on Build GRCh38
rs993173997 Chr.2: 144007619-144007619 on Build GRCh38
rs991657817 Chr.2: 144007705-144007705 on Build GRCh38
rs142813460 Chr.2: 144007858-144007858 on Build GRCh38
rs192415313 Chr.2: 144007653-144007653 on Build GRCh38
rs777966408 Chr.2: 144007560-144007560 on Build GRCh38
rs986860910 Chr.2: 144007825-144007825 on Build GRCh38
rs775004492 Chr.2: 144007543-144007543 on Build GRCh38
rs975868089 Chr.2: 144007864-144007864 on Build GRCh38
rs202049795 Chr.2: 144007468-144007468 on Build GRCh38
rs977666112 Chr.2: 144007763-144007763 on Build GRCh38
rs956274796 Chr.2: 144007774-144007774 on Build GRCh38
rs1022323499 Chr.2: 144007806-144007806 on Build GRCh38
rs1049909582 Chr.2: 144007517-144007517 on Build GRCh38
rs748328702 Chr.2: 144007474-144007474 on Build GRCh38
rs934986241 Chr.2: 144007798-144007798 on Build GRCh38
rs749442089 Chr.2: 144007718-144007718 on Build GRCh38
rs897444525 Chr.2: 144007593-144007593 on Build GRCh38
rs1039563102 Chr.2: 144007880-144007880 on Build GRCh38
rs144971379 Chr.2: 144007572-144007572 on Build GRCh38
rs184270940 Chr.2: 144007785-144007785 on Build GRCh38
rs1049482581 Chr.2: 144007634-144007634 on Build GRCh38
rs879568891 Chr.2: 144007895-144007895 on Build GRCh38
rs776674670 Chr.2: 144007581-144007581 on Build GRCh38
rs761797960 Chr.2: 144007587-144007587 on Build GRCh38
rs201214960 Chr.2: 144007470-144007470 on Build GRCh38
rs753469659 Chr.2: 144007475-144007475 on Build GRCh38
rs768452730 Chr.2: 144007917-144007917 on Build GRCh38
rs1002880092 Chr.2: 144007755-144007755 on Build GRCh38
rs772508349 Chr.2: 144007571-144007572 on Build GRCh38
rs756434344 Chr.2: 144007816-144007826 on Build GRCh38
rs768036580 Chr.2: 144007582-144007583 on Build GRCh38
rs1003381859 Chr.2: 144007594-144007594 on Build GRCh38
rs760678644 Chr.2: 144007472-144007472 on Build GRCh38
rs1035806554 Chr.2: 144007624-144007624 on Build GRCh38
rs760080365 Chr.2: 144007542-144007543 on Build GRCh38
rs528455231 Chr.2: 144007550-144007550 on Build GRCh38
rs75942008 Chr.2: 144007743-144007743 on Build GRCh38
rs760565143 Chr.2: 144007873-144007873 on Build GRCh38
rs145098096 Chr.2: 144007509-144007509 on Build GRCh38
rs1031676426 Chr.2: 144007756-144007756 on Build GRCh38
rs369224804 Chr.2: 144007492-144007492 on Build GRCh38
rs919003132 Chr.2: 144007869-144007869 on Build GRCh38
rs1046946010 Chr.2: 144007912-144007912 on Build GRCh38

View Full Product Details