Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 148178574-148179048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBD5
Entrez Gene ID
55777
Gene Name
methyl-CpG binding domain protein 5
Gene Aliases
MRD1
UniGene
Hs.458312
Gene Chromosome Location
Chr.2: 148021011-148516923 on Build GRCh38
Transcripts
NM_018328.4 XM_011511470.2 XM_011511472.2

SNP Detail

SNP ID Chromosome Location
rs780725807 Chr.2: 148178741-148178741 on Build GRCh38
rs548248613 Chr.2: 148178628-148178628 on Build GRCh38
rs746648847 Chr.2: 148178658-148178658 on Build GRCh38
rs934051758 Chr.2: 148178998-148178998 on Build GRCh38
rs981418205 Chr.2: 148178624-148178624 on Build GRCh38
rs942825691 Chr.2: 148178953-148178953 on Build GRCh38
rs921028393 Chr.2: 148178800-148178800 on Build GRCh38
rs946567989 Chr.2: 148178766-148178766 on Build GRCh38
rs538459122 Chr.2: 148178901-148178901 on Build GRCh38
rs1047118628 Chr.2: 148178999-148178999 on Build GRCh38
rs979417348 Chr.2: 148178926-148178926 on Build GRCh38
rs12465610 Chr.2: 148178611-148178611 on Build GRCh38
rs1039778344 Chr.2: 148178603-148178603 on Build GRCh38
rs1050752230 Chr.2: 148178853-148178853 on Build GRCh38
rs1016137155 Chr.2: 148178685-148178685 on Build GRCh38
rs1051695387 Chr.2: 148178655-148178655 on Build GRCh38
rs1039053137 Chr.2: 148178897-148178897 on Build GRCh38
rs571924213 Chr.2: 148179011-148179011 on Build GRCh38
rs967697022 Chr.2: 148178917-148178917 on Build GRCh38
rs898985723 Chr.2: 148178727-148178727 on Build GRCh38
rs956623269 Chr.2: 148178630-148178630 on Build GRCh38
rs988499117 Chr.2: 148178649-148178649 on Build GRCh38
rs139919398 Chr.2: 148178864-148178864 on Build GRCh38
rs1034018458 Chr.2: 148178768-148178768 on Build GRCh38
rs768351750 Chr.2: 148178682-148178682 on Build GRCh38
rs796052705 Chr.2: 148178802-148178803 on Build GRCh38
rs754998351 Chr.2: 148178895-148178898 on Build GRCh38
rs751272613 Chr.2: 148178977-148178977 on Build GRCh38
rs933746892 Chr.2: 148178799-148178823 on Build GRCh38
rs975514351 Chr.2: 148178954-148178954 on Build GRCh38
rs1005188022 Chr.2: 148178664-148178664 on Build GRCh38
rs959714540 Chr.2: 148178799-148178799 on Build GRCh38
rs915067401 Chr.2: 148178713-148178713 on Build GRCh38
rs942836150 Chr.2: 148178598-148178598 on Build GRCh38
rs530015911 Chr.2: 148178997-148178997 on Build GRCh38
rs926153763 Chr.2: 148178941-148178941 on Build GRCh38
rs1023493901 Chr.2: 148178604-148178604 on Build GRCh38

View Full Product Details