Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152651510-152652008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF40A
Entrez Gene ID
55660
Gene Name
pre-mRNA processing factor 40 homolog A
Gene Aliases
FBP-11, FBP11, FLAF1, FNBP3, HIP-10, HIP10, HYPA, NY-REN-6, Prp40
UniGene
Hs.643580
Gene Chromosome Location
Chr.2: 152651593-152718007 on Build GRCh38
Transcripts
NM_017892.3

SNP Detail

SNP ID Chromosome Location
rs961360946 Chr.2: 152651552-152651552 on Build GRCh38
rs923876041 Chr.2: 152651982-152651982 on Build GRCh38
rs1030099223 Chr.2: 152651928-152651928 on Build GRCh38
rs1022407250 Chr.2: 152651962-152651962 on Build GRCh38
rs1052367950 Chr.2: 152651871-152651871 on Build GRCh38
rs538190744 Chr.2: 152651799-152651799 on Build GRCh38
rs555221652 Chr.2: 152651810-152651810 on Build GRCh38
rs998194329 Chr.2: 152651921-152651921 on Build GRCh38
rs897434673 Chr.2: 152651911-152651911 on Build GRCh38
rs778005214 Chr.2: 152651571-152651571 on Build GRCh38
rs186614515 Chr.2: 152651970-152651970 on Build GRCh38
rs927893275 Chr.2: 152651540-152651543 on Build GRCh38
rs927642401 Chr.2: 152651858-152651858 on Build GRCh38
rs574980055 Chr.2: 152651828-152651828 on Build GRCh38
rs915107432 Chr.2: 152651632-152651632 on Build GRCh38
rs554311919 Chr.2: 152651943-152651943 on Build GRCh38
rs973539006 Chr.2: 152651754-152651754 on Build GRCh38
rs57704403 Chr.2: 152651570-152651571 on Build GRCh38
rs11685204 Chr.2: 152651574-152651574 on Build GRCh38
rs1005675804 Chr.2: 152651879-152651879 on Build GRCh38
rs904639863 Chr.2: 152651941-152651941 on Build GRCh38
rs868495801 Chr.2: 152651902-152651902 on Build GRCh38
rs200101751 Chr.2: 152651569-152651570 on Build GRCh38
rs1006952619 Chr.2: 152651949-152651949 on Build GRCh38
rs946546247 Chr.2: 152651636-152651636 on Build GRCh38
rs888588740 Chr.2: 152651861-152651861 on Build GRCh38
rs72511561 Chr.2: 152651574-152651574 on Build GRCh38
rs561530222 Chr.2: 152651845-152651845 on Build GRCh38
rs939306033 Chr.2: 152651868-152651871 on Build GRCh38
rs905110696 Chr.2: 152651594-152651594 on Build GRCh38
rs397948085 Chr.2: 152651573-152651574 on Build GRCh38
rs962211509 Chr.2: 152651743-152651743 on Build GRCh38
rs977166333 Chr.2: 152651553-152651553 on Build GRCh38
rs540781994 Chr.2: 152651937-152651937 on Build GRCh38
rs183437024 Chr.2: 152651631-152651631 on Build GRCh38
rs865788089 Chr.2: 152651974-152651974 on Build GRCh38
rs1015551482 Chr.2: 152651723-152651723 on Build GRCh38
rs946568278 Chr.2: 152651916-152651916 on Build GRCh38
rs938998737 Chr.2: 152651701-152651701 on Build GRCh38
rs1043133374 Chr.2: 152651596-152651596 on Build GRCh38
rs772819658 Chr.2: 152651725-152651725 on Build GRCh38
rs771022007 Chr.2: 152651571-152651572 on Build GRCh38
rs760094956 Chr.2: 152651908-152651908 on Build GRCh38
rs892298330 Chr.2: 152651576-152651576 on Build GRCh38
rs904587497 Chr.2: 152651616-152651616 on Build GRCh38
rs769030823 Chr.2: 152651590-152651590 on Build GRCh38
rs190701331 Chr.2: 152651557-152651557 on Build GRCh38
rs955160268 Chr.2: 152651665-152651665 on Build GRCh38
rs977979700 Chr.2: 152651654-152651654 on Build GRCh38
rs377475461 Chr.2: 152651626-152651626 on Build GRCh38
rs954140641 Chr.2: 152651938-152651938 on Build GRCh38
rs1050892673 Chr.2: 152651961-152651961 on Build GRCh38
rs947820797 Chr.2: 152651842-152651842 on Build GRCh38
rs1009148512 Chr.2: 152651714-152651714 on Build GRCh38
rs923857294 Chr.2: 152651662-152651662 on Build GRCh38
rs996152937 Chr.2: 152651623-152651623 on Build GRCh38
rs1056569953 Chr.2: 152651802-152651802 on Build GRCh38
rs866446164 Chr.2: 152651575-152651575 on Build GRCh38
rs913776134 Chr.2: 152651882-152651882 on Build GRCh38

View Full Product Details