Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159941481-159942005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2R1
Entrez Gene ID
22925
Gene Name
phospholipase A2 receptor 1
Gene Aliases
CLEC13C, PLA2-R, PLA2G1R, PLA2IR, PLA2R
UniGene
Hs.410477
Gene Chromosome Location
Chr.2: 159928384-160062902 on Build GRCh38
Transcripts
NM_001195641.1 NM_007366.4 XM_005246392.3 XM_011510820.2 XM_017003598.1 XM_017003600.1 XM_017003602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588932 c.4340T>C p.V1447A Chr.2: 159941830-159941830 on Build GRCh38
6587482 c.4354A>T p.N1452Y Chr.2: 159941816-159941816 on Build GRCh38
399444 c.4205C>T p.A1402V Chr.2: 159941965-159941965 on Build GRCh38
3300501 c.4199C>T p.P1400L Chr.2: 159941971-159941971 on Build GRCh38
3837140 c.4227C>T p.V1409V Chr.2: 159941943-159941943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757850432 Chr.2: 159941972-159941972 on Build GRCh38
rs1017833814 Chr.2: 159941827-159941827 on Build GRCh38
rs760757680 Chr.2: 159941801-159941801 on Build GRCh38
rs759917286 Chr.2: 159941734-159941734 on Build GRCh38
rs1044270161 Chr.2: 159941635-159941635 on Build GRCh38
rs762784868 Chr.2: 159941822-159941822 on Build GRCh38
rs765117802 Chr.2: 159941812-159941812 on Build GRCh38
rs893083780 Chr.2: 159941543-159941543 on Build GRCh38
rs747628224 Chr.2: 159941750-159941750 on Build GRCh38
rs757760363 Chr.2: 159941893-159941893 on Build GRCh38
rs144754264 Chr.2: 159941790-159941790 on Build GRCh38
rs1002627670 Chr.2: 159941740-159941740 on Build GRCh38
rs755614255 Chr.2: 159941867-159941867 on Build GRCh38
rs771393574 Chr.2: 159941795-159941795 on Build GRCh38
rs560415631 Chr.2: 159941821-159941821 on Build GRCh38
rs948622938 Chr.2: 159941596-159941596 on Build GRCh38
rs753328464 Chr.2: 159941969-159941969 on Build GRCh38
rs778466311 Chr.2: 159941875-159941875 on Build GRCh38
rs758511536 Chr.2: 159941564-159941565 on Build GRCh38
rs367882295 Chr.2: 159941965-159941965 on Build GRCh38
rs372712233 Chr.2: 159941966-159941966 on Build GRCh38
rs371803135 Chr.2: 159941942-159941942 on Build GRCh38
rs368059510 Chr.2: 159941908-159941908 on Build GRCh38
rs973694210 Chr.2: 159941511-159941511 on Build GRCh38
rs182334014 Chr.2: 159941866-159941866 on Build GRCh38
rs1001567103 Chr.2: 159941806-159941806 on Build GRCh38
rs747098064 Chr.2: 159941931-159941931 on Build GRCh38
rs932366430 Chr.2: 159941591-159941591 on Build GRCh38
rs995079225 Chr.2: 159941920-159941920 on Build GRCh38
rs556783031 Chr.2: 159941621-159941621 on Build GRCh38
rs780565639 Chr.2: 159941746-159941746 on Build GRCh38
rs912336028 Chr.2: 159941650-159941650 on Build GRCh38
rs947305059 Chr.2: 159941706-159941706 on Build GRCh38
rs774690617 Chr.2: 159941877-159941877 on Build GRCh38
rs772104361 Chr.2: 159941926-159941926 on Build GRCh38
rs913114131 Chr.2: 159941697-159941697 on Build GRCh38
rs943843633 Chr.2: 159941663-159941663 on Build GRCh38
rs767109719 Chr.2: 159941963-159941963 on Build GRCh38
rs774769426 Chr.2: 159941796-159941796 on Build GRCh38
rs756592979 Chr.2: 159941766-159941766 on Build GRCh38
rs780282214 Chr.2: 159941924-159941924 on Build GRCh38
rs560947497 Chr.2: 159941933-159941933 on Build GRCh38
rs746081413 Chr.2: 159941797-159941797 on Build GRCh38
rs754591691 Chr.2: 159941854-159941854 on Build GRCh38
rs896895848 Chr.2: 159941770-159941770 on Build GRCh38
rs374235754 Chr.2: 159941964-159941964 on Build GRCh38
rs545560852 Chr.2: 159941786-159941786 on Build GRCh38
rs766150509 Chr.2: 159941834-159941834 on Build GRCh38
rs774147212 Chr.2: 159941960-159941960 on Build GRCh38
rs990192399 Chr.2: 159941975-159941975 on Build GRCh38
rs773190521 Chr.2: 159941934-159941934 on Build GRCh38
rs578126904 Chr.2: 159941707-159941707 on Build GRCh38
rs151119000 Chr.2: 159941962-159941962 on Build GRCh38
rs373556051 Chr.2: 159941760-159941760 on Build GRCh38
rs746181323 Chr.2: 159941898-159941898 on Build GRCh38
rs1052892900 Chr.2: 159941685-159941685 on Build GRCh38
rs753057429 Chr.2: 159941900-159941907 on Build GRCh38
rs145231379 Chr.2: 159941788-159941788 on Build GRCh38
rs770788723 Chr.2: 159941945-159941945 on Build GRCh38
rs775442028 Chr.2: 159941800-159941800 on Build GRCh38
rs201567578 Chr.2: 159941897-159941897 on Build GRCh38
rs767141649 Chr.2: 159941856-159941856 on Build GRCh38
rs751281506 Chr.2: 159941852-159941852 on Build GRCh38

View Full Product Details