Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162343981-162344476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCA
Entrez Gene ID
25801
Gene Name
grancalcin
Gene Aliases
GCL
UniGene
Hs.377894
Gene Chromosome Location
Chr.2: 162318768-162371605 on Build GRCh38
Transcripts
NM_012198.3 XM_005246446.2 XM_006712398.3 XM_006712400.3 XM_006712401.3 XM_011510926.2 XM_011510927.2 XM_017003764.1 XM_017003765.1 XM_017003766.1 XM_017003767.1 XM_017003768.1 XM_017003769.1

SNP Detail

SNP ID Chromosome Location
rs778016125 Chr.2: 162344198-162344198 on Build GRCh38
rs989225047 Chr.2: 162344250-162344250 on Build GRCh38
rs548034993 Chr.2: 162344409-162344409 on Build GRCh38
rs994204567 Chr.2: 162344193-162344193 on Build GRCh38
rs994238875 Chr.2: 162344032-162344032 on Build GRCh38
rs371103030 Chr.2: 162344275-162344275 on Build GRCh38
rs986114960 Chr.2: 162344066-162344066 on Build GRCh38
rs749047786 Chr.2: 162344230-162344231 on Build GRCh38
rs541163281 Chr.2: 162344383-162344383 on Build GRCh38
rs7567026 Chr.2: 162344168-162344168 on Build GRCh38
rs951157772 Chr.2: 162344391-162344391 on Build GRCh38
rs766108785 Chr.2: 162344301-162344301 on Build GRCh38
rs772671473 Chr.2: 162344246-162344246 on Build GRCh38
rs764228048 Chr.2: 162344306-162344306 on Build GRCh38
rs1048890985 Chr.2: 162344387-162344387 on Build GRCh38
rs981859315 Chr.2: 162344274-162344274 on Build GRCh38
rs1048501665 Chr.2: 162344194-162344194 on Build GRCh38
rs537056010 Chr.2: 162344137-162344137 on Build GRCh38
rs749496273 Chr.2: 162344202-162344202 on Build GRCh38
rs149650493 Chr.2: 162344324-162344324 on Build GRCh38
rs762835855 Chr.2: 162344166-162344166 on Build GRCh38
rs569823913 Chr.2: 162344118-162344118 on Build GRCh38
rs556438498 Chr.2: 162344388-162344388 on Build GRCh38
rs776012181 Chr.2: 162344272-162344272 on Build GRCh38
rs927611334 Chr.2: 162344159-162344159 on Build GRCh38
rs897694811 Chr.2: 162344180-162344180 on Build GRCh38
rs888253399 Chr.2: 162344039-162344039 on Build GRCh38
rs756171068 Chr.2: 162344239-162344239 on Build GRCh38
rs780299879 Chr.2: 162344326-162344326 on Build GRCh38
rs777730174 Chr.2: 162344240-162344240 on Build GRCh38
rs776295741 Chr.2: 162344224-162344224 on Build GRCh38
rs564985243 Chr.2: 162344371-162344371 on Build GRCh38
rs376042341 Chr.2: 162344227-162344227 on Build GRCh38
rs927860780 Chr.2: 162344013-162344013 on Build GRCh38
rs372697444 Chr.2: 162344245-162344245 on Build GRCh38
rs1011122048 Chr.2: 162344374-162344374 on Build GRCh38
rs1007403817 Chr.2: 162344394-162344394 on Build GRCh38
rs762776897 Chr.2: 162344294-162344294 on Build GRCh38
rs559388510 Chr.2: 162344390-162344390 on Build GRCh38
rs377430052 Chr.2: 162344259-162344259 on Build GRCh38
rs1003729671 Chr.2: 162344385-162344385 on Build GRCh38
rs757623211 Chr.2: 162344243-162344243 on Build GRCh38
rs935343674 Chr.2: 162344364-162344364 on Build GRCh38
rs745601861 Chr.2: 162344042-162344042 on Build GRCh38
rs921125106 Chr.2: 162344424-162344424 on Build GRCh38
rs759394293 Chr.2: 162344304-162344304 on Build GRCh38
rs565566323 Chr.2: 162344040-162344040 on Build GRCh38
rs998428641 Chr.2: 162344439-162344439 on Build GRCh38
rs965633677 Chr.2: 162344214-162344214 on Build GRCh38
rs932073416 Chr.2: 162344344-162344344 on Build GRCh38
rs375545511 Chr.2: 162344308-162344308 on Build GRCh38
rs958178713 Chr.2: 162344143-162344143 on Build GRCh38
rs947784581 Chr.2: 162344152-162344152 on Build GRCh38
rs534733381 Chr.2: 162344205-162344205 on Build GRCh38
rs181421232 Chr.2: 162344322-162344322 on Build GRCh38
rs969544407 Chr.2: 162344266-162344266 on Build GRCh38
rs941247402 Chr.2: 162344317-162344317 on Build GRCh38
rs754048627 Chr.2: 162344242-162344242 on Build GRCh38
rs372896502 Chr.2: 162344225-162344225 on Build GRCh38
rs982192081 Chr.2: 162344153-162344153 on Build GRCh38
rs775456001 Chr.2: 162344138-162344138 on Build GRCh38
rs544084551 Chr.2: 162344353-162344353 on Build GRCh38
rs377659327 Chr.2: 162344311-162344311 on Build GRCh38
rs769494743 Chr.2: 162344287-162344287 on Build GRCh38
rs962620863 Chr.2: 162344419-162344419 on Build GRCh38
rs996572738 Chr.2: 162344087-162344087 on Build GRCh38
rs975273562 Chr.2: 162344422-162344422 on Build GRCh38
rs77262079 Chr.2: 162344373-162344373 on Build GRCh38
rs1031296240 Chr.2: 162344453-162344453 on Build GRCh38
rs955131766 Chr.2: 162344115-162344115 on Build GRCh38
rs762863330 Chr.2: 162344234-162344234 on Build GRCh38
rs930440254 Chr.2: 162344386-162344386 on Build GRCh38
rs763539843 Chr.2: 162344222-162344222 on Build GRCh38
rs116266748 Chr.2: 162344045-162344045 on Build GRCh38
rs1036271388 Chr.2: 162344382-162344382 on Build GRCh38
rs377110441 Chr.2: 162344162-162344162 on Build GRCh38
rs772969823 Chr.2: 162344293-162344293 on Build GRCh38
rs890475203 Chr.2: 162344357-162344357 on Build GRCh38
rs375649533 Chr.2: 162344151-162344151 on Build GRCh38
rs369137863 Chr.2: 162344235-162344235 on Build GRCh38
rs888532210 Chr.2: 162344197-162344197 on Build GRCh38
rs916330929 Chr.2: 162344149-162344149 on Build GRCh38
rs548481242 Chr.2: 162344104-162344104 on Build GRCh38
rs914901038 Chr.2: 162344147-162344147 on Build GRCh38
rs371966041 Chr.2: 162344099-162344099 on Build GRCh38
rs935049464 Chr.2: 162344123-162344123 on Build GRCh38
rs939272241 Chr.2: 162344376-162344376 on Build GRCh38
rs572691994 Chr.2: 162344263-162344263 on Build GRCh38
rs905348274 Chr.2: 162344378-162344378 on Build GRCh38
rs764942681 Chr.2: 162344226-162344226 on Build GRCh38
rs927876138 Chr.2: 162344165-162344165 on Build GRCh38
rs1051842287 Chr.2: 162344354-162344354 on Build GRCh38
rs1018489890 Chr.2: 162344076-162344076 on Build GRCh38
rs1035128844 Chr.2: 162344276-162344276 on Build GRCh38
rs772767492 Chr.2: 162344207-162344207 on Build GRCh38
rs547482928 Chr.2: 162344037-162344037 on Build GRCh38
rs969558942 Chr.2: 162344333-162344333 on Build GRCh38
rs768158188 Chr.2: 162344270-162344270 on Build GRCh38
rs773790858 Chr.2: 162344206-162344206 on Build GRCh38
rs751455584 Chr.2: 162344303-162344303 on Build GRCh38
rs2241662 Chr.2: 162344402-162344402 on Build GRCh38
rs779314338 Chr.2: 162344313-162344313 on Build GRCh38
rs760267924 Chr.2: 162344221-162344221 on Build GRCh38
rs976917683 Chr.2: 162344116-162344116 on Build GRCh38
rs751482886 Chr.2: 162344236-162344236 on Build GRCh38
rs774844613 Chr.2: 162344210-162344210 on Build GRCh38

View Full Product Details